Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 33007523-33008019 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BPIFB2
Entrez Gene ID
80341
Gene Name
BPI fold containing family B member 2
Gene Aliases
BPIL1, C20orf184, LPLUNC2, RYSR, dJ726C3.2
UniGene
Hs.257045
Gene Chromosome Location
Chr.20: 33007578-33023709 on Build GRCh38
Transcripts
NM_025227.2

SNP Detail

SNP ID Chromosome Location
rs1000445001 Chr.20: 33007662-33007662 on Build GRCh38
rs773394322 Chr.20: 33007845-33007845 on Build GRCh38
rs866852811 Chr.20: 33007824-33007824 on Build GRCh38
rs949865353 Chr.20: 33007847-33007847 on Build GRCh38
rs577747324 Chr.20: 33007898-33007898 on Build GRCh38
rs115165664 Chr.20: 33007773-33007773 on Build GRCh38
rs562532108 Chr.20: 33007858-33007858 on Build GRCh38
rs1055260708 Chr.20: 33007579-33007579 on Build GRCh38
rs966436143 Chr.20: 33007935-33007935 on Build GRCh38
rs961412315 Chr.20: 33007879-33007879 on Build GRCh38
rs1043941352 Chr.20: 33007694-33007694 on Build GRCh38
rs746433589 Chr.20: 33007922-33007922 on Build GRCh38
rs568359282 Chr.20: 33007684-33007684 on Build GRCh38
rs6579061 Chr.20: 33007703-33007703 on Build GRCh38
rs763322596 Chr.20: 33007768-33007768 on Build GRCh38
rs754762708 Chr.20: 33007648-33007648 on Build GRCh38
rs1054526168 Chr.20: 33007900-33007900 on Build GRCh38
rs573754878 Chr.20: 33007801-33007801 on Build GRCh38
rs780692451 Chr.20: 33007944-33007944 on Build GRCh38
rs1043969917 Chr.20: 33007971-33007971 on Build GRCh38
rs1037076226 Chr.20: 33007541-33007541 on Build GRCh38
rs965463599 Chr.20: 33007787-33007787 on Build GRCh38
rs557816267 Chr.20: 33007700-33007700 on Build GRCh38
rs764560752 Chr.20: 33007896-33007896 on Build GRCh38
rs1033207722 Chr.20: 33007670-33007670 on Build GRCh38
rs921136388 Chr.20: 33007706-33007706 on Build GRCh38
rs975222975 Chr.20: 33007704-33007704 on Build GRCh38
rs745558329 Chr.20: 33007947-33007947 on Build GRCh38
rs1044123724 Chr.20: 33007878-33007878 on Build GRCh38
rs984058717 Chr.20: 33007857-33007857 on Build GRCh38
rs188008777 Chr.20: 33007659-33007659 on Build GRCh38
rs560135566 Chr.20: 33007933-33007933 on Build GRCh38
rs143293880 Chr.20: 33007668-33007668 on Build GRCh38
rs770265093 Chr.20: 33007966-33007966 on Build GRCh38
rs750702696 Chr.20: 33007554-33007554 on Build GRCh38
rs770485446 Chr.20: 33007939-33007939 on Build GRCh38
rs368345368 Chr.20: 33007629-33007629 on Build GRCh38
rs1044578373 Chr.20: 33007643-33007643 on Build GRCh38
rs906066333 Chr.20: 33007891-33007891 on Build GRCh38
rs956071790 Chr.20: 33007677-33007677 on Build GRCh38
rs73907342 Chr.20: 33007555-33007555 on Build GRCh38
rs142272365 Chr.20: 33007818-33007819 on Build GRCh38
rs1036702969 Chr.20: 33007829-33007829 on Build GRCh38
rs973108566 Chr.20: 33007883-33007883 on Build GRCh38
rs534098211 Chr.20: 33007770-33007770 on Build GRCh38
rs879922610 Chr.20: 33007869-33007869 on Build GRCh38
rs73907343 Chr.20: 33007901-33007901 on Build GRCh38
rs191375079 Chr.20: 33007817-33007817 on Build GRCh38
rs953840259 Chr.20: 33007711-33007711 on Build GRCh38
rs1014382484 Chr.20: 33007630-33007630 on Build GRCh38
rs148335566 Chr.20: 33007685-33007685 on Build GRCh38

View Full Product Details