Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 33012712-33013210 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BPIFB2
Entrez Gene ID
80341
Gene Name
BPI fold containing family B member 2
Gene Aliases
BPIL1, C20orf184, LPLUNC2, RYSR, dJ726C3.2
UniGene
Hs.257045
Gene Chromosome Location
Chr.20: 33007578-33023709 on Build GRCh38
Transcripts
NM_025227.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4136825 c.208C>T p.R70W Chr.20: 33012807-33012807 on Build GRCh38
723089 c.211A>T p.I71F Chr.20: 33012810-33012810 on Build GRCh38
1025842 c.204-1_204insGATCC p.I71fs*4 Chr.20: 33012802-33012803 on Build GRCh38
6612337 c.308G>A p.R103H Chr.20: 33012907-33012907 on Build GRCh38
3545343 c.263G>A p.G88E Chr.20: 33012862-33012862 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778362267 Chr.20: 33012773-33012773 on Build GRCh38
rs778593567 Chr.20: 33012861-33012861 on Build GRCh38
rs780716507 Chr.20: 33012950-33012950 on Build GRCh38
rs560890032 Chr.20: 33013061-33013061 on Build GRCh38
rs774436795 Chr.20: 33012899-33012899 on Build GRCh38
rs781689215 Chr.20: 33012868-33012868 on Build GRCh38
rs777115352 Chr.20: 33012937-33012937 on Build GRCh38
rs573050966 Chr.20: 33012872-33012872 on Build GRCh38
rs183580046 Chr.20: 33012957-33012957 on Build GRCh38
rs574105155 Chr.20: 33012941-33012941 on Build GRCh38
rs760500106 Chr.20: 33012914-33012914 on Build GRCh38
rs1037251893 Chr.20: 33013135-33013135 on Build GRCh38
rs888170615 Chr.20: 33012928-33012928 on Build GRCh38
rs752313736 Chr.20: 33012931-33012931 on Build GRCh38
rs146634710 Chr.20: 33013050-33013050 on Build GRCh38
rs747678701 Chr.20: 33012784-33012784 on Build GRCh38
rs376681063 Chr.20: 33012757-33012757 on Build GRCh38
rs754619805 Chr.20: 33012860-33012860 on Build GRCh38
rs368011617 Chr.20: 33012766-33012766 on Build GRCh38
rs943910365 Chr.20: 33012840-33012840 on Build GRCh38
rs777009173 Chr.20: 33012794-33012794 on Build GRCh38
rs965375713 Chr.20: 33013006-33013006 on Build GRCh38
rs149802409 Chr.20: 33012818-33012818 on Build GRCh38
rs757877643 Chr.20: 33012867-33012867 on Build GRCh38
rs976664737 Chr.20: 33013115-33013115 on Build GRCh38
rs751145068 Chr.20: 33012940-33012940 on Build GRCh38
rs766371880 Chr.20: 33012925-33012925 on Build GRCh38
rs540459199 Chr.20: 33012896-33012896 on Build GRCh38
rs2024935 Chr.20: 33013185-33013185 on Build GRCh38
rs144442462 Chr.20: 33012955-33012955 on Build GRCh38
rs780565069 Chr.20: 33012877-33012877 on Build GRCh38
rs372195566 Chr.20: 33012796-33012796 on Build GRCh38
rs1012661375 Chr.20: 33012965-33012965 on Build GRCh38
rs776047785 Chr.20: 33012808-33012808 on Build GRCh38
rs761917219 Chr.20: 33012906-33012906 on Build GRCh38
rs984200898 Chr.20: 33013096-33013096 on Build GRCh38
rs764829953 Chr.20: 33012930-33012930 on Build GRCh38
rs1000318524 Chr.20: 33013012-33013012 on Build GRCh38
rs976790522 Chr.20: 33013020-33013021 on Build GRCh38
rs932752194 Chr.20: 33012839-33012839 on Build GRCh38
rs924625788 Chr.20: 33012768-33012768 on Build GRCh38
rs906754576 Chr.20: 33013140-33013140 on Build GRCh38
rs905855735 Chr.20: 33012979-33012979 on Build GRCh38
rs968558907 Chr.20: 33012977-33012977 on Build GRCh38
rs754672743 Chr.20: 33012771-33012771 on Build GRCh38
rs1031559844 Chr.20: 33013002-33013002 on Build GRCh38
rs199826775 Chr.20: 33012901-33012901 on Build GRCh38
rs750355484 Chr.20: 33012832-33012832 on Build GRCh38
rs762463179 Chr.20: 33012863-33012863 on Build GRCh38
rs1022943539 Chr.20: 33012965-33012965 on Build GRCh38
rs774563198 Chr.20: 33012822-33012822 on Build GRCh38
rs987989749 Chr.20: 33013051-33013051 on Build GRCh38
rs766138187 Chr.20: 33012852-33012852 on Build GRCh38
rs770078142 Chr.20: 33012807-33012807 on Build GRCh38
rs749731259 Chr.20: 33012880-33012880 on Build GRCh38
rs144717306 Chr.20: 33012871-33012871 on Build GRCh38
rs758117205 Chr.20: 33012932-33012932 on Build GRCh38
rs561853298 Chr.20: 33013126-33013126 on Build GRCh38
rs761815952 Chr.20: 33012824-33012824 on Build GRCh38
rs1014447537 Chr.20: 33012956-33012956 on Build GRCh38
rs771456069 Chr.20: 33012792-33012792 on Build GRCh38
rs1021901606 Chr.20: 33012977-33012977 on Build GRCh38
rs375320227 Chr.20: 33012831-33012831 on Build GRCh38
rs78767145 Chr.20: 33013029-33013029 on Build GRCh38
rs747636380 Chr.20: 33012864-33012864 on Build GRCh38
rs987939029 Chr.20: 33012830-33012830 on Build GRCh38
rs369655260 Chr.20: 33012929-33012929 on Build GRCh38
rs148804619 Chr.20: 33013067-33013067 on Build GRCh38
rs549506723 Chr.20: 33013100-33013100 on Build GRCh38
rs979235234 Chr.20: 33012804-33012804 on Build GRCh38
rs561715693 Chr.20: 33012913-33012913 on Build GRCh38
rs539753794 Chr.20: 33012735-33012735 on Build GRCh38
rs755886905 Chr.20: 33012845-33012845 on Build GRCh38
rs932759480 Chr.20: 33013081-33013081 on Build GRCh38
rs753671848 Chr.20: 33012855-33012855 on Build GRCh38

View Full Product Details