Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 33018547-33019049 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BPIFB2
Entrez Gene ID
80341
Gene Name
BPI fold containing family B member 2
Gene Aliases
BPIL1, C20orf184, LPLUNC2, RYSR, dJ726C3.2
UniGene
Hs.257045
Gene Chromosome Location
Chr.20: 33007578-33023709 on Build GRCh38
Transcripts
NM_025227.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6612340 c.823G>A p.G275S Chr.20: 33018790-33018790 on Build GRCh38
4665968 c.787C>T p.L263L Chr.20: 33018754-33018754 on Build GRCh38
5691413 c.707C>A p.P236H Chr.20: 33018674-33018674 on Build GRCh38
4804395 c.851A>G p.Q284R Chr.20: 33018818-33018818 on Build GRCh38
3911109 c.828C>T p.A276A Chr.20: 33018795-33018795 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1044491421 Chr.20: 33019010-33019010 on Build GRCh38
rs765978934 Chr.20: 33018639-33018639 on Build GRCh38
rs750940543 Chr.20: 33018733-33018733 on Build GRCh38
rs778295974 Chr.20: 33018587-33018587 on Build GRCh38
rs770303903 Chr.20: 33018677-33018677 on Build GRCh38
rs371034992 Chr.20: 33018624-33018624 on Build GRCh38
rs554959164 Chr.20: 33018993-33018993 on Build GRCh38
rs201596840 Chr.20: 33018841-33018841 on Build GRCh38
rs367964762 Chr.20: 33018779-33018779 on Build GRCh38
rs370560791 Chr.20: 33018852-33018852 on Build GRCh38
rs781218958 Chr.20: 33019020-33019020 on Build GRCh38
rs920978876 Chr.20: 33018913-33018913 on Build GRCh38
rs757811849 Chr.20: 33018935-33018935 on Build GRCh38
rs1028274431 Chr.20: 33018915-33018915 on Build GRCh38
rs533775789 Chr.20: 33018767-33018767 on Build GRCh38
rs148983624 Chr.20: 33018664-33018664 on Build GRCh38
rs375953081 Chr.20: 33018696-33018696 on Build GRCh38
rs913653109 Chr.20: 33018630-33018630 on Build GRCh38
rs752320176 Chr.20: 33018651-33018651 on Build GRCh38
rs770072964 Chr.20: 33018599-33018599 on Build GRCh38
rs750937264 Chr.20: 33018771-33018771 on Build GRCh38
rs775230446 Chr.20: 33018817-33018817 on Build GRCh38
rs746193081 Chr.20: 33018667-33018667 on Build GRCh38
rs971788086 Chr.20: 33018627-33018627 on Build GRCh38
rs748380610 Chr.20: 33018848-33018848 on Build GRCh38
rs757957519 Chr.20: 33018726-33018726 on Build GRCh38
rs773330016 Chr.20: 33018771-33018771 on Build GRCh38
rs144077950 Chr.20: 33018777-33018777 on Build GRCh38
rs1024793127 Chr.20: 33018570-33018570 on Build GRCh38
rs140858734 Chr.20: 33018663-33018663 on Build GRCh38
rs374022265 Chr.20: 33018833-33018833 on Build GRCh38
rs545689792 Chr.20: 33019014-33019014 on Build GRCh38
rs901175067 Chr.20: 33018910-33018910 on Build GRCh38
rs756512856 Chr.20: 33018835-33018848 on Build GRCh38
rs112294188 Chr.20: 33018812-33018812 on Build GRCh38
rs374100519 Chr.20: 33018601-33018601 on Build GRCh38
rs147086111 Chr.20: 33018678-33018678 on Build GRCh38
rs527834911 Chr.20: 33018586-33018586 on Build GRCh38
rs759107557 Chr.20: 33018721-33018721 on Build GRCh38
rs561806825 Chr.20: 33018596-33018596 on Build GRCh38
rs764753701 Chr.20: 33018789-33018789 on Build GRCh38
rs3746389 Chr.20: 33018933-33018933 on Build GRCh38
rs762561705 Chr.20: 33018823-33018823 on Build GRCh38
rs370240092 Chr.20: 33018717-33018717 on Build GRCh38
rs755101496 Chr.20: 33018754-33018754 on Build GRCh38
rs918899281 Chr.20: 33018654-33018655 on Build GRCh38
rs573822418 Chr.20: 33018785-33018785 on Build GRCh38
rs778256470 Chr.20: 33018643-33018643 on Build GRCh38
rs1004182689 Chr.20: 33018727-33018727 on Build GRCh38
rs761648157 Chr.20: 33018625-33018625 on Build GRCh38
rs992490525 Chr.20: 33018846-33018846 on Build GRCh38
rs748276111 Chr.20: 33018705-33018705 on Build GRCh38
rs750165753 Chr.20: 33018629-33018629 on Build GRCh38
rs537922118 Chr.20: 33018921-33018921 on Build GRCh38
rs373877757 Chr.20: 33018864-33018864 on Build GRCh38
rs894322981 Chr.20: 33018675-33018675 on Build GRCh38
rs138569633 Chr.20: 33018736-33018736 on Build GRCh38
rs1028323471 Chr.20: 33018746-33018746 on Build GRCh38
rs749717553 Chr.20: 33018740-33018740 on Build GRCh38
rs777391923 Chr.20: 33018964-33018964 on Build GRCh38
rs17853291 Chr.20: 33018737-33018737 on Build GRCh38
rs77962531 Chr.20: 33019001-33019001 on Build GRCh38
rs763662964 Chr.20: 33018832-33018832 on Build GRCh38
rs976520301 Chr.20: 33018898-33018898 on Build GRCh38
rs778957688 Chr.20: 33018847-33018847 on Build GRCh38
rs1053409811 Chr.20: 33018909-33018909 on Build GRCh38
rs1025264790 Chr.20: 33018744-33018744 on Build GRCh38
rs760469870 Chr.20: 33018633-33018633 on Build GRCh38
rs373815604 Chr.20: 33018735-33018735 on Build GRCh38
rs747151901 Chr.20: 33018867-33018867 on Build GRCh38
rs536970824 Chr.20: 33018922-33018922 on Build GRCh38
rs753512920 Chr.20: 33018641-33018641 on Build GRCh38
rs150989727 Chr.20: 33018611-33018611 on Build GRCh38
rs768088122 Chr.20: 33018730-33018730 on Build GRCh38
rs773168283 Chr.20: 33018711-33018711 on Build GRCh38
rs528983082 Chr.20: 33018597-33018597 on Build GRCh38
rs373846957 Chr.20: 33018992-33018992 on Build GRCh38
rs115981583 Chr.20: 33018981-33018981 on Build GRCh38
rs767552641 Chr.20: 33018628-33018628 on Build GRCh38
rs965540841 Chr.20: 33018875-33018875 on Build GRCh38
rs973371797 Chr.20: 33018790-33018790 on Build GRCh38
rs758660625 Chr.20: 33018849-33018849 on Build GRCh38
rs1017076219 Chr.20: 33018934-33018934 on Build GRCh38
rs752743349 Chr.20: 33018900-33018900 on Build GRCh38
rs752111967 Chr.20: 33019026-33019026 on Build GRCh38
rs377669667 Chr.20: 33018618-33018618 on Build GRCh38
rs764954423 Chr.20: 33018722-33018722 on Build GRCh38
rs555124864 Chr.20: 33018768-33018768 on Build GRCh38
rs146974702 Chr.20: 33018593-33018593 on Build GRCh38
rs568725348 Chr.20: 33018612-33018612 on Build GRCh38
rs779202645 Chr.20: 33018764-33018764 on Build GRCh38
rs776452780 Chr.20: 33018718-33018718 on Build GRCh38
rs755403380 Chr.20: 33018843-33018843 on Build GRCh38
rs767004197 Chr.20: 33018836-33018836 on Build GRCh38
rs542994217 Chr.20: 33018913-33018913 on Build GRCh38
rs371717562 Chr.20: 33018787-33018787 on Build GRCh38
rs183328639 Chr.20: 33018704-33018704 on Build GRCh38
rs771935340 Chr.20: 33018710-33018710 on Build GRCh38
rs940269196 Chr.20: 33018965-33018965 on Build GRCh38
rs560749027 Chr.20: 33018572-33018572 on Build GRCh38
rs1035643539 Chr.20: 33018980-33018980 on Build GRCh38
rs999160343 Chr.20: 33018707-33018707 on Build GRCh38

View Full Product Details