Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 33224907-33225327 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BPIFA3
Entrez Gene ID
128861
Gene Name
BPI fold containing family A member 3
Gene Aliases
C20orf71, SPLUNC3
UniGene
Hs.360989
Gene Chromosome Location
Chr.20: 33217310-33227806 on Build GRCh38
Transcripts
NM_001042439.2 NM_178466.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
337683 c.412A>T p.M138L Chr.20: 33225123-33225123 on Build GRCh38
6286440 c.407T>A p.V136E Chr.20: 33225118-33225118 on Build GRCh38
334324 c.446G>T p.W149L Chr.20: 33225157-33225157 on Build GRCh38
1198607 c.443T>C p.F148S Chr.20: 33225154-33225154 on Build GRCh38
7114499 c.483G>A p.V161V Chr.20: 33225194-33225194 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748556803 Chr.20: 33225076-33225076 on Build GRCh38
rs780582409 Chr.20: 33225070-33225070 on Build GRCh38
rs978746404 Chr.20: 33225245-33225245 on Build GRCh38
rs146096538 Chr.20: 33225153-33225153 on Build GRCh38
rs182863021 Chr.20: 33225004-33225004 on Build GRCh38
rs6057770 Chr.20: 33225019-33225019 on Build GRCh38
rs201399371 Chr.20: 33225144-33225144 on Build GRCh38
rs778554421 Chr.20: 33225268-33225268 on Build GRCh38
rs73904639 Chr.20: 33225256-33225256 on Build GRCh38
rs1007198822 Chr.20: 33225136-33225136 on Build GRCh38
rs779441425 Chr.20: 33225164-33225164 on Build GRCh38
rs751163653 Chr.20: 33225132-33225132 on Build GRCh38
rs770024851 Chr.20: 33225219-33225219 on Build GRCh38
rs762995342 Chr.20: 33225221-33225221 on Build GRCh38
rs926199855 Chr.20: 33225091-33225091 on Build GRCh38
rs546813990 Chr.20: 33225043-33225043 on Build GRCh38
rs769222408 Chr.20: 33225074-33225074 on Build GRCh38
rs564064028 Chr.20: 33225263-33225263 on Build GRCh38
rs758639274 Chr.20: 33224988-33224988 on Build GRCh38
rs150349800 Chr.20: 33225105-33225105 on Build GRCh38
rs763817105 Chr.20: 33225128-33225128 on Build GRCh38
rs138661121 Chr.20: 33225180-33225180 on Build GRCh38
rs767121694 Chr.20: 33225231-33225231 on Build GRCh38
rs568158557 Chr.20: 33225044-33225044 on Build GRCh38
rs528433568 Chr.20: 33225296-33225296 on Build GRCh38
rs1032773768 Chr.20: 33225302-33225302 on Build GRCh38
rs757480691 Chr.20: 33225268-33225268 on Build GRCh38
rs754522337 Chr.20: 33225184-33225184 on Build GRCh38
rs116819024 Chr.20: 33225206-33225206 on Build GRCh38
rs1005287771 Chr.20: 33224998-33224998 on Build GRCh38
rs370889922 Chr.20: 33225248-33225248 on Build GRCh38
rs563386656 Chr.20: 33225223-33225223 on Build GRCh38
rs147632261 Chr.20: 33225143-33225143 on Build GRCh38
rs755585157 Chr.20: 33225237-33225237 on Build GRCh38
rs200917820 Chr.20: 33225192-33225192 on Build GRCh38
rs1016361913 Chr.20: 33225137-33225137 on Build GRCh38
rs761510570 Chr.20: 33225230-33225230 on Build GRCh38
rs992729143 Chr.20: 33225018-33225018 on Build GRCh38
rs759739463 Chr.20: 33225210-33225210 on Build GRCh38
rs747189892 Chr.20: 33225198-33225198 on Build GRCh38
rs199894237 Chr.20: 33225240-33225240 on Build GRCh38
rs148428328 Chr.20: 33225229-33225229 on Build GRCh38
rs769935179 Chr.20: 33225289-33225289 on Build GRCh38
rs978593663 Chr.20: 33225032-33225032 on Build GRCh38
rs944390402 Chr.20: 33225107-33225107 on Build GRCh38
rs113913836 Chr.20: 33225249-33225249 on Build GRCh38
rs562352195 Chr.20: 33225181-33225181 on Build GRCh38
rs756862181 Chr.20: 33225133-33225133 on Build GRCh38
rs534574158 Chr.20: 33225026-33225026 on Build GRCh38
rs187062772 Chr.20: 33225112-33225112 on Build GRCh38
rs1004933425 Chr.20: 33225182-33225182 on Build GRCh38
rs762796606 Chr.20: 33225126-33225126 on Build GRCh38
rs749730720 Chr.20: 33225073-33225073 on Build GRCh38
rs6057771 Chr.20: 33225094-33225094 on Build GRCh38
rs142257117 Chr.20: 33225207-33225207 on Build GRCh38
rs35698429 Chr.20: 33225168-33225169 on Build GRCh38
rs6059168 Chr.20: 33225003-33225003 on Build GRCh38
rs746114654 Chr.20: 33225281-33225281 on Build GRCh38
rs376235721 Chr.20: 33225114-33225114 on Build GRCh38
rs773447902 Chr.20: 33225098-33225098 on Build GRCh38
rs539560503 Chr.20: 33225233-33225233 on Build GRCh38
rs3818222 Chr.20: 33225117-33225117 on Build GRCh38
rs758836054 Chr.20: 33225255-33225255 on Build GRCh38
rs780746298 Chr.20: 33225138-33225138 on Build GRCh38
rs535975604 Chr.20: 33225045-33225045 on Build GRCh38
rs140343618 Chr.20: 33225104-33225104 on Build GRCh38
rs761298488 Chr.20: 33225239-33225239 on Build GRCh38
rs747271153 Chr.20: 33225029-33225029 on Build GRCh38
rs1043461748 Chr.20: 33225083-33225083 on Build GRCh38
rs749899737 Chr.20: 33225235-33225235 on Build GRCh38
rs748689462 Chr.20: 33225170-33225170 on Build GRCh38
rs756744363 Chr.20: 33225064-33225064 on Build GRCh38
rs753528108 Chr.20: 33225063-33225064 on Build GRCh38
rs950866153 Chr.20: 33224992-33224992 on Build GRCh38
rs775518278 Chr.20: 33225220-33225220 on Build GRCh38
rs753148476 Chr.20: 33225254-33225254 on Build GRCh38
rs781605698 Chr.20: 33225270-33225270 on Build GRCh38

View Full Product Details