Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 34262731-34263174 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AHCY
Entrez Gene ID
191
Gene Name
adenosylhomocysteinase
Gene Aliases
SAHH, adoHcyase
UniGene
Hs.388004
Gene Chromosome Location
Chr.20: 34235012-34311976 on Build GRCh38
Transcripts
XM_011528657.2 NM_001672.2 XM_005260412.3 XM_011528820.2 XM_011528821.1

Gene Symbol
ASIP
Entrez Gene ID
434
Gene Name
agouti signaling protein
Gene Aliases
AGSW, AGTI, AGTIL, ASP, SHEP9
UniGene
Hs.659995
Gene Chromosome Location
Chr.20: 34186493-34269344 on Build GRCh38
Transcripts
XM_011528657.2 NM_001672.2 XM_005260412.3 XM_011528820.2 XM_011528821.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3545523 c.215C>T p.S72F Chr.20: 34262886-34262886 on Build GRCh38
5929999 c.222+6G>A p.? Chr.20: 34262899-34262899 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs986811413 Chr.20: 34263017-34263020 on Build GRCh38
rs549789591 Chr.20: 34262771-34262771 on Build GRCh38
rs780655512 Chr.20: 34262941-34262941 on Build GRCh38
rs754721917 Chr.20: 34262889-34262889 on Build GRCh38
rs753700502 Chr.20: 34262781-34262781 on Build GRCh38
rs752264573 Chr.20: 34262904-34262904 on Build GRCh38
rs950016975 Chr.20: 34262835-34262835 on Build GRCh38
rs375604124 Chr.20: 34263106-34263106 on Build GRCh38
rs771447427 Chr.20: 34262806-34262806 on Build GRCh38
rs770485831 Chr.20: 34262839-34262839 on Build GRCh38
rs990313829 Chr.20: 34262964-34262964 on Build GRCh38
rs749605159 Chr.20: 34262852-34262852 on Build GRCh38
rs149537165 Chr.20: 34262857-34262857 on Build GRCh38
rs565407899 Chr.20: 34262856-34262856 on Build GRCh38
rs77106176 Chr.20: 34263003-34263003 on Build GRCh38
rs369634657 Chr.20: 34262783-34262783 on Build GRCh38
rs912527886 Chr.20: 34263032-34263032 on Build GRCh38
rs765051639 Chr.20: 34262893-34262893 on Build GRCh38
rs112581222 Chr.20: 34262826-34262826 on Build GRCh38
rs554847477 Chr.20: 34262883-34262883 on Build GRCh38
rs1048863098 Chr.20: 34263141-34263141 on Build GRCh38
rs150059097 Chr.20: 34262924-34262924 on Build GRCh38
rs561768748 Chr.20: 34263102-34263102 on Build GRCh38
rs774388882 Chr.20: 34262946-34262949 on Build GRCh38
rs777139843 Chr.20: 34262915-34262915 on Build GRCh38
rs138013399 Chr.20: 34262833-34262833 on Build GRCh38
rs776143395 Chr.20: 34262840-34262840 on Build GRCh38
rs1040102459 Chr.20: 34262972-34262972 on Build GRCh38
rs1006496502 Chr.20: 34263111-34263111 on Build GRCh38
rs552958158 Chr.20: 34263103-34263103 on Build GRCh38
rs1014754780 Chr.20: 34263120-34263120 on Build GRCh38
rs543671099 Chr.20: 34262940-34262940 on Build GRCh38
rs761242573 Chr.20: 34262796-34262796 on Build GRCh38
rs537339407 Chr.20: 34262989-34262989 on Build GRCh38
rs181105100 Chr.20: 34262855-34262855 on Build GRCh38
rs748676213 Chr.20: 34262794-34262794 on Build GRCh38
rs901510056 Chr.20: 34262986-34262986 on Build GRCh38
rs972744141 Chr.20: 34262776-34262776 on Build GRCh38
rs778481016 Chr.20: 34262791-34262791 on Build GRCh38
rs760602587 Chr.20: 34262870-34262870 on Build GRCh38
rs746458725 Chr.20: 34262923-34262923 on Build GRCh38
rs201591152 Chr.20: 34262810-34262810 on Build GRCh38
rs773399233 Chr.20: 34262858-34262858 on Build GRCh38
rs766239012 Chr.20: 34262871-34262871 on Build GRCh38
rs909980932 Chr.20: 34262761-34262761 on Build GRCh38
rs1129414 Chr.20: 34262853-34262853 on Build GRCh38
rs759028141 Chr.20: 34262802-34262802 on Build GRCh38

View Full Product Details