Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 36366906-36367400 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DLGAP4
Entrez Gene ID
22839
Gene Name
DLG associated protein 4
Gene Aliases
DAP-4, DAP4, DLP4, SAPAP-4, SAPAP4
UniGene
Hs.249600
Gene Chromosome Location
Chr.20: 36306323-36528637 on Build GRCh38
Transcripts
NM_014902.5 XM_011528688.2 XM_011528689.2

SNP Detail

SNP ID Chromosome Location
rs760072654 Chr.20: 36367327-36367327 on Build GRCh38
rs147079279 Chr.20: 36367231-36367231 on Build GRCh38
rs889656425 Chr.20: 36367376-36367376 on Build GRCh38
rs553498782 Chr.20: 36367206-36367206 on Build GRCh38
rs1034223006 Chr.20: 36367194-36367194 on Build GRCh38
rs570164457 Chr.20: 36366980-36366980 on Build GRCh38
rs952506370 Chr.20: 36367149-36367149 on Build GRCh38
rs958405796 Chr.20: 36367214-36367214 on Build GRCh38
rs377203541 Chr.20: 36367347-36367347 on Build GRCh38
rs184915971 Chr.20: 36367288-36367288 on Build GRCh38
rs189760990 Chr.20: 36367291-36367291 on Build GRCh38
rs993991741 Chr.20: 36367340-36367340 on Build GRCh38
rs984008839 Chr.20: 36367154-36367154 on Build GRCh38
rs984698148 Chr.20: 36367235-36367235 on Build GRCh38
rs563222168 Chr.20: 36367245-36367245 on Build GRCh38
rs897802009 Chr.20: 36367309-36367309 on Build GRCh38
rs780906011 Chr.20: 36367116-36367116 on Build GRCh38
rs946261724 Chr.20: 36366934-36366934 on Build GRCh38
rs554742880 Chr.20: 36366955-36366955 on Build GRCh38
rs528360096 Chr.20: 36367292-36367292 on Build GRCh38
rs546936045 Chr.20: 36367320-36367320 on Build GRCh38
rs992437667 Chr.20: 36367216-36367216 on Build GRCh38
rs1050704092 Chr.20: 36367275-36367275 on Build GRCh38
rs541337546 Chr.20: 36367131-36367131 on Build GRCh38
rs558578430 Chr.20: 36367111-36367111 on Build GRCh38
rs769679330 Chr.20: 36367175-36367175 on Build GRCh38
rs969950194 Chr.20: 36367260-36367260 on Build GRCh38
rs568369979 Chr.20: 36367322-36367322 on Build GRCh38
rs192596006 Chr.20: 36367261-36367261 on Build GRCh38
rs947198994 Chr.20: 36367313-36367314 on Build GRCh38
rs931498090 Chr.20: 36366975-36366975 on Build GRCh38
rs750984569 Chr.20: 36366948-36366948 on Build GRCh38
rs574957644 Chr.20: 36367224-36367224 on Build GRCh38
rs537040696 Chr.20: 36367092-36367092 on Build GRCh38
rs745683689 Chr.20: 36367150-36367150 on Build GRCh38
rs577078659 Chr.20: 36367115-36367115 on Build GRCh38
rs936121455 Chr.20: 36367268-36367268 on Build GRCh38
rs1040788009 Chr.20: 36367296-36367296 on Build GRCh38
rs1050628361 Chr.20: 36367010-36367010 on Build GRCh38
rs1006842480 Chr.20: 36367195-36367195 on Build GRCh38
rs1019477649 Chr.20: 36367220-36367221 on Build GRCh38
rs28581719 Chr.20: 36366995-36366995 on Build GRCh38
rs904661407 Chr.20: 36366960-36366960 on Build GRCh38
rs948290761 Chr.20: 36367051-36367051 on Build GRCh38
rs952764223 Chr.20: 36367234-36367234 on Build GRCh38
rs889154106 Chr.20: 36367100-36367100 on Build GRCh38
rs917034788 Chr.20: 36367227-36367227 on Build GRCh38
rs751216745 Chr.20: 36367114-36367114 on Build GRCh38
rs985870362 Chr.20: 36366935-36366935 on Build GRCh38
rs1042858208 Chr.20: 36367191-36367191 on Build GRCh38
rs936256789 Chr.20: 36366961-36366961 on Build GRCh38
rs888312502 Chr.20: 36367369-36367369 on Build GRCh38
rs779944957 Chr.20: 36367246-36367246 on Build GRCh38
rs550790310 Chr.20: 36367370-36367370 on Build GRCh38
rs35077303 Chr.20: 36366951-36366952 on Build GRCh38
rs1008899173 Chr.20: 36367110-36367110 on Build GRCh38

View Full Product Details