Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 36439676-36440153 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DLGAP4
Entrez Gene ID
22839
Gene Name
DLG associated protein 4
Gene Aliases
DAP-4, DAP4, DLP4, SAPAP-4, SAPAP4
UniGene
Hs.249600
Gene Chromosome Location
Chr.20: 36306323-36528637 on Build GRCh38
Transcripts
NM_014902.5 XM_011528688.2 XM_011528689.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3423617 c.1306G>T p.D436Y Chr.20: 36439818-36439818 on Build GRCh38
6004576 c.1296C>T p.S432S Chr.20: 36439808-36439808 on Build GRCh38
6159857 c.1344C>G p.S448R Chr.20: 36439856-36439856 on Build GRCh38
6017878 c.1242G>A p.R414R Chr.20: 36439754-36439754 on Build GRCh38
5580838 c.1262C>T p.S421L Chr.20: 36439774-36439774 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753181982 Chr.20: 36439859-36439859 on Build GRCh38
rs988868200 Chr.20: 36439989-36439989 on Build GRCh38
rs575582414 Chr.20: 36439754-36439754 on Build GRCh38
rs568822083 Chr.20: 36439942-36439942 on Build GRCh38
rs767796965 Chr.20: 36440087-36440087 on Build GRCh38
rs979780893 Chr.20: 36439924-36439924 on Build GRCh38
rs371751848 Chr.20: 36440085-36440085 on Build GRCh38
rs1037607180 Chr.20: 36440010-36440010 on Build GRCh38
rs900692350 Chr.20: 36439793-36439793 on Build GRCh38
rs1005996597 Chr.20: 36439998-36439998 on Build GRCh38
rs963679913 Chr.20: 36439703-36439713 on Build GRCh38
rs745919299 Chr.20: 36439745-36439745 on Build GRCh38
rs762839707 Chr.20: 36440089-36440089 on Build GRCh38
rs912922603 Chr.20: 36440011-36440011 on Build GRCh38
rs112991757 Chr.20: 36440060-36440060 on Build GRCh38
rs764736387 Chr.20: 36439862-36439862 on Build GRCh38
rs747984385 Chr.20: 36439914-36439914 on Build GRCh38
rs970062778 Chr.20: 36439731-36439732 on Build GRCh38
rs762983600 Chr.20: 36439841-36439841 on Build GRCh38
rs776383755 Chr.20: 36439735-36439735 on Build GRCh38
rs779529145 Chr.20: 36439803-36439803 on Build GRCh38
rs900639982 Chr.20: 36440059-36440059 on Build GRCh38
rs6019198 Chr.20: 36440068-36440068 on Build GRCh38
rs761749097 Chr.20: 36439738-36439738 on Build GRCh38
rs1015315898 Chr.20: 36440117-36440117 on Build GRCh38
rs938465796 Chr.20: 36439981-36439981 on Build GRCh38
rs995795264 Chr.20: 36440126-36440126 on Build GRCh38
rs1001578632 Chr.20: 36440019-36440019 on Build GRCh38
rs138421401 Chr.20: 36439801-36439801 on Build GRCh38
rs147291353 Chr.20: 36439860-36439860 on Build GRCh38
rs78624845 Chr.20: 36439785-36439785 on Build GRCh38
rs532819572 Chr.20: 36439950-36439950 on Build GRCh38
rs764158047 Chr.20: 36440115-36440115 on Build GRCh38
rs778564325 Chr.20: 36439783-36439783 on Build GRCh38
rs190899702 Chr.20: 36439717-36439717 on Build GRCh38
rs74418648 Chr.20: 36439883-36439883 on Build GRCh38
rs1005290884 Chr.20: 36439810-36439810 on Build GRCh38
rs767819602 Chr.20: 36440014-36440014 on Build GRCh38
rs766512258 Chr.20: 36439842-36439842 on Build GRCh38
rs775331263 Chr.20: 36439714-36439714 on Build GRCh38
rs769689721 Chr.20: 36439919-36439919 on Build GRCh38
rs6066743 Chr.20: 36440116-36440116 on Build GRCh38
rs764357757 Chr.20: 36440077-36440077 on Build GRCh38
rs116155884 Chr.20: 36440013-36440013 on Build GRCh38
rs546453261 Chr.20: 36439782-36439782 on Build GRCh38
rs375444655 Chr.20: 36439731-36439731 on Build GRCh38
rs781640112 Chr.20: 36439765-36439765 on Build GRCh38
rs767710733 Chr.20: 36439848-36439848 on Build GRCh38
rs550445033 Chr.20: 36439938-36439938 on Build GRCh38
rs1043330223 Chr.20: 36440018-36440018 on Build GRCh38
rs774387764 Chr.20: 36439845-36439845 on Build GRCh38
rs750566233 Chr.20: 36439743-36439743 on Build GRCh38
rs928384121 Chr.20: 36439934-36439934 on Build GRCh38
rs776273851 Chr.20: 36439716-36439716 on Build GRCh38
rs145213052 Chr.20: 36439871-36439871 on Build GRCh38
rs11477851 Chr.20: 36440084-36440084 on Build GRCh38
rs899177502 Chr.20: 36440123-36440123 on Build GRCh38
rs748072313 Chr.20: 36439817-36439817 on Build GRCh38
rs775476284 Chr.20: 36439961-36439961 on Build GRCh38
rs1010715643 Chr.20: 36440069-36440069 on Build GRCh38
rs760670423 Chr.20: 36439759-36439759 on Build GRCh38
rs745417740 Chr.20: 36439795-36439795 on Build GRCh38
rs978922120 Chr.20: 36440086-36440086 on Build GRCh38
rs527846901 Chr.20: 36439835-36439835 on Build GRCh38
rs1036067537 Chr.20: 36440059-36440060 on Build GRCh38
rs754725636 Chr.20: 36439901-36439901 on Build GRCh38
rs538760499 Chr.20: 36439737-36439737 on Build GRCh38
rs145596733 Chr.20: 36439805-36439805 on Build GRCh38
rs868530871 Chr.20: 36439774-36439774 on Build GRCh38
rs769779205 Chr.20: 36439826-36439826 on Build GRCh38
rs766296601 Chr.20: 36439897-36439898 on Build GRCh38
rs368835058 Chr.20: 36439764-36439764 on Build GRCh38
rs765231597 Chr.20: 36439742-36439742 on Build GRCh38
rs199556260 Chr.20: 36439830-36439830 on Build GRCh38
rs566742165 Chr.20: 36440044-36440044 on Build GRCh38
rs746692284 Chr.20: 36439804-36439804 on Build GRCh38
rs780950987 Chr.20: 36439913-36439913 on Build GRCh38
rs932141430 Chr.20: 36440067-36440067 on Build GRCh38
rs745448433 Chr.20: 36439711-36439711 on Build GRCh38
rs779624775 Chr.20: 36439899-36439899 on Build GRCh38
rs961571945 Chr.20: 36439844-36439844 on Build GRCh38
rs140243609 Chr.20: 36439847-36439847 on Build GRCh38
rs751922344 Chr.20: 36439758-36439758 on Build GRCh38
rs753202130 Chr.20: 36439766-36439766 on Build GRCh38
rs750481285 Chr.20: 36440129-36440129 on Build GRCh38
rs960393782 Chr.20: 36439775-36439775 on Build GRCh38
rs763150160 Chr.20: 36439746-36439746 on Build GRCh38

View Full Product Details