Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 36442682-36443189 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DLGAP4
Entrez Gene ID
22839
Gene Name
DLG associated protein 4
Gene Aliases
DAP-4, DAP4, DLP4, SAPAP-4, SAPAP4
UniGene
Hs.249600
Gene Chromosome Location
Chr.20: 36306323-36528637 on Build GRCh38
Transcripts
NM_014902.5 XM_011528688.2 XM_011528689.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6856424 c.1380delC p.Q462fs*14 Chr.20: 36442750-36442750 on Build GRCh38
4098010 c.1370C>T p.A457V Chr.20: 36442740-36442740 on Build GRCh38
4798704 c.1401G>A p.E467E Chr.20: 36442771-36442771 on Build GRCh38
5391946 c.1374C>T p.S458S Chr.20: 36442744-36442744 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs370797844 Chr.20: 36443127-36443127 on Build GRCh38
rs369961067 Chr.20: 36442863-36442863 on Build GRCh38
rs901024589 Chr.20: 36443139-36443139 on Build GRCh38
rs749843033 Chr.20: 36442920-36442920 on Build GRCh38
rs1006112586 Chr.20: 36443166-36443166 on Build GRCh38
rs749118205 Chr.20: 36442804-36442804 on Build GRCh38
rs754334756 Chr.20: 36442745-36442745 on Build GRCh38
rs770971856 Chr.20: 36442817-36442817 on Build GRCh38
rs377745326 Chr.20: 36442723-36442723 on Build GRCh38
rs908957462 Chr.20: 36442786-36442786 on Build GRCh38
rs565160255 Chr.20: 36443116-36443116 on Build GRCh38
rs114705683 Chr.20: 36443156-36443156 on Build GRCh38
rs1016074626 Chr.20: 36443023-36443023 on Build GRCh38
rs73277335 Chr.20: 36443090-36443090 on Build GRCh38
rs752032650 Chr.20: 36442819-36442819 on Build GRCh38
rs778889202 Chr.20: 36443112-36443112 on Build GRCh38
rs978199833 Chr.20: 36443050-36443050 on Build GRCh38
rs765802034 Chr.20: 36442751-36442751 on Build GRCh38
rs768090682 Chr.20: 36442891-36442891 on Build GRCh38
rs949082738 Chr.20: 36443045-36443045 on Build GRCh38
rs767912293 Chr.20: 36442705-36442705 on Build GRCh38
rs184562191 Chr.20: 36442759-36442759 on Build GRCh38
rs971021293 Chr.20: 36443030-36443030 on Build GRCh38
rs754562927 Chr.20: 36442762-36442762 on Build GRCh38
rs1007005199 Chr.20: 36442937-36442937 on Build GRCh38
rs375972834 Chr.20: 36442704-36442704 on Build GRCh38
rs936936604 Chr.20: 36443089-36443089 on Build GRCh38
rs554259183 Chr.20: 36443024-36443024 on Build GRCh38
rs11906476 Chr.20: 36443138-36443138 on Build GRCh38
rs879380638 Chr.20: 36442973-36442973 on Build GRCh38
rs780865130 Chr.20: 36442768-36442768 on Build GRCh38
rs201450784 Chr.20: 36442792-36442792 on Build GRCh38
rs565156599 Chr.20: 36442959-36442959 on Build GRCh38
rs188893471 Chr.20: 36443162-36443162 on Build GRCh38
rs942734260 Chr.20: 36443119-36443119 on Build GRCh38
rs889862633 Chr.20: 36442929-36442929 on Build GRCh38
rs1054234151 Chr.20: 36442714-36442714 on Build GRCh38
rs990540756 Chr.20: 36443038-36443038 on Build GRCh38
rs911413739 Chr.20: 36443096-36443096 on Build GRCh38
rs761078347 Chr.20: 36442735-36442735 on Build GRCh38
rs549925410 Chr.20: 36442856-36442856 on Build GRCh38
rs924211127 Chr.20: 36443059-36443059 on Build GRCh38
rs1041142997 Chr.20: 36443120-36443120 on Build GRCh38
rs754794399 Chr.20: 36443055-36443055 on Build GRCh38
rs747830819 Chr.20: 36442784-36442784 on Build GRCh38
rs995469368 Chr.20: 36443159-36443159 on Build GRCh38
rs932365962 Chr.20: 36443091-36443091 on Build GRCh38
rs772307100 Chr.20: 36442823-36442823 on Build GRCh38
rs901186744 Chr.20: 36442953-36442953 on Build GRCh38
rs950669672 Chr.20: 36442992-36442992 on Build GRCh38
rs755654900 Chr.20: 36442933-36442933 on Build GRCh38
rs1047534514 Chr.20: 36443143-36443143 on Build GRCh38
rs561721953 Chr.20: 36442979-36442979 on Build GRCh38
rs370232246 Chr.20: 36442740-36442740 on Build GRCh38
rs757787809 Chr.20: 36442750-36442750 on Build GRCh38
rs753598491 Chr.20: 36443020-36443020 on Build GRCh38
rs370648240 Chr.20: 36442796-36442796 on Build GRCh38
rs1027366762 Chr.20: 36443164-36443164 on Build GRCh38
rs1027109989 Chr.20: 36442962-36442962 on Build GRCh38
rs572124621 Chr.20: 36442943-36442943 on Build GRCh38
rs991011789 Chr.20: 36442997-36442997 on Build GRCh38
rs936819022 Chr.20: 36442914-36442914 on Build GRCh38
rs576023075 Chr.20: 36443039-36443039 on Build GRCh38
rs892867526 Chr.20: 36442744-36442744 on Build GRCh38

View Full Product Details