Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 38308850-38309334 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BPI
Entrez Gene ID
671
Gene Name
bactericidal/permeability-increasing protein
Gene Aliases
BPIFD1, rBPI
UniGene
Hs.529019
Gene Chromosome Location
Chr.20: 38304150-38337503 on Build GRCh38
Transcripts
NM_001725.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4485322 c.291C>T p.S97S Chr.20: 38308963-38308963 on Build GRCh38
6539409 c.348C>G p.I116M Chr.20: 38309020-38309020 on Build GRCh38
7161568 c.375A>G p.Q125Q Chr.20: 38309047-38309047 on Build GRCh38
4912667 c.315G>T p.V105V Chr.20: 38308987-38308987 on Build GRCh38
4001899 c.268C>T p.R90C Chr.20: 38308940-38308940 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs183225213 Chr.20: 38309179-38309179 on Build GRCh38
rs750582767 Chr.20: 38308888-38308888 on Build GRCh38
rs5743502 Chr.20: 38309029-38309029 on Build GRCh38
rs142539838 Chr.20: 38309304-38309304 on Build GRCh38
rs890231476 Chr.20: 38309000-38309000 on Build GRCh38
rs866528318 Chr.20: 38308964-38308964 on Build GRCh38
rs781578237 Chr.20: 38308930-38308930 on Build GRCh38
rs187906913 Chr.20: 38309222-38309222 on Build GRCh38
rs5743501 Chr.20: 38308996-38308996 on Build GRCh38
rs932191753 Chr.20: 38308987-38308987 on Build GRCh38
rs758406160 Chr.20: 38308903-38308903 on Build GRCh38
rs1005962877 Chr.20: 38309067-38309067 on Build GRCh38
rs1051504249 Chr.20: 38308995-38308995 on Build GRCh38
rs772964825 Chr.20: 38308998-38308998 on Build GRCh38
rs745392616 Chr.20: 38308941-38308941 on Build GRCh38
rs757893170 Chr.20: 38309104-38309104 on Build GRCh38
rs761505075 Chr.20: 38308983-38308983 on Build GRCh38
rs753172909 Chr.20: 38309016-38309016 on Build GRCh38
rs949402779 Chr.20: 38308909-38308909 on Build GRCh38
rs990210145 Chr.20: 38309197-38309197 on Build GRCh38
rs368208078 Chr.20: 38309062-38309062 on Build GRCh38
rs766599239 Chr.20: 38308905-38308905 on Build GRCh38
rs759609264 Chr.20: 38309072-38309072 on Build GRCh38
rs368730780 Chr.20: 38309270-38309270 on Build GRCh38
rs763041129 Chr.20: 38309003-38309003 on Build GRCh38
rs765068736 Chr.20: 38308887-38308887 on Build GRCh38
rs893102413 Chr.20: 38308977-38308977 on Build GRCh38
rs5743500 Chr.20: 38308940-38308940 on Build GRCh38
rs189724791 Chr.20: 38309100-38309100 on Build GRCh38
rs558691834 Chr.20: 38309121-38309121 on Build GRCh38
rs146270213 Chr.20: 38309063-38309063 on Build GRCh38
rs768486656 Chr.20: 38309048-38309048 on Build GRCh38
rs370368653 Chr.20: 38308947-38308947 on Build GRCh38
rs775945607 Chr.20: 38308915-38308915 on Build GRCh38
rs542938869 Chr.20: 38309012-38309012 on Build GRCh38
rs370285193 Chr.20: 38308894-38308894 on Build GRCh38
rs201211140 Chr.20: 38309110-38309110 on Build GRCh38
rs61339792 Chr.20: 38309059-38309059 on Build GRCh38
rs746695936 Chr.20: 38308960-38308960 on Build GRCh38
rs773230970 Chr.20: 38309060-38309060 on Build GRCh38
rs116227767 Chr.20: 38309076-38309076 on Build GRCh38
rs200771214 Chr.20: 38309030-38309030 on Build GRCh38
rs537205807 Chr.20: 38309118-38309118 on Build GRCh38
rs6024750 Chr.20: 38309041-38309041 on Build GRCh38
rs1004648486 Chr.20: 38309004-38309004 on Build GRCh38
rs749971039 Chr.20: 38309128-38309128 on Build GRCh38
rs5743503 Chr.20: 38309077-38309077 on Build GRCh38
rs775307677 Chr.20: 38308949-38308949 on Build GRCh38
rs763939187 Chr.20: 38308883-38308883 on Build GRCh38
rs776543196 Chr.20: 38308976-38308976 on Build GRCh38
rs936709202 Chr.20: 38309108-38309108 on Build GRCh38
rs150762103 Chr.20: 38309197-38309198 on Build GRCh38
rs376494392 Chr.20: 38309103-38309103 on Build GRCh38
rs756703113 Chr.20: 38308934-38308934 on Build GRCh38
rs776079763 Chr.20: 38309054-38309054 on Build GRCh38
rs761854241 Chr.20: 38308884-38308884 on Build GRCh38
rs914592253 Chr.20: 38309257-38309257 on Build GRCh38
rs373704456 Chr.20: 38308932-38308932 on Build GRCh38
rs17197668 Chr.20: 38309127-38309127 on Build GRCh38
rs934746782 Chr.20: 38309180-38309180 on Build GRCh38
rs765285174 Chr.20: 38308974-38308974 on Build GRCh38
rs760562826 Chr.20: 38308881-38308881 on Build GRCh38
rs570447254 Chr.20: 38309042-38309042 on Build GRCh38
rs990910428 Chr.20: 38308876-38308876 on Build GRCh38
rs766404691 Chr.20: 38309010-38309010 on Build GRCh38
rs924793312 Chr.20: 38309141-38309141 on Build GRCh38
rs755368730 Chr.20: 38308929-38308929 on Build GRCh38
rs768398912 Chr.20: 38308968-38308968 on Build GRCh38
rs934757319 Chr.20: 38308943-38308943 on Build GRCh38
rs747988156 Chr.20: 38309056-38309056 on Build GRCh38
rs199987679 Chr.20: 38309105-38309105 on Build GRCh38
rs549880214 Chr.20: 38309033-38309033 on Build GRCh38
rs751729735 Chr.20: 38308914-38308914 on Build GRCh38
rs148087853 Chr.20: 38309011-38309011 on Build GRCh38
rs764328966 Chr.20: 38309102-38309102 on Build GRCh38
rs756370199 Chr.20: 38309018-38309018 on Build GRCh38

View Full Product Details