Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 38634658-38635203 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGAP40
Entrez Gene ID
343578
Gene Name
Rho GTPase activating protein 40
Gene Aliases
C20orf95, dJ1100H13.4
UniGene
Hs.451997
Gene Chromosome Location
Chr.20: 38601934-38650652 on Build GRCh38
Transcripts
NM_001164431.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6004584 c.57C>T p.L19L Chr.20: 38634739-38634739 on Build GRCh38
5662826 c.704C>G p.S235C Chr.20: 38634699-38634699 on Build GRCh38
6234437 c.727G>T p.A243S Chr.20: 38634722-38634722 on Build GRCh38
4961312 c.790+1G>A p.? Chr.20: 38634786-38634786 on Build GRCh38
6234438 c.40G>T p.A14S Chr.20: 38634722-38634722 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746394862 Chr.20: 38634726-38634726 on Build GRCh38
rs555966022 Chr.20: 38634974-38634974 on Build GRCh38
rs752769180 Chr.20: 38634721-38634721 on Build GRCh38
rs901127943 Chr.20: 38634756-38634756 on Build GRCh38
rs767340174 Chr.20: 38634906-38634906 on Build GRCh38
rs927921633 Chr.20: 38634877-38634880 on Build GRCh38
rs11481956 Chr.20: 38634887-38634888 on Build GRCh38
rs548790220 Chr.20: 38634951-38634951 on Build GRCh38
rs747244415 Chr.20: 38635024-38635024 on Build GRCh38
rs781138369 Chr.20: 38634715-38634715 on Build GRCh38
rs951303902 Chr.20: 38635105-38635105 on Build GRCh38
rs139698700 Chr.20: 38634947-38634947 on Build GRCh38
rs1032545347 Chr.20: 38634711-38634711 on Build GRCh38
rs969188726 Chr.20: 38635130-38635130 on Build GRCh38
rs529428617 Chr.20: 38634722-38634722 on Build GRCh38
rs866290962 Chr.20: 38634787-38634787 on Build GRCh38
rs868705841 Chr.20: 38635107-38635107 on Build GRCh38
rs902177592 Chr.20: 38635005-38635005 on Build GRCh38
rs911353885 Chr.20: 38634983-38634983 on Build GRCh38
rs144262104 Chr.20: 38634967-38634967 on Build GRCh38
rs112932208 Chr.20: 38634781-38634781 on Build GRCh38
rs749710569 Chr.20: 38634723-38634723 on Build GRCh38
rs917414399 Chr.20: 38635149-38635149 on Build GRCh38
rs16987449 Chr.20: 38634846-38634846 on Build GRCh38
rs1055910391 Chr.20: 38635148-38635148 on Build GRCh38
rs772571545 Chr.20: 38634739-38634739 on Build GRCh38
rs78673969 Chr.20: 38634765-38634765 on Build GRCh38
rs759408242 Chr.20: 38634815-38634815 on Build GRCh38
rs565139412 Chr.20: 38634959-38634959 on Build GRCh38
rs942874296 Chr.20: 38635004-38635004 on Build GRCh38
rs1032016150 Chr.20: 38635012-38635012 on Build GRCh38
rs773991586 Chr.20: 38634886-38634887 on Build GRCh38
rs79159966 Chr.20: 38634900-38634900 on Build GRCh38
rs539817449 Chr.20: 38635028-38635028 on Build GRCh38
rs532332399 Chr.20: 38635078-38635078 on Build GRCh38
rs909901026 Chr.20: 38635116-38635116 on Build GRCh38
rs779288510 Chr.20: 38634724-38634724 on Build GRCh38
rs35488620 Chr.20: 38634769-38634770 on Build GRCh38
rs778989865 Chr.20: 38635115-38635115 on Build GRCh38
rs868787629 Chr.20: 38634892-38634892 on Build GRCh38
rs956632216 Chr.20: 38635139-38635139 on Build GRCh38
rs397818479 Chr.20: 38634900-38634901 on Build GRCh38
rs749048560 Chr.20: 38634766-38634766 on Build GRCh38
rs371424502 Chr.20: 38634741-38634741 on Build GRCh38
rs1006660514 Chr.20: 38635056-38635056 on Build GRCh38
rs538243481 Chr.20: 38635007-38635007 on Build GRCh38
rs758477417 Chr.20: 38635171-38635171 on Build GRCh38
rs61737864 Chr.20: 38634740-38634740 on Build GRCh38
rs181720590 Chr.20: 38634960-38634960 on Build GRCh38
rs573578991 Chr.20: 38635099-38635099 on Build GRCh38
rs577610970 Chr.20: 38634994-38634994 on Build GRCh38
rs769476165 Chr.20: 38634746-38634746 on Build GRCh38
rs1041670084 Chr.20: 38635074-38635074 on Build GRCh38
rs934964160 Chr.20: 38634914-38634914 on Build GRCh38
rs75622778 Chr.20: 38634783-38634783 on Build GRCh38
rs1035296233 Chr.20: 38635138-38635138 on Build GRCh38
rs553686339 Chr.20: 38635041-38635041 on Build GRCh38
rs762613941 Chr.20: 38634760-38634760 on Build GRCh38
rs909919553 Chr.20: 38634871-38634871 on Build GRCh38
rs190809711 Chr.20: 38634823-38634823 on Build GRCh38
rs901438382 Chr.20: 38635081-38635081 on Build GRCh38
rs566663923 Chr.20: 38634945-38634945 on Build GRCh38
rs544943033 Chr.20: 38635131-38635131 on Build GRCh38
rs540899996 Chr.20: 38635118-38635118 on Build GRCh38
rs1018505904 Chr.20: 38635063-38635063 on Build GRCh38
rs368125920 Chr.20: 38634831-38634831 on Build GRCh38
rs754901517 Chr.20: 38634692-38634692 on Build GRCh38
rs772825570 Chr.20: 38634759-38634759 on Build GRCh38
rs952756429 Chr.20: 38634818-38634818 on Build GRCh38
rs984498131 Chr.20: 38635113-38635113 on Build GRCh38
rs879485744 Chr.20: 38635084-38635084 on Build GRCh38
rs914899755 Chr.20: 38635162-38635162 on Build GRCh38
rs943935455 Chr.20: 38634771-38634771 on Build GRCh38
rs1048462568 Chr.20: 38634860-38634860 on Build GRCh38

View Full Product Details