Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 38962256-38962741 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DHX35
Entrez Gene ID
60625
Gene Name
DEAH-box helicase 35
Gene Aliases
C20orf15, DDX35, KAIA0875
UniGene
Hs.444520
Gene Chromosome Location
Chr.20: 38962338-39039723 on Build GRCh38
Transcripts
NM_001190809.1 NM_021931.3 NR_033905.1 XM_011528972.1 XM_011528974.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7244626 c.40+8C>T p.? Chr.20: 38962415-38962415 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs938555224 Chr.20: 38962323-38962323 on Build GRCh38
rs532902757 Chr.20: 38962492-38962492 on Build GRCh38
rs566890776 Chr.20: 38962681-38962681 on Build GRCh38
rs763374555 Chr.20: 38962401-38962401 on Build GRCh38
rs757099608 Chr.20: 38962343-38962343 on Build GRCh38
rs896883893 Chr.20: 38962460-38962460 on Build GRCh38
rs770087110 Chr.20: 38962390-38962390 on Build GRCh38
rs545656904 Chr.20: 38962465-38962465 on Build GRCh38
rs980433819 Chr.20: 38962315-38962315 on Build GRCh38
rs961024083 Chr.20: 38962278-38962278 on Build GRCh38
rs748369566 Chr.20: 38962386-38962386 on Build GRCh38
rs927108371 Chr.20: 38962316-38962316 on Build GRCh38
rs890224011 Chr.20: 38962717-38962717 on Build GRCh38
rs532021618 Chr.20: 38962453-38962453 on Build GRCh38
rs1026968884 Chr.20: 38962581-38962581 on Build GRCh38
rs974560 Chr.20: 38962555-38962555 on Build GRCh38
rs913088022 Chr.20: 38962566-38962566 on Build GRCh38
rs777062738 Chr.20: 38962380-38962380 on Build GRCh38
rs753324859 Chr.20: 38962721-38962721 on Build GRCh38
rs554196926 Chr.20: 38962297-38962297 on Build GRCh38
rs368464643 Chr.20: 38962440-38962440 on Build GRCh38
rs563109353 Chr.20: 38962310-38962310 on Build GRCh38
rs772314837 Chr.20: 38962605-38962605 on Build GRCh38
rs949471508 Chr.20: 38962652-38962652 on Build GRCh38
rs1050693467 Chr.20: 38962634-38962634 on Build GRCh38
rs913345045 Chr.20: 38962368-38962368 on Build GRCh38
rs760294601 Chr.20: 38962420-38962420 on Build GRCh38
rs752174517 Chr.20: 38962356-38962356 on Build GRCh38
rs961891515 Chr.20: 38962331-38962331 on Build GRCh38
rs1019361471 Chr.20: 38962454-38962454 on Build GRCh38
rs534149650 Chr.20: 38962692-38962692 on Build GRCh38
rs946108745 Chr.20: 38962573-38962573 on Build GRCh38
rs900155893 Chr.20: 38962504-38962504 on Build GRCh38
rs1027188840 Chr.20: 38962714-38962714 on Build GRCh38
rs144338277 Chr.20: 38962637-38962637 on Build GRCh38
rs769579320 Chr.20: 38962526-38962526 on Build GRCh38
rs370695428 Chr.20: 38962407-38962407 on Build GRCh38
rs773718942 Chr.20: 38962397-38962397 on Build GRCh38
rs1028519299 Chr.20: 38962292-38962292 on Build GRCh38
rs540282495 Chr.20: 38962285-38962285 on Build GRCh38
rs938118831 Chr.20: 38962404-38962404 on Build GRCh38
rs368443589 Chr.20: 38962379-38962379 on Build GRCh38
rs374592674 Chr.20: 38962417-38962417 on Build GRCh38
rs972421917 Chr.20: 38962286-38962286 on Build GRCh38
rs574172267 Chr.20: 38962715-38962715 on Build GRCh38
rs758389648 Chr.20: 38962349-38962349 on Build GRCh38
rs936601916 Chr.20: 38962680-38962680 on Build GRCh38
rs755533669 Chr.20: 38962325-38962325 on Build GRCh38
rs779807363 Chr.20: 38962365-38962365 on Build GRCh38
rs1009105605 Chr.20: 38962444-38962444 on Build GRCh38
rs550769325 Chr.20: 38962430-38962430 on Build GRCh38
rs898003950 Chr.20: 38962706-38962706 on Build GRCh38
rs574184454 Chr.20: 38962300-38962300 on Build GRCh38
rs977970345 Chr.20: 38962653-38962653 on Build GRCh38
rs753480397 Chr.20: 38962341-38962341 on Build GRCh38
rs752287525 Chr.20: 38962324-38962324 on Build GRCh38
rs1034262089 Chr.20: 38962535-38962535 on Build GRCh38
rs151212426 Chr.20: 38962377-38962377 on Build GRCh38
rs1051832221 Chr.20: 38962709-38962709 on Build GRCh38
rs557073357 Chr.20: 38962470-38962470 on Build GRCh38
rs995913347 Chr.20: 38962284-38962284 on Build GRCh38
rs980038623 Chr.20: 38962530-38962530 on Build GRCh38
rs546284855 Chr.20: 38962501-38962501 on Build GRCh38
rs1057134677 Chr.20: 38962333-38962333 on Build GRCh38
rs890740680 Chr.20: 38962645-38962645 on Build GRCh38
rs925939563 Chr.20: 38962600-38962600 on Build GRCh38
rs568737285 Chr.20: 38962565-38962565 on Build GRCh38
rs140245638 Chr.20: 38962406-38962406 on Build GRCh38
rs974559 Chr.20: 38962718-38962718 on Build GRCh38
rs1015855572 Chr.20: 38962318-38962318 on Build GRCh38
rs543240824 Chr.20: 38962309-38962309 on Build GRCh38
rs1036502615 Chr.20: 38962697-38962697 on Build GRCh38
rs1053473200 Chr.20: 38962488-38962491 on Build GRCh38
rs373968541 Chr.20: 38962347-38962347 on Build GRCh38
rs140789922 Chr.20: 38962470-38962470 on Build GRCh38
rs974561 Chr.20: 38962541-38962541 on Build GRCh38
rs553925245 Chr.20: 38962707-38962707 on Build GRCh38
rs932219223 Chr.20: 38962416-38962416 on Build GRCh38
rs900208164 Chr.20: 38962687-38962687 on Build GRCh38
rs995960769 Chr.20: 38962524-38962524 on Build GRCh38
rs1026615601 Chr.20: 38962509-38962509 on Build GRCh38
rs946030207 Chr.20: 38962371-38962371 on Build GRCh38
rs3752294 Chr.20: 38962515-38962515 on Build GRCh38
rs537664905 Chr.20: 38962593-38962593 on Build GRCh38
rs763844769 Chr.20: 38962327-38962327 on Build GRCh38
rs1051123617 Chr.20: 38962424-38962424 on Build GRCh38
rs923751654 Chr.20: 38962656-38962656 on Build GRCh38
rs551617660 Chr.20: 38962594-38962594 on Build GRCh38
rs959366235 Chr.20: 38962584-38962585 on Build GRCh38

View Full Product Details