Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 43620721-43621221 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IFT52
Entrez Gene ID
51098
Gene Name
intraflagellar transport 52
Gene Aliases
C20orf9, CGI-53, NGD2, NGD5
UniGene
Hs.444332
Gene Chromosome Location
Chr.20: 43590613-43647296 on Build GRCh38
Transcripts
NM_001303458.2 NM_001303459.2 NM_001323578.1 NM_001323579.1 NM_001323580.1 NM_001323581.1 NM_016004.4 XM_017027863.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6017905 c.736C>T p.H246Y Chr.20: 43620893-43620893 on Build GRCh38
4973006 c.768+1G>A p.? Chr.20: 43620926-43620926 on Build GRCh38
1026991 c.723G>A p.T241T Chr.20: 43620880-43620880 on Build GRCh38
4691878 c.704T>C p.V235A Chr.20: 43620861-43620861 on Build GRCh38
5366160 c.745C>G p.Q249E Chr.20: 43620902-43620902 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs983999352 Chr.20: 43621126-43621126 on Build GRCh38
rs1024279707 Chr.20: 43620990-43620990 on Build GRCh38
rs762776083 Chr.20: 43620935-43620935 on Build GRCh38
rs781324889 Chr.20: 43620861-43620861 on Build GRCh38
rs779305915 Chr.20: 43620880-43620880 on Build GRCh38
rs775338091 Chr.20: 43620885-43620886 on Build GRCh38
rs766658166 Chr.20: 43621104-43621104 on Build GRCh38
rs746968962 Chr.20: 43620840-43620840 on Build GRCh38
rs774276132 Chr.20: 43620945-43620945 on Build GRCh38
rs1046988374 Chr.20: 43621036-43621039 on Build GRCh38
rs970040715 Chr.20: 43621009-43621009 on Build GRCh38
rs779645327 Chr.20: 43621100-43621100 on Build GRCh38
rs753909529 Chr.20: 43620972-43620972 on Build GRCh38
rs760384960 Chr.20: 43620814-43620814 on Build GRCh38
rs567507538 Chr.20: 43620839-43620840 on Build GRCh38
rs956296240 Chr.20: 43620893-43620893 on Build GRCh38
rs1018351366 Chr.20: 43621090-43621090 on Build GRCh38
rs772567125 Chr.20: 43620888-43620888 on Build GRCh38
rs562928780 Chr.20: 43621103-43621103 on Build GRCh38
rs11483104 Chr.20: 43620840-43620841 on Build GRCh38
rs772818867 Chr.20: 43620934-43620934 on Build GRCh38
rs878885044 Chr.20: 43620993-43620993 on Build GRCh38
rs756402041 Chr.20: 43620962-43620962 on Build GRCh38
rs754799617 Chr.20: 43620858-43620858 on Build GRCh38
rs373726921 Chr.20: 43620879-43620879 on Build GRCh38
rs773463562 Chr.20: 43621044-43621044 on Build GRCh38
rs1038245937 Chr.20: 43620986-43620986 on Build GRCh38
rs750312800 Chr.20: 43620834-43620834 on Build GRCh38
rs925442531 Chr.20: 43620865-43620865 on Build GRCh38
rs949383853 Chr.20: 43620913-43620913 on Build GRCh38
rs908035774 Chr.20: 43621138-43621138 on Build GRCh38
rs199638691 Chr.20: 43620856-43620856 on Build GRCh38
rs543133112 Chr.20: 43620984-43620984 on Build GRCh38
rs747866295 Chr.20: 43620914-43620914 on Build GRCh38
rs1039975036 Chr.20: 43620774-43620774 on Build GRCh38
rs766057363 Chr.20: 43620849-43620849 on Build GRCh38
rs777978967 Chr.20: 43620874-43620874 on Build GRCh38
rs150320619 Chr.20: 43620839-43620839 on Build GRCh38
rs113286819 Chr.20: 43620744-43620744 on Build GRCh38
rs530334997 Chr.20: 43621155-43621155 on Build GRCh38
rs769180754 Chr.20: 43620927-43620927 on Build GRCh38
rs944488195 Chr.20: 43620769-43620769 on Build GRCh38
rs760635591 Chr.20: 43620850-43620851 on Build GRCh38
rs746190968 Chr.20: 43620882-43620882 on Build GRCh38
rs374545109 Chr.20: 43621045-43621045 on Build GRCh38
rs993925333 Chr.20: 43620992-43620992 on Build GRCh38
rs752931776 Chr.20: 43620868-43620868 on Build GRCh38
rs141077205 Chr.20: 43620838-43620838 on Build GRCh38
rs747990194 Chr.20: 43620838-43620839 on Build GRCh38
rs746453842 Chr.20: 43621119-43621119 on Build GRCh38
rs776449925 Chr.20: 43620816-43620816 on Build GRCh38
rs5841515 Chr.20: 43620849-43620850 on Build GRCh38
rs767359586 Chr.20: 43620955-43620955 on Build GRCh38
rs187230020 Chr.20: 43620815-43620815 on Build GRCh38
rs995832187 Chr.20: 43620800-43620800 on Build GRCh38
rs1555348 Chr.20: 43620758-43620758 on Build GRCh38
rs367609987 Chr.20: 43620953-43620953 on Build GRCh38
rs869169609 Chr.20: 43620850-43620851 on Build GRCh38
rs1001025251 Chr.20: 43621110-43621110 on Build GRCh38
rs891026019 Chr.20: 43621056-43621056 on Build GRCh38
rs1032061258 Chr.20: 43621147-43621147 on Build GRCh38
rs776361854 Chr.20: 43620906-43620906 on Build GRCh38
rs766299974 Chr.20: 43620936-43620936 on Build GRCh38
rs763756258 Chr.20: 43620851-43620852 on Build GRCh38
rs753946862 Chr.20: 43620933-43620933 on Build GRCh38
rs756176279 Chr.20: 43620869-43620869 on Build GRCh38
rs1007924567 Chr.20: 43621060-43621060 on Build GRCh38
rs199949818 Chr.20: 43620850-43620850 on Build GRCh38
rs761337893 Chr.20: 43620827-43620827 on Build GRCh38
rs113765987 Chr.20: 43620959-43620959 on Build GRCh38
rs900184829 Chr.20: 43620789-43620789 on Build GRCh38
rs751404101 Chr.20: 43620854-43620854 on Build GRCh38
rs764813414 Chr.20: 43620828-43620828 on Build GRCh38
rs764062708 Chr.20: 43620970-43620970 on Build GRCh38
rs952341734 Chr.20: 43621120-43621120 on Build GRCh38
rs749731647 Chr.20: 43620877-43620877 on Build GRCh38
rs980140183 Chr.20: 43621033-43621033 on Build GRCh38

View Full Product Details