Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 44622509-44623019 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADA
Entrez Gene ID
100
Gene Name
adenosine deaminase
Gene Aliases
-
UniGene
Hs.654536
Gene Chromosome Location
Chr.20: 44619519-44651758 on Build GRCh38
Transcripts
NM_000022.3 NM_001322050.1 NM_001322051.1 NR_136160.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3546906 c.777C>T p.F259F Chr.20: 44622832-44622832 on Build GRCh38
4098722 c.695A>T p.K232M Chr.20: 44622914-44622914 on Build GRCh38
6851689 c.672A>C p.V224V Chr.20: 44623013-44623013 on Build GRCh38
3546907 c.679-1G>A p.? Chr.20: 44622931-44622931 on Build GRCh38
4952897 c.778G>A p.E260K Chr.20: 44622831-44622831 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs546350234 Chr.20: 44622769-44622769 on Build GRCh38
rs764386125 Chr.20: 44622695-44622695 on Build GRCh38
rs762216021 Chr.20: 44622856-44622856 on Build GRCh38
rs528680964 Chr.20: 44622596-44622596 on Build GRCh38
rs148994526 Chr.20: 44622851-44622851 on Build GRCh38
rs749159675 Chr.20: 44622820-44622820 on Build GRCh38
rs775436011 Chr.20: 44622921-44622921 on Build GRCh38
rs759743522 Chr.20: 44622687-44622687 on Build GRCh38
rs773349681 Chr.20: 44622559-44622559 on Build GRCh38
rs994812351 Chr.20: 44622752-44622752 on Build GRCh38
rs899193277 Chr.20: 44622740-44622740 on Build GRCh38
rs368646586 Chr.20: 44622810-44622810 on Build GRCh38
rs886337650 Chr.20: 44622766-44622766 on Build GRCh38
rs904442047 Chr.20: 44622965-44622965 on Build GRCh38
rs772279816 Chr.20: 44622918-44622918 on Build GRCh38
rs754172068 Chr.20: 44622701-44622701 on Build GRCh38
rs768596356 Chr.20: 44622924-44622924 on Build GRCh38
rs780717460 Chr.20: 44622796-44622796 on Build GRCh38
rs751244757 Chr.20: 44622863-44622863 on Build GRCh38
rs763198534 Chr.20: 44622571-44622571 on Build GRCh38
rs1047697626 Chr.20: 44622925-44622925 on Build GRCh38
rs121908738 Chr.20: 44622612-44622612 on Build GRCh38
rs761204032 Chr.20: 44622692-44622692 on Build GRCh38
rs778809577 Chr.20: 44622906-44622906 on Build GRCh38
rs562095440 Chr.20: 44622611-44622611 on Build GRCh38
rs182482421 Chr.20: 44622753-44622753 on Build GRCh38
rs201219441 Chr.20: 44622794-44622794 on Build GRCh38
rs756023598 Chr.20: 44622808-44622808 on Build GRCh38
rs904398666 Chr.20: 44622705-44622705 on Build GRCh38
rs375562913 Chr.20: 44622936-44622936 on Build GRCh38
rs370902329 Chr.20: 44622968-44622968 on Build GRCh38
rs781040248 Chr.20: 44622640-44622640 on Build GRCh38
rs1044495128 Chr.20: 44622738-44622738 on Build GRCh38
rs368087583 Chr.20: 44622785-44622785 on Build GRCh38
rs776548198 Chr.20: 44622550-44622550 on Build GRCh38
rs1000953688 Chr.20: 44622734-44622736 on Build GRCh38
rs757614845 Chr.20: 44622783-44622783 on Build GRCh38
rs368607629 Chr.20: 44622982-44622982 on Build GRCh38
rs559798694 Chr.20: 44622589-44622589 on Build GRCh38
rs748088317 Chr.20: 44622648-44622648 on Build GRCh38
rs772309012 Chr.20: 44622832-44622832 on Build GRCh38
rs374812597 Chr.20: 44622656-44622656 on Build GRCh38
rs757796081 Chr.20: 44622647-44622648 on Build GRCh38
rs761098217 Chr.20: 44622844-44622844 on Build GRCh38
rs774759287 Chr.20: 44622823-44622823 on Build GRCh38
rs550893200 Chr.20: 44622666-44622666 on Build GRCh38
rs934826988 Chr.20: 44622689-44622689 on Build GRCh38
rs116962828 Chr.20: 44622599-44622599 on Build GRCh38
rs762269064 Chr.20: 44622956-44622956 on Build GRCh38
rs759196142 Chr.20: 44622977-44622977 on Build GRCh38
rs371862936 Chr.20: 44622822-44622822 on Build GRCh38
rs1013028412 Chr.20: 44622723-44622723 on Build GRCh38
rs756480576 Chr.20: 44622606-44622606 on Build GRCh38
rs755578452 Chr.20: 44622880-44622880 on Build GRCh38
rs1033841912 Chr.20: 44622812-44622812 on Build GRCh38
rs751635016 Chr.20: 44622588-44622588 on Build GRCh38
rs749544011 Chr.20: 44622994-44622994 on Build GRCh38
rs770035190 Chr.20: 44622865-44622866 on Build GRCh38
rs757155433 Chr.20: 44622901-44622901 on Build GRCh38
rs752274920 Chr.20: 44622988-44622988 on Build GRCh38
rs781605283 Chr.20: 44622655-44622656 on Build GRCh38
rs545935345 Chr.20: 44622558-44622558 on Build GRCh38
rs977746470 Chr.20: 44622802-44622802 on Build GRCh38
rs769511813 Chr.20: 44622661-44622661 on Build GRCh38
rs145863450 Chr.20: 44622839-44622839 on Build GRCh38
rs6031679 Chr.20: 44622793-44622793 on Build GRCh38
rs746370947 Chr.20: 44622690-44622690 on Build GRCh38
rs570514588 Chr.20: 44622707-44622707 on Build GRCh38
rs948856707 Chr.20: 44622887-44622887 on Build GRCh38
rs370450947 Chr.20: 44622633-44622633 on Build GRCh38
rs770169599 Chr.20: 44622959-44622959 on Build GRCh38
rs773510057 Chr.20: 44622964-44622964 on Build GRCh38
rs748062599 Chr.20: 44622803-44622803 on Build GRCh38
rs747172905 Chr.20: 44622922-44622922 on Build GRCh38
rs777820729 Chr.20: 44622894-44622894 on Build GRCh38
rs773016155 Chr.20: 44622664-44622664 on Build GRCh38
rs766590645 Chr.20: 44622587-44622587 on Build GRCh38
rs1008700370 Chr.20: 44622779-44622779 on Build GRCh38
rs778167423 Chr.20: 44622608-44622608 on Build GRCh38
rs79281338 Chr.20: 44622905-44622905 on Build GRCh38
rs761572248 Chr.20: 44622554-44622554 on Build GRCh38
rs886056707 Chr.20: 44622938-44622938 on Build GRCh38
rs1057292007 Chr.20: 44622710-44622710 on Build GRCh38
rs746052951 Chr.20: 44622828-44622828 on Build GRCh38
rs201944717 Chr.20: 44622852-44622852 on Build GRCh38
rs760746989 Chr.20: 44622533-44622533 on Build GRCh38
rs760277918 Chr.20: 44622993-44622993 on Build GRCh38
rs754578956 Chr.20: 44622870-44622870 on Build GRCh38
rs771213761 Chr.20: 44622669-44622669 on Build GRCh38
rs577317154 Chr.20: 44622895-44622895 on Build GRCh38
rs766952213 Chr.20: 44622876-44622876 on Build GRCh38
rs774708098 Chr.20: 44622674-44622674 on Build GRCh38
rs542229902 Chr.20: 44622602-44622602 on Build GRCh38
rs775691787 Chr.20: 44622961-44622962 on Build GRCh38
rs895973555 Chr.20: 44622719-44622719 on Build GRCh38
rs547700655 Chr.20: 44622995-44622995 on Build GRCh38
rs973595980 Chr.20: 44622534-44622534 on Build GRCh38
rs121908729 Chr.20: 44622911-44622911 on Build GRCh38
rs767679026 Chr.20: 44622691-44622691 on Build GRCh38
rs765555325 Chr.20: 44622861-44622861 on Build GRCh38
rs968126152 Chr.20: 44622801-44622801 on Build GRCh38

View Full Product Details