Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 45801798-45802300 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNTTIP1
Entrez Gene ID
116092
Gene Name
deoxynucleotidyltransferase, terminal, interacting protein 1
Gene Aliases
C20orf167, Tdif1, dJ447F3.4
UniGene
Hs.472852
Gene Chromosome Location
Chr.20: 45791937-45811427 on Build GRCh38
Transcripts
NM_052951.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4098861 c.535G>A p.D179N Chr.20: 45802035-45802035 on Build GRCh38
6876608 c.539G>A p.R180Q Chr.20: 45802039-45802039 on Build GRCh38
3841133 c.505G>A p.G169R Chr.20: 45802005-45802005 on Build GRCh38
1027281 c.508C>T p.R170W Chr.20: 45802008-45802008 on Build GRCh38
3841134 c.506G>A p.G169E Chr.20: 45802006-45802006 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs575335661 Chr.20: 45802050-45802050 on Build GRCh38
rs761023465 Chr.20: 45802066-45802066 on Build GRCh38
rs750255975 Chr.20: 45801922-45801922 on Build GRCh38
rs774736151 Chr.20: 45801997-45801997 on Build GRCh38
rs746406509 Chr.20: 45802071-45802071 on Build GRCh38
rs901772315 Chr.20: 45802121-45802121 on Build GRCh38
rs553678868 Chr.20: 45801902-45801902 on Build GRCh38
rs1050449211 Chr.20: 45802126-45802126 on Build GRCh38
rs547826766 Chr.20: 45801844-45801844 on Build GRCh38
rs779805577 Chr.20: 45801967-45801967 on Build GRCh38
rs566920726 Chr.20: 45801942-45801942 on Build GRCh38
rs1004854552 Chr.20: 45802132-45802132 on Build GRCh38
rs779850414 Chr.20: 45801952-45801952 on Build GRCh38
rs748016365 Chr.20: 45802090-45802090 on Build GRCh38
rs371351416 Chr.20: 45802082-45802082 on Build GRCh38
rs767804559 Chr.20: 45802017-45802017 on Build GRCh38
rs773469565 Chr.20: 45801971-45801971 on Build GRCh38
rs576950227 Chr.20: 45802204-45802204 on Build GRCh38
rs368778767 Chr.20: 45802060-45802060 on Build GRCh38
rs930456793 Chr.20: 45801901-45801901 on Build GRCh38
rs185750172 Chr.20: 45802039-45802039 on Build GRCh38
rs4810471 Chr.20: 45802130-45802130 on Build GRCh38
rs781472079 Chr.20: 45801985-45801986 on Build GRCh38
rs539951817 Chr.20: 45802150-45802150 on Build GRCh38
rs6094212 Chr.20: 45801898-45801898 on Build GRCh38
rs377103727 Chr.20: 45802083-45802083 on Build GRCh38
rs750000048 Chr.20: 45802034-45802034 on Build GRCh38
rs772085900 Chr.20: 45801951-45801951 on Build GRCh38
rs765834846 Chr.20: 45802079-45802079 on Build GRCh38
rs770211942 Chr.20: 45802049-45802049 on Build GRCh38
rs1011700853 Chr.20: 45802250-45802250 on Build GRCh38
rs201221160 Chr.20: 45802008-45802008 on Build GRCh38
rs916974570 Chr.20: 45802073-45802073 on Build GRCh38
rs759984735 Chr.20: 45802058-45802058 on Build GRCh38
rs767984941 Chr.20: 45802251-45802251 on Build GRCh38
rs746878056 Chr.20: 45801954-45801954 on Build GRCh38
rs950224149 Chr.20: 45802086-45802086 on Build GRCh38
rs41282768 Chr.20: 45802101-45802101 on Build GRCh38
rs996556831 Chr.20: 45802122-45802123 on Build GRCh38
rs148955121 Chr.20: 45802027-45802027 on Build GRCh38
rs1044637688 Chr.20: 45801828-45801828 on Build GRCh38
rs772080042 Chr.20: 45802158-45802158 on Build GRCh38
rs542207919 Chr.20: 45802209-45802209 on Build GRCh38
rs41282766 Chr.20: 45802041-45802041 on Build GRCh38
rs901889436 Chr.20: 45802241-45802241 on Build GRCh38
rs766970182 Chr.20: 45801992-45801992 on Build GRCh38
rs1005398892 Chr.20: 45801912-45801912 on Build GRCh38
rs779556936 Chr.20: 45802038-45802038 on Build GRCh38
rs995049216 Chr.20: 45801882-45801882 on Build GRCh38
rs750088161 Chr.20: 45802071-45802071 on Build GRCh38
rs778908757 Chr.20: 45802102-45802102 on Build GRCh38
rs200334919 Chr.20: 45802028-45802028 on Build GRCh38
rs781152194 Chr.20: 45802046-45802046 on Build GRCh38
rs886454904 Chr.20: 45802112-45802112 on Build GRCh38
rs199577746 Chr.20: 45802069-45802069 on Build GRCh38
rs147664923 Chr.20: 45802056-45802056 on Build GRCh38
rs201888261 Chr.20: 45801985-45801985 on Build GRCh38
rs367936627 Chr.20: 45801938-45801938 on Build GRCh38
rs745359179 Chr.20: 45801950-45801950 on Build GRCh38
rs770922215 Chr.20: 45802051-45802051 on Build GRCh38
rs758000448 Chr.20: 45802077-45802077 on Build GRCh38
rs141065268 Chr.20: 45801861-45801861 on Build GRCh38
rs768346835 Chr.20: 45801965-45801965 on Build GRCh38
rs777645712 Chr.20: 45802093-45802093 on Build GRCh38
rs757830632 Chr.20: 45802037-45802037 on Build GRCh38
rs544129761 Chr.20: 45802259-45802259 on Build GRCh38
rs369671027 Chr.20: 45802099-45802099 on Build GRCh38
rs753011518 Chr.20: 45802022-45802022 on Build GRCh38
rs1040721544 Chr.20: 45802111-45802111 on Build GRCh38
rs408911 Chr.20: 45802047-45802047 on Build GRCh38
rs73306486 Chr.20: 45802157-45802157 on Build GRCh38
rs1048874473 Chr.20: 45802089-45802089 on Build GRCh38
rs536221701 Chr.20: 45802065-45802065 on Build GRCh38
rs754916575 Chr.20: 45802085-45802085 on Build GRCh38
rs370375348 Chr.20: 45802026-45802026 on Build GRCh38
rs781274936 Chr.20: 45802088-45802088 on Build GRCh38
rs990520848 Chr.20: 45802053-45802053 on Build GRCh38

View Full Product Details