Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 46174096-46174609 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDH22
Entrez Gene ID
64405
Gene Name
cadherin 22
Gene Aliases
C20orf25, dJ998H6.1
UniGene
Hs.472861
Gene Chromosome Location
Chr.20: 46173733-46308498 on Build GRCh38
Transcripts
NM_021248.2 XM_011528994.2 XM_011528995.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7299321 c.2420C>T p.P807L Chr.20: 46174573-46174573 on Build GRCh38
5646041 c.2470G>A p.E824K Chr.20: 46174523-46174523 on Build GRCh38
3389795 c.2429G>A p.R810Q Chr.20: 46174564-46174564 on Build GRCh38
6443891 c.2462G>A p.G821E Chr.20: 46174531-46174531 on Build GRCh38
6441383 c.2475_2476CC>TT p.Q826* Chr.20: 46174517-46174518 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs571611226 Chr.20: 46174360-46174360 on Build GRCh38
rs914060491 Chr.20: 46174203-46174203 on Build GRCh38
rs953108370 Chr.20: 46174168-46174168 on Build GRCh38
rs1050821099 Chr.20: 46174228-46174228 on Build GRCh38
rs1025002620 Chr.20: 46174385-46174385 on Build GRCh38
rs1007333924 Chr.20: 46174437-46174437 on Build GRCh38
rs892542390 Chr.20: 46174508-46174508 on Build GRCh38
rs113966253 Chr.20: 46174499-46174499 on Build GRCh38
rs957719813 Chr.20: 46174493-46174493 on Build GRCh38
rs747375025 Chr.20: 46174529-46174529 on Build GRCh38
rs924954207 Chr.20: 46174190-46174190 on Build GRCh38
rs745508542 Chr.20: 46174489-46174489 on Build GRCh38
rs1013693322 Chr.20: 46174514-46174514 on Build GRCh38
rs1000710304 Chr.20: 46174320-46174320 on Build GRCh38
rs551168889 Chr.20: 46174429-46174429 on Build GRCh38
rs997717315 Chr.20: 46174378-46174378 on Build GRCh38
rs565204995 Chr.20: 46174326-46174326 on Build GRCh38
rs890107665 Chr.20: 46174177-46174177 on Build GRCh38
rs925196438 Chr.20: 46174488-46174488 on Build GRCh38
rs778330224 Chr.20: 46174456-46174456 on Build GRCh38
rs1016938489 Chr.20: 46174391-46174391 on Build GRCh38
rs547074596 Chr.20: 46174238-46174238 on Build GRCh38
rs79797488 Chr.20: 46174554-46174554 on Build GRCh38
rs938110764 Chr.20: 46174252-46174252 on Build GRCh38
rs879133117 Chr.20: 46174176-46174176 on Build GRCh38
rs905253453 Chr.20: 46174250-46174250 on Build GRCh38
rs1002623206 Chr.20: 46174542-46174542 on Build GRCh38
rs766967938 Chr.20: 46174535-46174535 on Build GRCh38
rs929761213 Chr.20: 46174405-46174405 on Build GRCh38
rs573657441 Chr.20: 46174167-46174167 on Build GRCh38
rs926814015 Chr.20: 46174557-46174557 on Build GRCh38
rs373065110 Chr.20: 46174365-46174365 on Build GRCh38
rs986854581 Chr.20: 46174422-46174422 on Build GRCh38
rs758040732 Chr.20: 46174492-46174492 on Build GRCh38
rs918332293 Chr.20: 46174394-46174394 on Build GRCh38
rs888472716 Chr.20: 46174283-46174283 on Build GRCh38
rs970025185 Chr.20: 46174388-46174388 on Build GRCh38
rs942658147 Chr.20: 46174256-46174256 on Build GRCh38
rs3746822 Chr.20: 46174358-46174358 on Build GRCh38
rs988409018 Chr.20: 46174500-46174500 on Build GRCh38
rs367642695 Chr.20: 46174336-46174336 on Build GRCh38
rs936883435 Chr.20: 46174191-46174191 on Build GRCh38
rs528617859 Chr.20: 46174233-46174233 on Build GRCh38
rs1006083266 Chr.20: 46174289-46174289 on Build GRCh38
rs1036720541 Chr.20: 46174339-46174339 on Build GRCh38
rs905161469 Chr.20: 46174541-46174541 on Build GRCh38
rs769091962 Chr.20: 46174550-46174550 on Build GRCh38
rs1048360451 Chr.20: 46174271-46174271 on Build GRCh38
rs938081050 Chr.20: 46174561-46174561 on Build GRCh38
rs769886200 Chr.20: 46174573-46174573 on Build GRCh38
rs1031055071 Chr.20: 46174328-46174328 on Build GRCh38
rs981020933 Chr.20: 46174390-46174390 on Build GRCh38
rs530686454 Chr.20: 46174120-46174120 on Build GRCh38
rs949624076 Chr.20: 46174210-46174210 on Build GRCh38
rs780459936 Chr.20: 46174515-46174515 on Build GRCh38
rs963479780 Chr.20: 46174392-46174392 on Build GRCh38
rs914101390 Chr.20: 46174505-46174505 on Build GRCh38
rs975847397 Chr.20: 46174136-46174136 on Build GRCh38
rs988821119 Chr.20: 46174192-46174192 on Build GRCh38
rs1046563101 Chr.20: 46174215-46174215 on Build GRCh38
rs561320436 Chr.20: 46174186-46174186 on Build GRCh38
rs748186611 Chr.20: 46174572-46174572 on Build GRCh38
rs963636374 Chr.20: 46174576-46174576 on Build GRCh38
rs903660519 Chr.20: 46174313-46174315 on Build GRCh38
rs1030170100 Chr.20: 46174409-46174409 on Build GRCh38

View Full Product Details