Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 49058341-49058821 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSE1L
Entrez Gene ID
1434
Gene Name
chromosome segregation 1 like
Gene Aliases
CAS, CSE1, XPO2
UniGene
Hs.90073
Gene Chromosome Location
Chr.20: 49046246-49096960 on Build GRCh38
Transcripts
NM_001256135.1 NM_001316.3 NR_045796.1 XM_011528599.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6160012 c.16G>T p.A6S Chr.20: 49058479-49058479 on Build GRCh38
1307496 c.13G>A p.D5N Chr.20: 49058476-49058476 on Build GRCh38
3991766 c.25C>A p.Q9K Chr.20: 49058488-49058488 on Build GRCh38
4932302 c.54A>G p.T18T Chr.20: 49058517-49058517 on Build GRCh38
5623491 c.61C>A p.P21T Chr.20: 49058524-49058524 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1028723422 Chr.20: 49058474-49058474 on Build GRCh38
rs185750141 Chr.20: 49058384-49058384 on Build GRCh38
rs528654513 Chr.20: 49058390-49058390 on Build GRCh38
rs139770170 Chr.20: 49058575-49058575 on Build GRCh38
rs939889415 Chr.20: 49058743-49058744 on Build GRCh38
rs773565545 Chr.20: 49058456-49058456 on Build GRCh38
rs112688645 Chr.20: 49058654-49058654 on Build GRCh38
rs1006733513 Chr.20: 49058583-49058583 on Build GRCh38
rs535966776 Chr.20: 49058617-49058617 on Build GRCh38
rs760623788 Chr.20: 49058475-49058475 on Build GRCh38
rs770183164 Chr.20: 49058453-49058453 on Build GRCh38
rs772419337 Chr.20: 49058413-49058413 on Build GRCh38
rs778601313 Chr.20: 49058511-49058511 on Build GRCh38
rs940842993 Chr.20: 49058740-49058740 on Build GRCh38
rs886950682 Chr.20: 49058394-49058394 on Build GRCh38
rs768554048 Chr.20: 49058795-49058795 on Build GRCh38
rs981603137 Chr.20: 49058667-49058667 on Build GRCh38
rs532820443 Chr.20: 49058563-49058563 on Build GRCh38
rs371482865 Chr.20: 49058690-49058690 on Build GRCh38
rs1038287579 Chr.20: 49058426-49058426 on Build GRCh38
rs775310683 Chr.20: 49058600-49058600 on Build GRCh38
rs937058539 Chr.20: 49058707-49058707 on Build GRCh38
rs747648935 Chr.20: 49058632-49058632 on Build GRCh38
rs750060417 Chr.20: 49058517-49058517 on Build GRCh38
rs769155749 Chr.20: 49058542-49058542 on Build GRCh38
rs192118244 Chr.20: 49058770-49058770 on Build GRCh38
rs917453107 Chr.20: 49058734-49058734 on Build GRCh38
rs1057189043 Chr.20: 49058383-49058383 on Build GRCh38
rs927066081 Chr.20: 49058697-49058697 on Build GRCh38
rs752585427 Chr.20: 49058469-49058469 on Build GRCh38
rs902587509 Chr.20: 49058775-49058775 on Build GRCh38
rs763201685 Chr.20: 49058582-49058582 on Build GRCh38
rs533303897 Chr.20: 49058749-49058749 on Build GRCh38
rs533925713 Chr.20: 49058750-49058750 on Build GRCh38
rs937542227 Chr.20: 49058373-49058373 on Build GRCh38
rs1008741552 Chr.20: 49058531-49058531 on Build GRCh38
rs747502351 Chr.20: 49058540-49058540 on Build GRCh38
rs147405039 Chr.20: 49058437-49058437 on Build GRCh38
rs756726793 Chr.20: 49058502-49058502 on Build GRCh38
rs372551577 Chr.20: 49058476-49058476 on Build GRCh38
rs1056797703 Chr.20: 49058720-49058720 on Build GRCh38
rs771789550 Chr.20: 49058677-49058677 on Build GRCh38
rs747487456 Chr.20: 49058429-49058429 on Build GRCh38
rs1021492677 Chr.20: 49058773-49058773 on Build GRCh38
rs867152620 Chr.20: 49058666-49058666 on Build GRCh38
rs898438998 Chr.20: 49058755-49058755 on Build GRCh38
rs748306421 Chr.20: 49058564-49058564 on Build GRCh38
rs959593374 Chr.20: 49058651-49058651 on Build GRCh38
rs188215367 Chr.20: 49058436-49058436 on Build GRCh38
rs767284976 Chr.20: 49058463-49058463 on Build GRCh38
rs377402001 Chr.20: 49058440-49058440 on Build GRCh38
rs899386045 Chr.20: 49058765-49058765 on Build GRCh38
rs771797256 Chr.20: 49058593-49058593 on Build GRCh38
rs759462614 Chr.20: 49058461-49058461 on Build GRCh38
rs753367257 Chr.20: 49058599-49058599 on Build GRCh38
rs965200625 Chr.20: 49058597-49058597 on Build GRCh38
rs145758236 Chr.20: 49058496-49058496 on Build GRCh38
rs568803842 Chr.20: 49058608-49058608 on Build GRCh38
rs753398302 Chr.20: 49058477-49058477 on Build GRCh38
rs372875852 Chr.20: 49058414-49058414 on Build GRCh38
rs899187216 Chr.20: 49058462-49058462 on Build GRCh38
rs2064853 Chr.20: 49058639-49058639 on Build GRCh38
rs758772975 Chr.20: 49058527-49058527 on Build GRCh38
rs780459783 Chr.20: 49058490-49058490 on Build GRCh38
rs932589398 Chr.20: 49058766-49058766 on Build GRCh38
rs55727893 Chr.20: 49058660-49058660 on Build GRCh38
rs773582029 Chr.20: 49058578-49058578 on Build GRCh38
rs148515114 Chr.20: 49058771-49058771 on Build GRCh38

View Full Product Details