Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 50841663-50842207 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCAS4
Entrez Gene ID
55653
Gene Name
breast carcinoma amplified sequence 4
Gene Aliases
CNOL
UniGene
Hs.381178
Gene Chromosome Location
Chr.20: 50794894-50877736 on Build GRCh38
Transcripts
NM_001010974.2 NM_017843.4 NM_198799.3 XM_011528886.2 XM_011528887.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
127428 c.405G>C p.V135V Chr.20: 50841816-50841816 on Build GRCh38
3547665 c.474C>T p.L158L Chr.20: 50841885-50841885 on Build GRCh38
6200185 c.407T>G p.L136R Chr.20: 50841818-50841818 on Build GRCh38
6427036 c.419G>A p.R140Q Chr.20: 50841830-50841830 on Build GRCh38
3547668 c.480C>T p.S160S Chr.20: 50841891-50841891 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774255921 Chr.20: 50841771-50841771 on Build GRCh38
rs201242829 Chr.20: 50841829-50841829 on Build GRCh38
rs372999758 Chr.20: 50841908-50841908 on Build GRCh38
rs770203707 Chr.20: 50841851-50841851 on Build GRCh38
rs933961471 Chr.20: 50842174-50842174 on Build GRCh38
rs906989771 Chr.20: 50842041-50842041 on Build GRCh38
rs375993366 Chr.20: 50841971-50841971 on Build GRCh38
rs8117696 Chr.20: 50842101-50842101 on Build GRCh38
rs752585676 Chr.20: 50841785-50841785 on Build GRCh38
rs941323177 Chr.20: 50841703-50841703 on Build GRCh38
rs769546070 Chr.20: 50841782-50841782 on Build GRCh38
rs386814936 Chr.20: 50842068-50842069 on Build GRCh38
rs367585023 Chr.20: 50841754-50841754 on Build GRCh38
rs867893000 Chr.20: 50842118-50842118 on Build GRCh38
rs756600384 Chr.20: 50841952-50841952 on Build GRCh38
rs6063570 Chr.20: 50842068-50842068 on Build GRCh38
rs1022360514 Chr.20: 50842094-50842094 on Build GRCh38
rs992427265 Chr.20: 50842023-50842024 on Build GRCh38
rs368879764 Chr.20: 50841830-50841830 on Build GRCh38
rs929490246 Chr.20: 50842180-50842180 on Build GRCh38
rs571438318 Chr.20: 50841875-50841875 on Build GRCh38
rs371565876 Chr.20: 50842037-50842037 on Build GRCh38
rs377159322 Chr.20: 50841909-50841909 on Build GRCh38
rs781464792 Chr.20: 50841836-50841836 on Build GRCh38
rs776804680 Chr.20: 50841755-50841755 on Build GRCh38
rs111708986 Chr.20: 50841876-50841876 on Build GRCh38
rs550628520 Chr.20: 50841877-50841877 on Build GRCh38
rs763173953 Chr.20: 50841857-50841857 on Build GRCh38
rs575839349 Chr.20: 50842025-50842025 on Build GRCh38
rs992378126 Chr.20: 50842126-50842126 on Build GRCh38
rs532334457 Chr.20: 50842076-50842076 on Build GRCh38
rs775855073 Chr.20: 50841738-50841738 on Build GRCh38
rs533797906 Chr.20: 50841929-50841929 on Build GRCh38
rs758020860 Chr.20: 50841888-50841888 on Build GRCh38
rs899891059 Chr.20: 50841776-50841776 on Build GRCh38
rs926847194 Chr.20: 50841994-50841995 on Build GRCh38
rs372574118 Chr.20: 50841747-50841747 on Build GRCh38
rs748308643 Chr.20: 50841901-50841901 on Build GRCh38
rs769023749 Chr.20: 50842083-50842083 on Build GRCh38
rs772692182 Chr.20: 50841737-50841737 on Build GRCh38
rs934468084 Chr.20: 50841999-50841999 on Build GRCh38
rs2145693 Chr.20: 50841890-50841890 on Build GRCh38
rs773306389 Chr.20: 50841853-50841853 on Build GRCh38
rs771780945 Chr.20: 50841859-50841859 on Build GRCh38
rs1051091943 Chr.20: 50841982-50841982 on Build GRCh38
rs764675901 Chr.20: 50841941-50841941 on Build GRCh38
rs546572057 Chr.20: 50841795-50841795 on Build GRCh38
rs771151453 Chr.20: 50841910-50841910 on Build GRCh38
rs901530873 Chr.20: 50842047-50842047 on Build GRCh38
rs750057692 Chr.20: 50841881-50841881 on Build GRCh38
rs142613237 Chr.20: 50841781-50841781 on Build GRCh38
rs755994429 Chr.20: 50841788-50841788 on Build GRCh38
rs116620165 Chr.20: 50842098-50842098 on Build GRCh38
rs201941810 Chr.20: 50841761-50841761 on Build GRCh38
rs542842685 Chr.20: 50842028-50842028 on Build GRCh38
rs375780908 Chr.20: 50841814-50841814 on Build GRCh38
rs191473028 Chr.20: 50842088-50842088 on Build GRCh38
rs951486773 Chr.20: 50841933-50841933 on Build GRCh38
rs554896122 Chr.20: 50841892-50841892 on Build GRCh38
rs1040090394 Chr.20: 50841958-50841958 on Build GRCh38
rs778416220 Chr.20: 50841802-50841802 on Build GRCh38
rs965844636 Chr.20: 50841702-50841702 on Build GRCh38
rs929474189 Chr.20: 50842027-50842027 on Build GRCh38
rs369028398 Chr.20: 50841951-50841951 on Build GRCh38
rs763995627 Chr.20: 50841870-50841870 on Build GRCh38
rs960698031 Chr.20: 50842119-50842119 on Build GRCh38
rs375476882 Chr.20: 50841889-50841890 on Build GRCh38
rs748426468 Chr.20: 50842129-50842129 on Build GRCh38
rs986861522 Chr.20: 50842175-50842175 on Build GRCh38
rs769774324 Chr.20: 50841905-50841905 on Build GRCh38
rs187032416 Chr.20: 50841985-50841985 on Build GRCh38
rs776554277 Chr.20: 50841932-50841932 on Build GRCh38
rs2145692 Chr.20: 50841889-50841889 on Build GRCh38
rs915224056 Chr.20: 50841796-50841796 on Build GRCh38
rs77973389 Chr.20: 50842039-50842039 on Build GRCh38
rs202162521 Chr.20: 50841860-50841860 on Build GRCh38
rs6096118 Chr.20: 50841942-50841942 on Build GRCh38
rs775196324 Chr.20: 50841916-50841916 on Build GRCh38
rs370380983 Chr.20: 50841917-50841917 on Build GRCh38
rs762159407 Chr.20: 50841767-50841767 on Build GRCh38
rs779288945 Chr.20: 50841716-50841716 on Build GRCh38
rs759240930 Chr.20: 50841772-50841772 on Build GRCh38
rs752031069 Chr.20: 50841946-50841946 on Build GRCh38
rs898007337 Chr.20: 50841886-50841886 on Build GRCh38
rs1054398218 Chr.20: 50842001-50842001 on Build GRCh38
rs750554818 Chr.20: 50841963-50841963 on Build GRCh38
rs746196357 Chr.20: 50841728-50841728 on Build GRCh38
rs528740006 Chr.20: 50842045-50842045 on Build GRCh38
rs747485073 Chr.20: 50841744-50841744 on Build GRCh38
rs36084823 Chr.20: 50842107-50842108 on Build GRCh38
rs145066991 Chr.20: 50841685-50841685 on Build GRCh38
rs368180767 Chr.20: 50841813-50841813 on Build GRCh38
rs921267803 Chr.20: 50842148-50842148 on Build GRCh38
rs371799931 Chr.20: 50841793-50841793 on Build GRCh38
rs748218673 Chr.20: 50841841-50841841 on Build GRCh38
rs763623373 Chr.20: 50841792-50841792 on Build GRCh38
rs752057828 Chr.20: 50842038-50842038 on Build GRCh38
rs114935198 Chr.20: 50842069-50842069 on Build GRCh38
rs983329218 Chr.20: 50842095-50842095 on Build GRCh38
rs369934786 Chr.20: 50841934-50841934 on Build GRCh38
rs200416545 Chr.20: 50841903-50841903 on Build GRCh38
rs2145694 Chr.20: 50841947-50841947 on Build GRCh38
rs909886409 Chr.20: 50841690-50841690 on Build GRCh38
rs566645256 Chr.20: 50841900-50841900 on Build GRCh38
rs934076030 Chr.20: 50841974-50841974 on Build GRCh38

View Full Product Details