Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 54162564-54163064 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP24A1
Entrez Gene ID
1591
Gene Name
cytochrome P450 family 24 subfamily A member 1
Gene Aliases
CP24, CYP24, HCAI, HCINF1, P450-CC24
UniGene
Hs.89663
Gene Chromosome Location
Chr.20: 54145731-54173985 on Build GRCh38
Transcripts
NM_000782.4 NM_001128915.1 XM_005260304.4 XM_017027691.1 XM_017027692.1 XM_017027693.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1291133 c.945A>C p.E315D Chr.20: 54162762-54162762 on Build GRCh38
4675895 c.865C>T p.R289W Chr.20: 54162842-54162842 on Build GRCh38
4675894 c.953C>T p.A318V Chr.20: 54162754-54162754 on Build GRCh38
5529491 c.990+1G>A p.? Chr.20: 54162716-54162716 on Build GRCh38
7177132 c.856A>C p.I286L Chr.20: 54162851-54162851 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1015246623 Chr.20: 54162619-54162619 on Build GRCh38
rs929027030 Chr.20: 54162861-54162861 on Build GRCh38
rs763631953 Chr.20: 54162865-54162865 on Build GRCh38
rs886056788 Chr.20: 54162846-54162846 on Build GRCh38
rs370297190 Chr.20: 54162756-54162756 on Build GRCh38
rs768841085 Chr.20: 54162822-54162822 on Build GRCh38
rs908976061 Chr.20: 54162982-54162982 on Build GRCh38
rs753379986 Chr.20: 54162868-54162868 on Build GRCh38
rs760822781 Chr.20: 54162706-54162706 on Build GRCh38
rs541392563 Chr.20: 54162603-54162603 on Build GRCh38
rs758868434 Chr.20: 54162909-54162909 on Build GRCh38
rs779209362 Chr.20: 54162777-54162777 on Build GRCh38
rs562355310 Chr.20: 54162701-54162701 on Build GRCh38
rs1036721674 Chr.20: 54162798-54162798 on Build GRCh38
rs767209123 Chr.20: 54162730-54162730 on Build GRCh38
rs749784738 Chr.20: 54162848-54162849 on Build GRCh38
rs953583839 Chr.20: 54162656-54162656 on Build GRCh38
rs778754341 Chr.20: 54162864-54162864 on Build GRCh38
rs746266995 Chr.20: 54162778-54162778 on Build GRCh38
rs926504558 Chr.20: 54162722-54162722 on Build GRCh38
rs1047915718 Chr.20: 54162943-54162943 on Build GRCh38
rs765747500 Chr.20: 54162888-54162888 on Build GRCh38
rs758491296 Chr.20: 54162779-54162779 on Build GRCh38
rs757588339 Chr.20: 54162773-54162773 on Build GRCh38
rs58658819 Chr.20: 54162636-54162636 on Build GRCh38
rs966201029 Chr.20: 54163000-54163000 on Build GRCh38
rs184453665 Chr.20: 54162750-54162750 on Build GRCh38
rs1028306385 Chr.20: 54162647-54162647 on Build GRCh38
rs561627955 Chr.20: 54162716-54162716 on Build GRCh38
rs749195896 Chr.20: 54162841-54162841 on Build GRCh38
rs769114522 Chr.20: 54162857-54162858 on Build GRCh38
rs201825042 Chr.20: 54162849-54162849 on Build GRCh38
rs76747058 Chr.20: 54162799-54162799 on Build GRCh38
rs371873677 Chr.20: 54162826-54162826 on Build GRCh38
rs1019160610 Chr.20: 54162999-54162999 on Build GRCh38
rs946592997 Chr.20: 54162663-54162663 on Build GRCh38
rs973703361 Chr.20: 54162633-54162633 on Build GRCh38
rs929056052 Chr.20: 54162693-54162693 on Build GRCh38
rs866441419 Chr.20: 54162851-54162851 on Build GRCh38
rs147022974 Chr.20: 54162786-54162786 on Build GRCh38
rs757714766 Chr.20: 54162887-54162887 on Build GRCh38
rs986678211 Chr.20: 54162669-54162669 on Build GRCh38
rs200624428 Chr.20: 54162856-54162856 on Build GRCh38
rs572368697 Chr.20: 54162596-54162596 on Build GRCh38
rs1029723770 Chr.20: 54162590-54162590 on Build GRCh38
rs387907324 Chr.20: 54162743-54162743 on Build GRCh38
rs6097816 Chr.20: 54162673-54162673 on Build GRCh38
rs774227656 Chr.20: 54162842-54162842 on Build GRCh38
rs749648570 Chr.20: 54162675-54162675 on Build GRCh38
rs6091828 Chr.20: 54162671-54162671 on Build GRCh38
rs955438162 Chr.20: 54162753-54162753 on Build GRCh38
rs775384646 Chr.20: 54162704-54162704 on Build GRCh38
rs979301542 Chr.20: 54162667-54162667 on Build GRCh38
rs552310427 Chr.20: 54162718-54162718 on Build GRCh38
rs373314547 Chr.20: 54162695-54162695 on Build GRCh38
rs368404770 Chr.20: 54162759-54162759 on Build GRCh38
rs761930941 Chr.20: 54162872-54162872 on Build GRCh38
rs386815206 Chr.20: 54162671-54162673 on Build GRCh38
rs756018085 Chr.20: 54162678-54162678 on Build GRCh38
rs762830804 Chr.20: 54162714-54162714 on Build GRCh38
rs6022993 Chr.20: 54162708-54162708 on Build GRCh38
rs773353239 Chr.20: 54162711-54162711 on Build GRCh38
rs1056187605 Chr.20: 54162740-54162740 on Build GRCh38
rs756296470 Chr.20: 54162794-54162794 on Build GRCh38
rs955646930 Chr.20: 54162598-54162598 on Build GRCh38
rs896196218 Chr.20: 54162686-54162686 on Build GRCh38
rs772125898 Chr.20: 54162882-54162883 on Build GRCh38
rs752080791 Chr.20: 54162734-54162734 on Build GRCh38
rs771340916 Chr.20: 54162681-54162681 on Build GRCh38
rs1007738059 Chr.20: 54162998-54162998 on Build GRCh38
rs531371253 Chr.20: 54162703-54162703 on Build GRCh38
rs750687191 Chr.20: 54162893-54162893 on Build GRCh38
rs764570392 Chr.20: 54162748-54162748 on Build GRCh38
rs375976660 Chr.20: 54162848-54162848 on Build GRCh38
rs962688665 Chr.20: 54162627-54162627 on Build GRCh38
rs745483961 Chr.20: 54162965-54162965 on Build GRCh38
rs774812969 Chr.20: 54162863-54162864 on Build GRCh38
rs749064329 Chr.20: 54162726-54162726 on Build GRCh38

View Full Product Details