Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 57174840-57175341 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BMP7
Entrez Gene ID
655
Gene Name
bone morphogenetic protein 7
Gene Aliases
OP-1
UniGene
Hs.473163
Gene Chromosome Location
Chr.20: 57168753-57266651 on Build GRCh38
Transcripts
NM_001719.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4407855 c.1019G>A p.R340Q Chr.20: 57174947-57174947 on Build GRCh38
1713710 c.973G>A p.D325N Chr.20: 57174993-57174993 on Build GRCh38
6415903 c.962A>G p.N321S Chr.20: 57175004-57175004 on Build GRCh38
1751514 c.1011C>T p.V337V Chr.20: 57174955-57174955 on Build GRCh38
4773891 c.959-7A>C p.? Chr.20: 57175014-57175014 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs112630476 Chr.20: 57174875-57174875 on Build GRCh38
rs994123101 Chr.20: 57175170-57175170 on Build GRCh38
rs761532385 Chr.20: 57174947-57174947 on Build GRCh38
rs762810018 Chr.20: 57174881-57174882 on Build GRCh38
rs199953994 Chr.20: 57175032-57175032 on Build GRCh38
rs987758965 Chr.20: 57175164-57175164 on Build GRCh38
rs200825690 Chr.20: 57174993-57174993 on Build GRCh38
rs373485547 Chr.20: 57174887-57174887 on Build GRCh38
rs185828910 Chr.20: 57175155-57175155 on Build GRCh38
rs989066587 Chr.20: 57175101-57175101 on Build GRCh38
rs771357020 Chr.20: 57175153-57175153 on Build GRCh38
rs563837208 Chr.20: 57175069-57175069 on Build GRCh38
rs574025413 Chr.20: 57175118-57175118 on Build GRCh38
rs201801316 Chr.20: 57174994-57174994 on Build GRCh38
rs530565073 Chr.20: 57175187-57175187 on Build GRCh38
rs994051225 Chr.20: 57174980-57174980 on Build GRCh38
rs1000623313 Chr.20: 57175269-57175269 on Build GRCh38
rs772689429 Chr.20: 57174966-57174966 on Build GRCh38
rs969603385 Chr.20: 57175247-57175247 on Build GRCh38
rs1035957339 Chr.20: 57175048-57175051 on Build GRCh38
rs546249439 Chr.20: 57175259-57175259 on Build GRCh38
rs559576947 Chr.20: 57175138-57175138 on Build GRCh38
rs779093350 Chr.20: 57174898-57174898 on Build GRCh38
rs61733438 Chr.20: 57175004-57175004 on Build GRCh38
rs757552780 Chr.20: 57175029-57175029 on Build GRCh38
rs751126578 Chr.20: 57174990-57174990 on Build GRCh38
rs955918110 Chr.20: 57175160-57175160 on Build GRCh38
rs895772857 Chr.20: 57175062-57175062 on Build GRCh38
rs551625986 Chr.20: 57175158-57175158 on Build GRCh38
rs764632412 Chr.20: 57175025-57175025 on Build GRCh38
rs1051253447 Chr.20: 57175121-57175121 on Build GRCh38
rs760087726 Chr.20: 57175300-57175300 on Build GRCh38
rs61733437 Chr.20: 57174967-57174967 on Build GRCh38
rs565001021 Chr.20: 57175186-57175186 on Build GRCh38
rs754520569 Chr.20: 57174981-57174981 on Build GRCh38
rs2182435 Chr.20: 57175148-57175148 on Build GRCh38
rs987802111 Chr.20: 57175299-57175299 on Build GRCh38
rs879812703 Chr.20: 57175184-57175184 on Build GRCh38
rs375215962 Chr.20: 57175103-57175103 on Build GRCh38
rs1057495281 Chr.20: 57175270-57175270 on Build GRCh38
rs769591631 Chr.20: 57174948-57174948 on Build GRCh38
rs1023481282 Chr.20: 57175241-57175241 on Build GRCh38
rs770879190 Chr.20: 57174897-57174897 on Build GRCh38
rs920330115 Chr.20: 57175213-57175213 on Build GRCh38
rs753980150 Chr.20: 57175020-57175020 on Build GRCh38
rs75877339 Chr.20: 57175205-57175205 on Build GRCh38
rs898958752 Chr.20: 57175012-57175012 on Build GRCh38
rs572448202 Chr.20: 57174931-57174931 on Build GRCh38
rs182913460 Chr.20: 57175123-57175123 on Build GRCh38
rs773568800 Chr.20: 57175074-57175074 on Build GRCh38
rs1051658305 Chr.20: 57174866-57174866 on Build GRCh38
rs1032059747 Chr.20: 57175276-57175276 on Build GRCh38
rs754824184 Chr.20: 57175294-57175294 on Build GRCh38
rs113035312 Chr.20: 57175045-57175045 on Build GRCh38
rs778234407 Chr.20: 57175017-57175017 on Build GRCh38
rs913477369 Chr.20: 57175110-57175110 on Build GRCh38
rs376331503 Chr.20: 57175044-57175044 on Build GRCh38
rs114761082 Chr.20: 57174925-57174925 on Build GRCh38
rs529732230 Chr.20: 57175233-57175233 on Build GRCh38
rs887243725 Chr.20: 57175266-57175266 on Build GRCh38
rs372806147 Chr.20: 57174914-57174914 on Build GRCh38
rs113547893 Chr.20: 57175043-57175043 on Build GRCh38
rs369886884 Chr.20: 57174896-57174896 on Build GRCh38
rs565934733 Chr.20: 57175277-57175277 on Build GRCh38
rs768638308 Chr.20: 57174915-57174915 on Build GRCh38
rs1013363794 Chr.20: 57175018-57175018 on Build GRCh38
rs550285431 Chr.20: 57175188-57175188 on Build GRCh38
rs756501676 Chr.20: 57175015-57175015 on Build GRCh38

View Full Product Details