Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 57265623-57266123 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BMP7
Entrez Gene ID
655
Gene Name
bone morphogenetic protein 7
Gene Aliases
OP-1
UniGene
Hs.473163
Gene Chromosome Location
Chr.20: 57168753-57266651 on Build GRCh38
Transcripts
NM_001719.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5580347 c.317C>T p.P106L Chr.20: 57265806-57265806 on Build GRCh38
3953079 c.162G>C p.M54I Chr.20: 57265961-57265961 on Build GRCh38
4651415 c.165G>A p.Q55Q Chr.20: 57265958-57265958 on Build GRCh38
6396706 c.235G>A p.A79T Chr.20: 57265888-57265888 on Build GRCh38
7357598 c.314A>G p.Y105C Chr.20: 57265809-57265809 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs925783954 Chr.20: 57265829-57265829 on Build GRCh38
rs149276258 Chr.20: 57265693-57265693 on Build GRCh38
rs924570866 Chr.20: 57265771-57265771 on Build GRCh38
rs758480033 Chr.20: 57265755-57265755 on Build GRCh38
rs1021408170 Chr.20: 57266087-57266087 on Build GRCh38
rs765378952 Chr.20: 57265846-57265846 on Build GRCh38
rs1005812409 Chr.20: 57265688-57265688 on Build GRCh38
rs769113411 Chr.20: 57265919-57265919 on Build GRCh38
rs931743425 Chr.20: 57265893-57265893 on Build GRCh38
rs753143487 Chr.20: 57265810-57265812 on Build GRCh38
rs763198817 Chr.20: 57265973-57265973 on Build GRCh38
rs866060383 Chr.20: 57265758-57265758 on Build GRCh38
rs776026323 Chr.20: 57265826-57265826 on Build GRCh38
rs377359930 Chr.20: 57265723-57265723 on Build GRCh38
rs961649397 Chr.20: 57265709-57265709 on Build GRCh38
rs868564328 Chr.20: 57266056-57266056 on Build GRCh38
rs1016222909 Chr.20: 57265699-57265702 on Build GRCh38
rs6099519 Chr.20: 57265817-57265817 on Build GRCh38
rs940219541 Chr.20: 57265858-57265858 on Build GRCh38
rs900152094 Chr.20: 57265890-57265890 on Build GRCh38
rs745308771 Chr.20: 57265811-57265811 on Build GRCh38
rs778560295 Chr.20: 57265807-57265807 on Build GRCh38
rs559666513 Chr.20: 57265841-57265841 on Build GRCh38
rs774351201 Chr.20: 57266007-57266007 on Build GRCh38
rs781124338 Chr.20: 57265756-57265756 on Build GRCh38
rs777610187 Chr.20: 57265868-57265868 on Build GRCh38
rs768202871 Chr.20: 57265680-57265680 on Build GRCh38
rs777202520 Chr.20: 57265955-57265955 on Build GRCh38
rs757079660 Chr.20: 57266075-57266075 on Build GRCh38
rs766457017 Chr.20: 57265976-57265976 on Build GRCh38
rs770163557 Chr.20: 57265970-57265970 on Build GRCh38
rs747542314 Chr.20: 57265917-57265917 on Build GRCh38
rs371423711 Chr.20: 57265695-57265695 on Build GRCh38
rs754059991 Chr.20: 57265706-57265706 on Build GRCh38
rs779479867 Chr.20: 57265910-57265910 on Build GRCh38
rs867354171 Chr.20: 57265914-57265914 on Build GRCh38
rs774053418 Chr.20: 57265668-57265668 on Build GRCh38
rs921311051 Chr.20: 57265778-57265778 on Build GRCh38
rs980184645 Chr.20: 57266057-57266057 on Build GRCh38
rs868147897 Chr.20: 57265987-57265987 on Build GRCh38
rs376083170 Chr.20: 57265853-57265853 on Build GRCh38
rs112053109 Chr.20: 57265704-57265704 on Build GRCh38
rs755895161 Chr.20: 57265789-57265789 on Build GRCh38
rs370363790 Chr.20: 57265735-57265735 on Build GRCh38
rs766396823 Chr.20: 57265860-57265860 on Build GRCh38
rs765291185 Chr.20: 57265728-57265728 on Build GRCh38
rs748948590 Chr.20: 57265792-57265792 on Build GRCh38
rs779457752 Chr.20: 57265721-57265726 on Build GRCh38
rs888626278 Chr.20: 57265963-57265963 on Build GRCh38
rs775529228 Chr.20: 57265866-57265866 on Build GRCh38
rs770533058 Chr.20: 57265664-57265664 on Build GRCh38
rs770763971 Chr.20: 57265798-57265798 on Build GRCh38
rs970017958 Chr.20: 57266053-57266053 on Build GRCh38
rs776921476 Chr.20: 57265843-57265843 on Build GRCh38
rs745548111 Chr.20: 57265894-57265894 on Build GRCh38
rs771580937 Chr.20: 57265895-57265895 on Build GRCh38
rs144422198 Chr.20: 57265710-57265710 on Build GRCh38
rs777464786 Chr.20: 57265790-57265790 on Build GRCh38
rs748919336 Chr.20: 57265657-57265657 on Build GRCh38
rs367800755 Chr.20: 57265865-57265865 on Build GRCh38
rs897430051 Chr.20: 57266006-57266006 on Build GRCh38
rs758852938 Chr.20: 57265739-57265739 on Build GRCh38
rs755317863 Chr.20: 57265679-57265680 on Build GRCh38
rs775877188 Chr.20: 57265683-57265683 on Build GRCh38
rs752595818 Chr.20: 57265775-57265775 on Build GRCh38
rs762109775 Chr.20: 57265958-57265958 on Build GRCh38
rs760656364 Chr.20: 57266017-57266017 on Build GRCh38
rs750655217 Chr.20: 57265850-57265850 on Build GRCh38
rs761126812 Chr.20: 57265832-57265832 on Build GRCh38
rs763868573 Chr.20: 57266021-57266021 on Build GRCh38
rs368156920 Chr.20: 57265679-57265679 on Build GRCh38
rs771732616 Chr.20: 57265821-57265821 on Build GRCh38
rs976788767 Chr.20: 57265777-57265777 on Build GRCh38
rs865925053 Chr.20: 57266062-57266062 on Build GRCh38
rs371542500 Chr.20: 57265889-57265889 on Build GRCh38
rs536220019 Chr.20: 57265962-57265962 on Build GRCh38
rs767991677 Chr.20: 57265738-57265738 on Build GRCh38
rs199724512 Chr.20: 57266027-57266027 on Build GRCh38
rs61733434 Chr.20: 57265844-57265844 on Build GRCh38
rs753750823 Chr.20: 57265888-57265888 on Build GRCh38
rs1048768000 Chr.20: 57265916-57265916 on Build GRCh38

View Full Product Details