Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 58839611-58840106 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNAS
Entrez Gene ID
2778
Gene Name
GNAS complex locus
Gene Aliases
AHO, C20orf45, GNAS1, GPSA, GSA, GSP, NESP, POH, SCG6, SgVI
UniGene
Hs.125898
Gene Chromosome Location
Chr.20: 58839681-58911196 on Build GRCh38
Transcripts
NM_016592.3 XM_017027821.1 XM_017027822.1 NR_002785.2

Gene Symbol
GNAS-AS1
Entrez Gene ID
149775
Gene Name
GNAS antisense RNA 1
Gene Aliases
GNAS-AS, GNAS1AS, GNASAS, NCRNA00075, NESP-AS, NESPAS, SANG
UniGene
Hs.122718
Gene Chromosome Location
Chr.20: 58818918-58850903 on Build GRCh38
Transcripts
NM_016592.3 XM_017027821.1 XM_017027822.1 NR_002785.2

SNP Detail

SNP ID Chromosome Location
rs1056143210 Chr.20: 58839745-58839745 on Build GRCh38
rs1006953165 Chr.20: 58839974-58839974 on Build GRCh38
rs1036645642 Chr.20: 58840015-58840015 on Build GRCh38
rs918794407 Chr.20: 58839997-58839997 on Build GRCh38
rs951616510 Chr.20: 58840004-58840004 on Build GRCh38
rs1052821754 Chr.20: 58839849-58839849 on Build GRCh38
rs1029632865 Chr.20: 58839896-58839896 on Build GRCh38
rs1044894538 Chr.20: 58839799-58839799 on Build GRCh38
rs955464674 Chr.20: 58839690-58839690 on Build GRCh38
rs1018431016 Chr.20: 58839975-58839975 on Build GRCh38
rs532186771 Chr.20: 58840052-58840052 on Build GRCh38
rs896307016 Chr.20: 58839635-58839635 on Build GRCh38
rs369024311 Chr.20: 58839797-58839797 on Build GRCh38
rs537345739 Chr.20: 58839760-58839760 on Build GRCh38
rs748024920 Chr.20: 58840024-58840024 on Build GRCh38
rs1042166761 Chr.20: 58839662-58839662 on Build GRCh38
rs947775276 Chr.20: 58840045-58840045 on Build GRCh38
rs945731825 Chr.20: 58839865-58839865 on Build GRCh38
rs544068079 Chr.20: 58839990-58839990 on Build GRCh38
rs370380052 Chr.20: 58839893-58839893 on Build GRCh38
rs985564902 Chr.20: 58839695-58839695 on Build GRCh38
rs974758829 Chr.20: 58839721-58839721 on Build GRCh38
rs991793016 Chr.20: 58840025-58840025 on Build GRCh38
rs956271320 Chr.20: 58839632-58839632 on Build GRCh38
rs922895264 Chr.20: 58839959-58839961 on Build GRCh38
rs1006783741 Chr.20: 58840008-58840008 on Build GRCh38
rs929666005 Chr.20: 58839740-58839740 on Build GRCh38
rs911399131 Chr.20: 58839707-58839707 on Build GRCh38
rs867648879 Chr.20: 58839655-58839655 on Build GRCh38
rs902223345 Chr.20: 58839670-58839670 on Build GRCh38
rs892386658 Chr.20: 58839863-58839863 on Build GRCh38
rs918250739 Chr.20: 58839730-58839730 on Build GRCh38
rs768408948 Chr.20: 58840063-58840063 on Build GRCh38
rs917606178 Chr.20: 58840044-58840044 on Build GRCh38
rs61558260 Chr.20: 58839812-58839812 on Build GRCh38
rs913132536 Chr.20: 58839859-58839859 on Build GRCh38
rs554584733 Chr.20: 58839645-58839645 on Build GRCh38
rs531607158 Chr.20: 58840040-58840040 on Build GRCh38
rs1040062014 Chr.20: 58839874-58839875 on Build GRCh38
rs1800900 Chr.20: 58840055-58840055 on Build GRCh38
rs768318168 Chr.20: 58840077-58840078 on Build GRCh38
rs577379485 Chr.20: 58839835-58839835 on Build GRCh38
rs553878748 Chr.20: 58839989-58839989 on Build GRCh38
rs541643892 Chr.20: 58839970-58839970 on Build GRCh38
rs151179443 Chr.20: 58839992-58839992 on Build GRCh38
rs560873163 Chr.20: 58840011-58840011 on Build GRCh38
rs917637308 Chr.20: 58839748-58839748 on Build GRCh38
rs1006487792 Chr.20: 58839706-58839706 on Build GRCh38
rs1010160378 Chr.20: 58839883-58839883 on Build GRCh38
rs140203361 Chr.20: 58840050-58840050 on Build GRCh38
rs1050620349 Chr.20: 58839688-58839688 on Build GRCh38
rs996387771 Chr.20: 58839667-58839667 on Build GRCh38
rs949196195 Chr.20: 58839659-58839659 on Build GRCh38
rs1013527043 Chr.20: 58839731-58839731 on Build GRCh38
rs962974464 Chr.20: 58839708-58839708 on Build GRCh38
rs931758983 Chr.20: 58839968-58839968 on Build GRCh38
rs1015569363 Chr.20: 58839723-58839723 on Build GRCh38
rs1029664100 Chr.20: 58839687-58839687 on Build GRCh38
rs569974334 Chr.20: 58839677-58839677 on Build GRCh38
rs761171827 Chr.20: 58840078-58840078 on Build GRCh38

View Full Product Details