Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 58839995-58840485 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNAS
Entrez Gene ID
2778
Gene Name
GNAS complex locus
Gene Aliases
AHO, C20orf45, GNAS1, GPSA, GSA, GSP, NESP, POH, SCG6, SgVI
UniGene
Hs.125898
Gene Chromosome Location
Chr.20: 58839681-58911196 on Build GRCh38
Transcripts
NM_016592.3 XM_017027821.1 XM_017027822.1 NR_002785.2

Gene Symbol
GNAS-AS1
Entrez Gene ID
149775
Gene Name
GNAS antisense RNA 1
Gene Aliases
GNAS-AS, GNAS1AS, GNASAS, NCRNA00075, NESP-AS, NESPAS, SANG
UniGene
Hs.122718
Gene Chromosome Location
Chr.20: 58818918-58850903 on Build GRCh38
Transcripts
NM_016592.3 XM_017027821.1 XM_017027822.1 NR_002785.2

SNP Detail

SNP ID Chromosome Location
rs975964450 Chr.20: 58840216-58840216 on Build GRCh38
rs747367482 Chr.20: 58840445-58840445 on Build GRCh38
rs753525197 Chr.20: 58840418-58840418 on Build GRCh38
rs780943650 Chr.20: 58840163-58840163 on Build GRCh38
rs951583745 Chr.20: 58840370-58840370 on Build GRCh38
rs760919160 Chr.20: 58840210-58840210 on Build GRCh38
rs778940007 Chr.20: 58840144-58840144 on Build GRCh38
rs767745741 Chr.20: 58840196-58840196 on Build GRCh38
rs770356690 Chr.20: 58840094-58840094 on Build GRCh38
rs1800901 Chr.20: 58840289-58840289 on Build GRCh38
rs532186771 Chr.20: 58840052-58840052 on Build GRCh38
rs762096209 Chr.20: 58840371-58840371 on Build GRCh38
rs558969394 Chr.20: 58840223-58840223 on Build GRCh38
rs947775276 Chr.20: 58840045-58840045 on Build GRCh38
rs563844600 Chr.20: 58840372-58840372 on Build GRCh38
rs772759887 Chr.20: 58840169-58840169 on Build GRCh38
rs761688848 Chr.20: 58840109-58840110 on Build GRCh38
rs543435005 Chr.20: 58840437-58840437 on Build GRCh38
rs764128447 Chr.20: 58840214-58840214 on Build GRCh38
rs777423607 Chr.20: 58840299-58840299 on Build GRCh38
rs991793016 Chr.20: 58840025-58840025 on Build GRCh38
rs777218004 Chr.20: 58840239-58840239 on Build GRCh38
rs565827536 Chr.20: 58840101-58840101 on Build GRCh38
rs746422746 Chr.20: 58840328-58840328 on Build GRCh38
rs144308406 Chr.20: 58840116-58840116 on Build GRCh38
rs772553938 Chr.20: 58840339-58840339 on Build GRCh38
rs575933995 Chr.20: 58840433-58840433 on Build GRCh38
rs146532795 Chr.20: 58840244-58840244 on Build GRCh38
rs969325846 Chr.20: 58840096-58840096 on Build GRCh38
rs373276011 Chr.20: 58840415-58840415 on Build GRCh38
rs747808766 Chr.20: 58840235-58840235 on Build GRCh38
rs768408948 Chr.20: 58840063-58840063 on Build GRCh38
rs201307445 Chr.20: 58840440-58840440 on Build GRCh38
rs760197051 Chr.20: 58840432-58840433 on Build GRCh38
rs766199286 Chr.20: 58840445-58840480 on Build GRCh38
rs569736160 Chr.20: 58840174-58840174 on Build GRCh38
rs531607158 Chr.20: 58840040-58840040 on Build GRCh38
rs752452409 Chr.20: 58840410-58840410 on Build GRCh38
rs908942214 Chr.20: 58840295-58840298 on Build GRCh38
rs1800900 Chr.20: 58840055-58840055 on Build GRCh38
rs768318168 Chr.20: 58840077-58840078 on Build GRCh38
rs367861404 Chr.20: 58840322-58840322 on Build GRCh38
rs898582390 Chr.20: 58840320-58840320 on Build GRCh38
rs587778388 Chr.20: 58840438-58840438 on Build GRCh38
rs750367565 Chr.20: 58840146-58840146 on Build GRCh38
rs754780507 Chr.20: 58840234-58840234 on Build GRCh38
rs781674491 Chr.20: 58840451-58840451 on Build GRCh38
rs557714686 Chr.20: 58840242-58840242 on Build GRCh38
rs758474465 Chr.20: 58840229-58840229 on Build GRCh38
rs761171827 Chr.20: 58840078-58840078 on Build GRCh38
rs776589538 Chr.20: 58840260-58840260 on Build GRCh38
rs759617960 Chr.20: 58840392-58840392 on Build GRCh38
rs755781835 Chr.20: 58840237-58840237 on Build GRCh38
rs141238454 Chr.20: 58840273-58840273 on Build GRCh38
rs767657815 Chr.20: 58840120-58840120 on Build GRCh38
rs769104795 Chr.20: 58840446-58840446 on Build GRCh38
rs774788451 Chr.20: 58840247-58840247 on Build GRCh38
rs759833927 Chr.20: 58840188-58840188 on Build GRCh38
rs755729283 Chr.20: 58840290-58840290 on Build GRCh38
rs967292772 Chr.20: 58840182-58840182 on Build GRCh38
rs746736450 Chr.20: 58840161-58840161 on Build GRCh38
rs370175837 Chr.20: 58840232-58840232 on Build GRCh38
rs775901436 Chr.20: 58840257-58840257 on Build GRCh38
rs369511579 Chr.20: 58840369-58840369 on Build GRCh38
rs767440362 Chr.20: 58840111-58840112 on Build GRCh38
rs754947062 Chr.20: 58840272-58840272 on Build GRCh38
rs761937008 Chr.20: 58840270-58840270 on Build GRCh38
rs587778387 Chr.20: 58840451-58840462 on Build GRCh38
rs748024920 Chr.20: 58840024-58840024 on Build GRCh38
rs187415363 Chr.20: 58840311-58840311 on Build GRCh38
rs755419386 Chr.20: 58840456-58840485 on Build GRCh38
rs780051524 Chr.20: 58840149-58840149 on Build GRCh38
rs779115304 Chr.20: 58840458-58840459 on Build GRCh38
rs202131370 Chr.20: 58840113-58840113 on Build GRCh38
rs942246524 Chr.20: 58840365-58840365 on Build GRCh38
rs202166182 Chr.20: 58840352-58840352 on Build GRCh38
rs144845382 Chr.20: 58840382-58840382 on Build GRCh38
rs774390751 Chr.20: 58840103-58840105 on Build GRCh38
rs375054629 Chr.20: 58840364-58840364 on Build GRCh38
rs747330417 Chr.20: 58840356-58840356 on Build GRCh38
rs148910090 Chr.20: 58840427-58840427 on Build GRCh38
rs747725720 Chr.20: 58840164-58840164 on Build GRCh38
rs752831442 Chr.20: 58840141-58840141 on Build GRCh38
rs374513507 Chr.20: 58840106-58840106 on Build GRCh38
rs375337259 Chr.20: 58840269-58840269 on Build GRCh38
rs139302910 Chr.20: 58840114-58840114 on Build GRCh38
rs917606178 Chr.20: 58840044-58840044 on Build GRCh38
rs764371346 Chr.20: 58840258-58840258 on Build GRCh38
rs752818515 Chr.20: 58840202-58840202 on Build GRCh38
rs756711953 Chr.20: 58840312-58840312 on Build GRCh38
rs748443274 Chr.20: 58840457-58840457 on Build GRCh38
rs772464689 Chr.20: 58840253-58840253 on Build GRCh38
rs774700482 Chr.20: 58840183-58840183 on Build GRCh38
rs112532266 Chr.20: 58840230-58840230 on Build GRCh38
rs756880829 Chr.20: 58840426-58840426 on Build GRCh38
rs1800902 Chr.20: 58840400-58840400 on Build GRCh38
rs199948732 Chr.20: 58840252-58840252 on Build GRCh38
rs773416371 Chr.20: 58840386-58840386 on Build GRCh38
rs548132075 Chr.20: 58840147-58840147 on Build GRCh38
rs1002304696 Chr.20: 58840111-58840111 on Build GRCh38
rs140203361 Chr.20: 58840050-58840050 on Build GRCh38
rs537714939 Chr.20: 58840177-58840177 on Build GRCh38
rs749968330 Chr.20: 58840342-58840342 on Build GRCh38
rs200003575 Chr.20: 58840306-58840306 on Build GRCh38

View Full Product Details