Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 58840764-58841254 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNAS
Entrez Gene ID
2778
Gene Name
GNAS complex locus
Gene Aliases
AHO, C20orf45, GNAS1, GPSA, GSA, GSP, NESP, POH, SCG6, SgVI
UniGene
Hs.125898
Gene Chromosome Location
Chr.20: 58839681-58911196 on Build GRCh38
Transcripts
NM_016592.3 XM_017027815.1 XM_017027821.1 XM_017027822.1 NR_002785.2

Gene Symbol
GNAS-AS1
Entrez Gene ID
149775
Gene Name
GNAS antisense RNA 1
Gene Aliases
GNAS-AS, GNAS1AS, GNASAS, NCRNA00075, NESP-AS, NESPAS, SANG
UniGene
Hs.122718
Gene Chromosome Location
Chr.20: 58818918-58850903 on Build GRCh38
Transcripts
NM_016592.3 XM_017027815.1 XM_017027821.1 XM_017027822.1 NR_002785.2

SNP Detail

SNP ID Chromosome Location
rs760317470 Chr.20: 58840909-58840909 on Build GRCh38
rs1047219979 Chr.20: 58840990-58840990 on Build GRCh38
rs903043489 Chr.20: 58840904-58840904 on Build GRCh38
rs759229542 Chr.20: 58840784-58840784 on Build GRCh38
rs954468866 Chr.20: 58841125-58841125 on Build GRCh38
rs964966767 Chr.20: 58841151-58841151 on Build GRCh38
rs41305799 Chr.20: 58840912-58840912 on Build GRCh38
rs1032026231 Chr.20: 58840806-58840806 on Build GRCh38
rs780175640 Chr.20: 58840838-58840838 on Build GRCh38
rs887195993 Chr.20: 58841023-58841023 on Build GRCh38
rs971804131 Chr.20: 58841029-58841029 on Build GRCh38
rs539043629 Chr.20: 58840929-58840929 on Build GRCh38
rs1032410869 Chr.20: 58841116-58841116 on Build GRCh38
rs536634729 Chr.20: 58840947-58840947 on Build GRCh38
rs749143784 Chr.20: 58840855-58840855 on Build GRCh38
rs750784274 Chr.20: 58840925-58840925 on Build GRCh38
rs1002484726 Chr.20: 58841108-58841108 on Build GRCh38
rs918987128 Chr.20: 58841030-58841030 on Build GRCh38
rs147123433 Chr.20: 58841039-58841039 on Build GRCh38
rs79709641 Chr.20: 58840786-58840786 on Build GRCh38
rs6100247 Chr.20: 58841137-58841137 on Build GRCh38
rs778743536 Chr.20: 58840867-58840867 on Build GRCh38
rs111921690 Chr.20: 58841138-58841138 on Build GRCh38
rs1800905 Chr.20: 58840940-58840940 on Build GRCh38
rs1800904 Chr.20: 58840907-58840907 on Build GRCh38
rs773279644 Chr.20: 58840795-58840795 on Build GRCh38
rs547189576 Chr.20: 58840854-58840854 on Build GRCh38
rs765578743 Chr.20: 58840923-58840923 on Build GRCh38
rs987236342 Chr.20: 58841131-58841131 on Build GRCh38
rs771753113 Chr.20: 58840796-58840796 on Build GRCh38
rs938067305 Chr.20: 58840785-58840785 on Build GRCh38
rs769227032 Chr.20: 58840880-58840880 on Build GRCh38
rs931294068 Chr.20: 58841165-58841165 on Build GRCh38
rs775141554 Chr.20: 58840870-58840870 on Build GRCh38
rs745570313 Chr.20: 58840869-58840869 on Build GRCh38
rs963047641 Chr.20: 58841020-58841020 on Build GRCh38
rs756061865 Chr.20: 58840852-58840852 on Build GRCh38
rs1040085351 Chr.20: 58840924-58840924 on Build GRCh38
rs761327919 Chr.20: 58840807-58840807 on Build GRCh38
rs80141274 Chr.20: 58840835-58840835 on Build GRCh38
rs775124978 Chr.20: 58840800-58840800 on Build GRCh38
rs373908908 Chr.20: 58841124-58841124 on Build GRCh38
rs112322423 Chr.20: 58840826-58840826 on Build GRCh38
rs957762610 Chr.20: 58841118-58841118 on Build GRCh38
rs201650638 Chr.20: 58840850-58840850 on Build GRCh38
rs1047103815 Chr.20: 58841166-58841166 on Build GRCh38
rs760203169 Chr.20: 58840878-58840878 on Build GRCh38
rs987267791 Chr.20: 58840876-58840876 on Build GRCh38
rs78536121 Chr.20: 58840856-58840856 on Build GRCh38
rs6092699 Chr.20: 58840916-58840916 on Build GRCh38
rs6100246 Chr.20: 58840974-58840974 on Build GRCh38
rs910402426 Chr.20: 58841148-58841148 on Build GRCh38
rs373737048 Chr.20: 58840829-58840829 on Build GRCh38
rs905420938 Chr.20: 58841104-58841104 on Build GRCh38
rs908601226 Chr.20: 58841192-58841192 on Build GRCh38
rs554684203 Chr.20: 58840930-58840930 on Build GRCh38
rs982235893 Chr.20: 58841095-58841095 on Build GRCh38
rs79527543 Chr.20: 58840821-58840821 on Build GRCh38
rs764628148 Chr.20: 58840819-58840819 on Build GRCh38
rs1009728200 Chr.20: 58841120-58841120 on Build GRCh38
rs540948441 Chr.20: 58841157-58841157 on Build GRCh38
rs771945640 Chr.20: 58841152-58841152 on Build GRCh38

View Full Product Details