Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 78004827-78005330 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJB4
Entrez Gene ID
11080
Gene Name
DnaJ heat shock protein family (Hsp40) member B4
Gene Aliases
DNAJW, DjB4, HLJ1
UniGene
Hs.13852
Gene Chromosome Location
Chr.1: 77979158-78017964 on Build GRCh38
Transcripts
NM_001317099.1 NM_001317100.1 NM_001317101.1 NM_001317102.1 NM_001317103.1 NM_007034.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
369219 c.49G>A p.D17N Chr.1: 78005159-78005159 on Build GRCh38
5590638 c.115C>T p.P39S Chr.1: 78005225-78005225 on Build GRCh38
5587658 c.73C>A p.R25R Chr.1: 78005183-78005183 on Build GRCh38
4010010 c.64A>T p.K22* Chr.1: 78005174-78005174 on Build GRCh38
2156303 c.38A>G p.K13R Chr.1: 78005148-78005148 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758021849 Chr.1: 78005194-78005194 on Build GRCh38
rs967403845 Chr.1: 78004878-78004878 on Build GRCh38
rs768785050 Chr.1: 78005263-78005263 on Build GRCh38
rs942421679 Chr.1: 78005283-78005283 on Build GRCh38
rs752215075 Chr.1: 78005170-78005170 on Build GRCh38
rs754621075 Chr.1: 78005162-78005162 on Build GRCh38
rs1010245687 Chr.1: 78005057-78005057 on Build GRCh38
rs759396618 Chr.1: 78005223-78005225 on Build GRCh38
rs553472649 Chr.1: 78004930-78004930 on Build GRCh38
rs746240855 Chr.1: 78005233-78005233 on Build GRCh38
rs769950532 Chr.1: 78005234-78005234 on Build GRCh38
rs942349097 Chr.1: 78004907-78004907 on Build GRCh38
rs957545850 Chr.1: 78005034-78005034 on Build GRCh38
rs1039649696 Chr.1: 78004918-78004918 on Build GRCh38
rs1018963471 Chr.1: 78004871-78004871 on Build GRCh38
rs996669150 Chr.1: 78005272-78005272 on Build GRCh38
rs41309165 Chr.1: 78004950-78004950 on Build GRCh38
rs557127367 Chr.1: 78005255-78005255 on Build GRCh38
rs144880380 Chr.1: 78005151-78005151 on Build GRCh38
rs374712714 Chr.1: 78005284-78005284 on Build GRCh38
rs374281571 Chr.1: 78005128-78005128 on Build GRCh38
rs776438104 Chr.1: 78005296-78005296 on Build GRCh38
rs1001938068 Chr.1: 78005122-78005122 on Build GRCh38
rs753581354 Chr.1: 78005154-78005154 on Build GRCh38
rs767414778 Chr.1: 78005141-78005141 on Build GRCh38
rs145620690 Chr.1: 78005164-78005164 on Build GRCh38
rs759867091 Chr.1: 78005082-78005082 on Build GRCh38
rs909521625 Chr.1: 78004898-78004898 on Build GRCh38
rs111489789 Chr.1: 78005197-78005197 on Build GRCh38
rs756793714 Chr.1: 78004958-78004961 on Build GRCh38
rs766243388 Chr.1: 78005279-78005279 on Build GRCh38
rs929672565 Chr.1: 78004863-78004863 on Build GRCh38
rs771194087 Chr.1: 78005215-78005215 on Build GRCh38
rs148334107 Chr.1: 78005209-78005209 on Build GRCh38
rs545787787 Chr.1: 78005061-78005061 on Build GRCh38
rs949813342 Chr.1: 78005142-78005142 on Build GRCh38
rs1052188512 Chr.1: 78004959-78004959 on Build GRCh38
rs759436082 Chr.1: 78005300-78005300 on Build GRCh38
rs546518950 Chr.1: 78005040-78005040 on Build GRCh38
rs760595061 Chr.1: 78005277-78005277 on Build GRCh38
rs781550817 Chr.1: 78005228-78005228 on Build GRCh38
rs771664003 Chr.1: 78005193-78005193 on Build GRCh38
rs201314004 Chr.1: 78005232-78005232 on Build GRCh38
rs150408892 Chr.1: 78005266-78005266 on Build GRCh38
rs977665808 Chr.1: 78004947-78004947 on Build GRCh38
rs370923954 Chr.1: 78005108-78005108 on Build GRCh38
rs773989896 Chr.1: 78005123-78005124 on Build GRCh38
rs776703958 Chr.1: 78005069-78005069 on Build GRCh38
rs770505305 Chr.1: 78005068-78005069 on Build GRCh38
rs763039133 Chr.1: 78005131-78005131 on Build GRCh38
rs901085328 Chr.1: 78004941-78004941 on Build GRCh38
rs760377085 Chr.1: 78005153-78005153 on Build GRCh38
rs915626751 Chr.1: 78005111-78005111 on Build GRCh38
rs747304458 Chr.1: 78005060-78005060 on Build GRCh38
rs376353018 Chr.1: 78005091-78005091 on Build GRCh38

View Full Product Details