Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 62877642-62878138 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DIDO1
Entrez Gene ID
11083
Gene Name
death inducer-obliterator 1
Gene Aliases
BYE1, C20orf158, DATF-1, DATF1, DIDO2, DIDO3, DIO-1, DIO1, dJ885L7.8
UniGene
Hs.517172
Gene Chromosome Location
Chr.20: 62877738-62937952 on Build GRCh38
Transcripts
NM_001193369.1 NM_033081.2 XM_011528504.1 XM_011528505.1 XM_011528506.1 XM_011528507.1 XM_011528508.1

SNP Detail

SNP ID Chromosome Location
rs1017534169 Chr.20: 62877861-62877861 on Build GRCh38
rs750515589 Chr.20: 62877945-62877945 on Build GRCh38
rs903696850 Chr.20: 62877821-62877821 on Build GRCh38
rs1020835874 Chr.20: 62877682-62877682 on Build GRCh38
rs560974549 Chr.20: 62877683-62877683 on Build GRCh38
rs76041129 Chr.20: 62878020-62878020 on Build GRCh38
rs923389333 Chr.20: 62877679-62877679 on Build GRCh38
rs984049882 Chr.20: 62877696-62877696 on Build GRCh38
rs533631304 Chr.20: 62877706-62877706 on Build GRCh38
rs983569622 Chr.20: 62877996-62877996 on Build GRCh38
rs573417386 Chr.20: 62878056-62878056 on Build GRCh38
rs896255796 Chr.20: 62877799-62877799 on Build GRCh38
rs546181624 Chr.20: 62878033-62878033 on Build GRCh38
rs950838970 Chr.20: 62877952-62877952 on Build GRCh38
rs1049090618 Chr.20: 62877767-62877767 on Build GRCh38
rs180942415 Chr.20: 62877966-62877966 on Build GRCh38
rs542192415 Chr.20: 62877998-62877998 on Build GRCh38
rs938136872 Chr.20: 62877787-62877787 on Build GRCh38
rs758452343 Chr.20: 62877971-62877971 on Build GRCh38
rs919494767 Chr.20: 62877747-62877747 on Build GRCh38
rs917610902 Chr.20: 62878096-62878096 on Build GRCh38
rs1042587873 Chr.20: 62877819-62877819 on Build GRCh38
rs1001176297 Chr.20: 62877927-62877930 on Build GRCh38
rs926254783 Chr.20: 62877731-62877731 on Build GRCh38
rs1019336996 Chr.20: 62878000-62878000 on Build GRCh38
rs1032029287 Chr.20: 62877934-62877934 on Build GRCh38
rs574668305 Chr.20: 62877997-62877997 on Build GRCh38
rs929312775 Chr.20: 62877766-62877766 on Build GRCh38
rs887762274 Chr.20: 62877771-62877771 on Build GRCh38
rs964703561 Chr.20: 62877908-62877908 on Build GRCh38
rs979096444 Chr.20: 62877689-62877689 on Build GRCh38
rs985444884 Chr.20: 62878090-62878090 on Build GRCh38
rs564283968 Chr.20: 62878044-62878044 on Build GRCh38
rs997551475 Chr.20: 62877923-62877923 on Build GRCh38
rs1001155250 Chr.20: 62877822-62877822 on Build GRCh38
rs937719732 Chr.20: 62877762-62877762 on Build GRCh38
rs902398795 Chr.20: 62877849-62877849 on Build GRCh38
rs541312327 Chr.20: 62877678-62877678 on Build GRCh38
rs1032517864 Chr.20: 62877697-62877697 on Build GRCh38
rs985187192 Chr.20: 62877727-62877727 on Build GRCh38
rs909402000 Chr.20: 62877703-62877703 on Build GRCh38
rs761225133 Chr.20: 62878002-62878002 on Build GRCh38
rs1055126994 Chr.20: 62877791-62877798 on Build GRCh38
rs976746699 Chr.20: 62877745-62877745 on Build GRCh38
rs529904056 Chr.20: 62877852-62877852 on Build GRCh38
rs774405162 Chr.20: 62877707-62877707 on Build GRCh38
rs1021307835 Chr.20: 62877828-62877828 on Build GRCh38
rs144289212 Chr.20: 62878028-62878028 on Build GRCh38
rs1030407990 Chr.20: 62877949-62877949 on Build GRCh38
rs750021030 Chr.20: 62878092-62878092 on Build GRCh38
rs937663835 Chr.20: 62878068-62878068 on Build GRCh38
rs1013490823 Chr.20: 62877681-62877681 on Build GRCh38
rs896330933 Chr.20: 62877798-62877798 on Build GRCh38
rs1056136443 Chr.20: 62877768-62877768 on Build GRCh38
rs553194725 Chr.20: 62877800-62877800 on Build GRCh38
rs944824793 Chr.20: 62877732-62877732 on Build GRCh38

View Full Product Details