Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 10454666-10454120 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BAGE2
Entrez Gene ID
85319
Gene Name
B melanoma antigen family member 2
Gene Aliases
CT2.2
UniGene
Hs.545789
Gene Chromosome Location
Chr.21: 10413532-10491615 on Build GRCh38
Transcripts
NM_182482.2

View Full Product Details