Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 29056200-29056714 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCT8
Entrez Gene ID
10694
Gene Name
chaperonin containing TCP1 subunit 8
Gene Aliases
C21orf112, Cctq, D21S246, PRED71
UniGene
Hs.125113
Gene Chromosome Location
Chr.21: 29056322-29073797 on Build GRCh38
Transcripts
NM_001282907.1 NM_001282908.1 NM_001282909.1 NM_006585.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1632442 c.1639A>C p.N547H Chr.21: 29056483-29056483 on Build GRCh38
3841690 c.1633G>C p.D545H Chr.21: 29056489-29056489 on Build GRCh38
4670316 c.1599C>G p.P533P Chr.21: 29056523-29056523 on Build GRCh38
3841691 c.1593T>G p.G531G Chr.21: 29056529-29056529 on Build GRCh38
5813611 c.1640A>G p.N547S Chr.21: 29056482-29056482 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1012558860 Chr.21: 29056646-29056648 on Build GRCh38
rs572283260 Chr.21: 29056513-29056513 on Build GRCh38
rs962268813 Chr.21: 29056464-29056464 on Build GRCh38
rs903673357 Chr.21: 29056687-29056687 on Build GRCh38
rs769805660 Chr.21: 29056313-29056313 on Build GRCh38
rs1038152269 Chr.21: 29056304-29056304 on Build GRCh38
rs914789413 Chr.21: 29056396-29056396 on Build GRCh38
rs758419786 Chr.21: 29056600-29056600 on Build GRCh38
rs896794323 Chr.21: 29056305-29056305 on Build GRCh38
rs896908090 Chr.21: 29056606-29056606 on Build GRCh38
rs767348406 Chr.21: 29056311-29056311 on Build GRCh38
rs563986044 Chr.21: 29056630-29056630 on Build GRCh38
rs972123857 Chr.21: 29056243-29056243 on Build GRCh38
rs191263186 Chr.21: 29056450-29056450 on Build GRCh38
rs763337716 Chr.21: 29056476-29056476 on Build GRCh38
rs757293791 Chr.21: 29056442-29056442 on Build GRCh38
rs929621219 Chr.21: 29056614-29056614 on Build GRCh38
rs1024615143 Chr.21: 29056509-29056509 on Build GRCh38
rs34016221 Chr.21: 29056393-29056394 on Build GRCh38
rs761892081 Chr.21: 29056490-29056490 on Build GRCh38
rs756468808 Chr.21: 29056521-29056521 on Build GRCh38
rs778087735 Chr.21: 29056559-29056561 on Build GRCh38
rs776033779 Chr.21: 29056347-29056347 on Build GRCh38
rs754318740 Chr.21: 29056571-29056571 on Build GRCh38
rs909368329 Chr.21: 29056491-29056491 on Build GRCh38
rs143353810 Chr.21: 29056225-29056225 on Build GRCh38
rs764480542 Chr.21: 29056536-29056536 on Build GRCh38
rs760361197 Chr.21: 29056690-29056691 on Build GRCh38
rs183564802 Chr.21: 29056350-29056350 on Build GRCh38
rs888918572 Chr.21: 29056447-29056447 on Build GRCh38
rs980524538 Chr.21: 29056274-29056274 on Build GRCh38
rs749862733 Chr.21: 29056439-29056439 on Build GRCh38
rs993850404 Chr.21: 29056319-29056319 on Build GRCh38
rs112006909 Chr.21: 29056273-29056273 on Build GRCh38
rs757708879 Chr.21: 29056596-29056596 on Build GRCh38
rs58847852 Chr.21: 29056573-29056573 on Build GRCh38
rs747202852 Chr.21: 29056574-29056575 on Build GRCh38
rs1016124102 Chr.21: 29056454-29056454 on Build GRCh38
rs901859649 Chr.21: 29056435-29056435 on Build GRCh38
rs746759590 Chr.21: 29056451-29056451 on Build GRCh38
rs764569340 Chr.21: 29056641-29056641 on Build GRCh38
rs573233238 Chr.21: 29056387-29056391 on Build GRCh38
rs200555208 Chr.21: 29056444-29056444 on Build GRCh38
rs758362073 Chr.21: 29056500-29056501 on Build GRCh38
rs191712667 Chr.21: 29056266-29056266 on Build GRCh38
rs75832146 Chr.21: 29056415-29056415 on Build GRCh38
rs751231473 Chr.21: 29056519-29056519 on Build GRCh38
rs772739239 Chr.21: 29056503-29056503 on Build GRCh38
rs887993395 Chr.21: 29056287-29056287 on Build GRCh38
rs937526007 Chr.21: 29056331-29056331 on Build GRCh38
rs914779065 Chr.21: 29056656-29056656 on Build GRCh38
rs750450841 Chr.21: 29056598-29056598 on Build GRCh38
rs1034902219 Chr.21: 29056329-29056329 on Build GRCh38
rs941072286 Chr.21: 29056299-29056299 on Build GRCh38
rs186406731 Chr.21: 29056425-29056425 on Build GRCh38
rs371509519 Chr.21: 29056457-29056457 on Build GRCh38
rs926374184 Chr.21: 29056564-29056564 on Build GRCh38
rs145225862 Chr.21: 29056428-29056430 on Build GRCh38
rs35958494 Chr.21: 29056380-29056381 on Build GRCh38
rs1012199077 Chr.21: 29056372-29056372 on Build GRCh38
rs980494741 Chr.21: 29056546-29056546 on Build GRCh38
rs147534472 Chr.21: 29056258-29056258 on Build GRCh38
rs776680408 Chr.21: 29056489-29056489 on Build GRCh38
rs752837551 Chr.21: 29056465-29056466 on Build GRCh38
rs772298151 Chr.21: 29056446-29056446 on Build GRCh38
rs778241996 Chr.21: 29056430-29056430 on Build GRCh38
rs112843637 Chr.21: 29056673-29056673 on Build GRCh38
rs932137827 Chr.21: 29056245-29056245 on Build GRCh38
rs765288666 Chr.21: 29056500-29056500 on Build GRCh38
rs762377991 Chr.21: 29056504-29056504 on Build GRCh38

View Full Product Details