Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 29062189-29062727 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCT8
Entrez Gene ID
10694
Gene Name
chaperonin containing TCP1 subunit 8
Gene Aliases
C21orf112, Cctq, D21S246, PRED71
UniGene
Hs.125113
Gene Chromosome Location
Chr.21: 29056322-29073797 on Build GRCh38
Transcripts
NM_001282907.1 NM_001282908.1 NM_001282909.1 NM_006585.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4844475 c.1027G>C p.E343Q Chr.21: 29062397-29062397 on Build GRCh38
1029775 c.1137C>A p.G379G Chr.21: 29062203-29062203 on Build GRCh38
1029776 c.1132C>T p.R378* Chr.21: 29062208-29062208 on Build GRCh38
4101023 c.1133G>A p.R378Q Chr.21: 29062207-29062207 on Build GRCh38
6623358 c.972T>A p.L324L Chr.21: 29062526-29062526 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756575146 Chr.21: 29062602-29062602 on Build GRCh38
rs780515483 Chr.21: 29062292-29062292 on Build GRCh38
rs950309198 Chr.21: 29062332-29062332 on Build GRCh38
rs774138325 Chr.21: 29062217-29062217 on Build GRCh38
rs773393355 Chr.21: 29062317-29062317 on Build GRCh38
rs542007088 Chr.21: 29062230-29062230 on Build GRCh38
rs750110241 Chr.21: 29062573-29062573 on Build GRCh38
rs889053737 Chr.21: 29062625-29062625 on Build GRCh38
rs199996167 Chr.21: 29062445-29062445 on Build GRCh38
rs376145854 Chr.21: 29062486-29062486 on Build GRCh38
rs142359495 Chr.21: 29062213-29062213 on Build GRCh38
rs144490490 Chr.21: 29062633-29062633 on Build GRCh38
rs1016002276 Chr.21: 29062268-29062268 on Build GRCh38
rs768735474 Chr.21: 29062548-29062548 on Build GRCh38
rs759471310 Chr.21: 29062218-29062218 on Build GRCh38
rs764827078 Chr.21: 29062414-29062414 on Build GRCh38
rs537872864 Chr.21: 29062324-29062324 on Build GRCh38
rs201833105 Chr.21: 29062379-29062379 on Build GRCh38
rs373495330 Chr.21: 29062645-29062645 on Build GRCh38
rs763820874 Chr.21: 29062261-29062261 on Build GRCh38
rs901693513 Chr.21: 29062569-29062569 on Build GRCh38
rs756885428 Chr.21: 29062412-29062412 on Build GRCh38
rs753608433 Chr.21: 29062276-29062276 on Build GRCh38
rs532778136 Chr.21: 29062605-29062605 on Build GRCh38
rs981736665 Chr.21: 29062350-29062350 on Build GRCh38
rs988106685 Chr.21: 29062639-29062639 on Build GRCh38
rs1015970797 Chr.21: 29062634-29062634 on Build GRCh38
rs139875242 Chr.21: 29062373-29062373 on Build GRCh38
rs774158410 Chr.21: 29062346-29062346 on Build GRCh38
rs138368687 Chr.21: 29062305-29062305 on Build GRCh38
rs747353614 Chr.21: 29062299-29062299 on Build GRCh38
rs373381503 Chr.21: 29062427-29062427 on Build GRCh38
rs961757315 Chr.21: 29062641-29062641 on Build GRCh38
rs201716852 Chr.21: 29062341-29062341 on Build GRCh38
rs749361467 Chr.21: 29062460-29062460 on Build GRCh38
rs984482434 Chr.21: 29062248-29062248 on Build GRCh38
rs748215410 Chr.21: 29062306-29062306 on Build GRCh38
rs764929960 Chr.21: 29062570-29062570 on Build GRCh38
rs937331765 Chr.21: 29062389-29062389 on Build GRCh38
rs371142113 Chr.21: 29062592-29062592 on Build GRCh38
rs755357363 Chr.21: 29062428-29062428 on Build GRCh38
rs370460323 Chr.21: 29062498-29062498 on Build GRCh38
rs759916213 Chr.21: 29062574-29062574 on Build GRCh38
rs760742565 Chr.21: 29062533-29062533 on Build GRCh38
rs151240700 Chr.21: 29062216-29062216 on Build GRCh38
rs971810202 Chr.21: 29062651-29062651 on Build GRCh38
rs571770028 Chr.21: 29062426-29062426 on Build GRCh38
rs781662485 Chr.21: 29062433-29062433 on Build GRCh38
rs377423345 Chr.21: 29062517-29062517 on Build GRCh38
rs961829217 Chr.21: 29062297-29062297 on Build GRCh38
rs943368295 Chr.21: 29062257-29062257 on Build GRCh38
rs771085615 Chr.21: 29062329-29062329 on Build GRCh38
rs759257153 Chr.21: 29062360-29062360 on Build GRCh38
rs767389625 Chr.21: 29062227-29062227 on Build GRCh38
rs971603353 Chr.21: 29062672-29062672 on Build GRCh38
rs767428116 Chr.21: 29062372-29062372 on Build GRCh38
rs749564968 Chr.21: 29062319-29062319 on Build GRCh38
rs189177098 Chr.21: 29062669-29062669 on Build GRCh38
rs987568174 Chr.21: 29062215-29062215 on Build GRCh38
rs752184452 Chr.21: 29062229-29062229 on Build GRCh38
rs554540181 Chr.21: 29062388-29062388 on Build GRCh38
rs954548223 Chr.21: 29062686-29062686 on Build GRCh38
rs556337944 Chr.21: 29062695-29062695 on Build GRCh38
rs376132055 Chr.21: 29062594-29062594 on Build GRCh38
rs61736413 Chr.21: 29062505-29062505 on Build GRCh38
rs927550868 Chr.21: 29062703-29062703 on Build GRCh38
rs369154122 Chr.21: 29062301-29062301 on Build GRCh38
rs761233152 Chr.21: 29062566-29062566 on Build GRCh38
rs760312150 Chr.21: 29062248-29062249 on Build GRCh38
rs745659979 Chr.21: 29062483-29062483 on Build GRCh38
rs1028896257 Chr.21: 29062612-29062612 on Build GRCh38
rs774447008 Chr.21: 29062479-29062479 on Build GRCh38
rs777785122 Chr.21: 29062457-29062457 on Build GRCh38
rs563173788 Chr.21: 29062664-29062664 on Build GRCh38
rs758658243 Chr.21: 29062279-29062279 on Build GRCh38
rs767920038 Chr.21: 29062576-29062576 on Build GRCh38
rs563709441 Chr.21: 29062562-29062562 on Build GRCh38
rs748693987 Chr.21: 29062436-29062436 on Build GRCh38
rs753518517 Chr.21: 29062410-29062410 on Build GRCh38
rs771355266 Chr.21: 29062249-29062249 on Build GRCh38
rs150997216 Chr.21: 29062560-29062561 on Build GRCh38
rs765797471 Chr.21: 29062591-29062594 on Build GRCh38
rs771120941 Chr.21: 29062471-29062471 on Build GRCh38
rs763734366 Chr.21: 29062381-29062381 on Build GRCh38
rs776988085 Chr.21: 29062466-29062467 on Build GRCh38

View Full Product Details