Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 29063035-29063578 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCT8
Entrez Gene ID
10694
Gene Name
chaperonin containing TCP1 subunit 8
Gene Aliases
C21orf112, Cctq, D21S246, PRED71
UniGene
Hs.125113
Gene Chromosome Location
Chr.21: 29056322-29073797 on Build GRCh38
Transcripts
NM_001282907.1 NM_001282908.1 NM_001282909.1 NM_006585.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
219967 c.928A>G p.I310V Chr.21: 29063365-29063365 on Build GRCh38
6399300 c.914C>A p.A305E Chr.21: 29063379-29063379 on Build GRCh38
6623357 c.826A>G p.M276V Chr.21: 29063467-29063467 on Build GRCh38
1307699 c.924T>C p.Y308Y Chr.21: 29063369-29063369 on Build GRCh38
5986842 c.837A>C p.Q279H Chr.21: 29063456-29063456 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752151561 Chr.21: 29063338-29063338 on Build GRCh38
rs958906932 Chr.21: 29063085-29063085 on Build GRCh38
rs34511083 Chr.21: 29063240-29063240 on Build GRCh38
rs764247884 Chr.21: 29063326-29063327 on Build GRCh38
rs983413051 Chr.21: 29063324-29063325 on Build GRCh38
rs145989778 Chr.21: 29063203-29063203 on Build GRCh38
rs769794366 Chr.21: 29063534-29063534 on Build GRCh38
rs573032869 Chr.21: 29063435-29063435 on Build GRCh38
rs200392501 Chr.21: 29063324-29063324 on Build GRCh38
rs199710268 Chr.21: 29063420-29063420 on Build GRCh38
rs779186383 Chr.21: 29063315-29063315 on Build GRCh38
rs910395190 Chr.21: 29063271-29063271 on Build GRCh38
rs548038821 Chr.21: 29063284-29063284 on Build GRCh38
rs543402449 Chr.21: 29063205-29063205 on Build GRCh38
rs185760796 Chr.21: 29063218-29063218 on Build GRCh38
rs139983851 Chr.21: 29063514-29063514 on Build GRCh38
rs980395818 Chr.21: 29063327-29063327 on Build GRCh38
rs201375014 Chr.21: 29063423-29063423 on Build GRCh38
rs766054334 Chr.21: 29063378-29063378 on Build GRCh38
rs765558716 Chr.21: 29063102-29063102 on Build GRCh38
rs1042110888 Chr.21: 29063213-29063213 on Build GRCh38
rs762467646 Chr.21: 29063375-29063375 on Build GRCh38
rs147651768 Chr.21: 29063489-29063489 on Build GRCh38
rs1011725777 Chr.21: 29063371-29063371 on Build GRCh38
rs967639253 Chr.21: 29063466-29063466 on Build GRCh38
rs914553112 Chr.21: 29063189-29063189 on Build GRCh38
rs180857608 Chr.21: 29063339-29063339 on Build GRCh38
rs200482519 Chr.21: 29063325-29063326 on Build GRCh38
rs772560443 Chr.21: 29063547-29063547 on Build GRCh38
rs142165839 Chr.21: 29063500-29063500 on Build GRCh38
rs1042058151 Chr.21: 29063382-29063382 on Build GRCh38
rs550109441 Chr.21: 29063316-29063316 on Build GRCh38
rs771967633 Chr.21: 29063326-29063326 on Build GRCh38
rs368757807 Chr.21: 29063401-29063401 on Build GRCh38
rs986632573 Chr.21: 29063548-29063548 on Build GRCh38
rs893424979 Chr.21: 29063119-29063119 on Build GRCh38
rs748087114 Chr.21: 29063524-29063524 on Build GRCh38
rs563177959 Chr.21: 29063216-29063216 on Build GRCh38
rs557412691 Chr.21: 29063144-29063144 on Build GRCh38
rs763133309 Chr.21: 29063326-29063326 on Build GRCh38
rs75366973 Chr.21: 29063337-29063337 on Build GRCh38
rs757335304 Chr.21: 29063312-29063312 on Build GRCh38
rs61736414 Chr.21: 29063492-29063492 on Build GRCh38
rs376913863 Chr.21: 29063367-29063367 on Build GRCh38
rs769229817 Chr.21: 29063364-29063364 on Build GRCh38
rs577689730 Chr.21: 29063152-29063152 on Build GRCh38
rs1038952132 Chr.21: 29063439-29063439 on Build GRCh38
rs555255537 Chr.21: 29063360-29063360 on Build GRCh38
rs190082888 Chr.21: 29063291-29063291 on Build GRCh38
rs74679876 Chr.21: 29063336-29063336 on Build GRCh38
rs1017826091 Chr.21: 29063237-29063237 on Build GRCh38
rs752997338 Chr.21: 29063389-29063389 on Build GRCh38
rs768066904 Chr.21: 29063274-29063274 on Build GRCh38
rs781511332 Chr.21: 29063413-29063413 on Build GRCh38
rs936188346 Chr.21: 29063346-29063346 on Build GRCh38
rs372863710 Chr.21: 29063356-29063356 on Build GRCh38
rs1017818325 Chr.21: 29063388-29063388 on Build GRCh38
rs778906454 Chr.21: 29063424-29063424 on Build GRCh38
rs1009245880 Chr.21: 29063158-29063158 on Build GRCh38
rs753901251 Chr.21: 29063305-29063305 on Build GRCh38
rs533308190 Chr.21: 29063304-29063304 on Build GRCh38
rs562266631 Chr.21: 29063177-29063178 on Build GRCh38
rs181509608 Chr.21: 29063072-29063072 on Build GRCh38
rs914989603 Chr.21: 29063365-29063367 on Build GRCh38
rs758651319 Chr.21: 29063450-29063450 on Build GRCh38
rs369911647 Chr.21: 29063352-29063352 on Build GRCh38
rs977448136 Chr.21: 29063487-29063487 on Build GRCh38
rs554795807 Chr.21: 29063283-29063283 on Build GRCh38
rs1034968073 Chr.21: 29063068-29063068 on Build GRCh38
rs923374242 Chr.21: 29063230-29063230 on Build GRCh38
rs993164193 Chr.21: 29063313-29063313 on Build GRCh38
rs565670373 Chr.21: 29063422-29063422 on Build GRCh38
rs761044325 Chr.21: 29063395-29063395 on Build GRCh38
rs570410962 Chr.21: 29063253-29063253 on Build GRCh38
rs557577495 Chr.21: 29063481-29063481 on Build GRCh38
rs750537848 Chr.21: 29063436-29063436 on Build GRCh38

View Full Product Details