Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 29064852-29065358 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCT8
Entrez Gene ID
10694
Gene Name
chaperonin containing TCP1 subunit 8
Gene Aliases
C21orf112, Cctq, D21S246, PRED71
UniGene
Hs.125113
Gene Chromosome Location
Chr.21: 29056322-29073797 on Build GRCh38
Transcripts
NM_001282907.1 NM_001282908.1 NM_001282909.1 NM_006585.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5883624 c.625-1G>A p.? Chr.21: 29065106-29065106 on Build GRCh38
5631512 c.715A>G p.I239V Chr.21: 29065015-29065015 on Build GRCh38
4924299 c.687T>G p.G229G Chr.21: 29065043-29065043 on Build GRCh38
279699 c.627C>T p.G209G Chr.21: 29065103-29065103 on Build GRCh38
1681908 c.754G>T p.E252* Chr.21: 29064976-29064976 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776818853 Chr.21: 29065015-29065015 on Build GRCh38
rs930886224 Chr.21: 29065243-29065243 on Build GRCh38
rs112581414 Chr.21: 29065003-29065003 on Build GRCh38
rs748787550 Chr.21: 29064989-29064989 on Build GRCh38
rs558149122 Chr.21: 29064896-29064896 on Build GRCh38
rs780802809 Chr.21: 29064959-29064959 on Build GRCh38
rs748447177 Chr.21: 29065153-29065153 on Build GRCh38
rs921305818 Chr.21: 29065304-29065304 on Build GRCh38
rs766615097 Chr.21: 29065036-29065036 on Build GRCh38
rs770260379 Chr.21: 29065156-29065156 on Build GRCh38
rs147091611 Chr.21: 29064880-29064880 on Build GRCh38
rs939409055 Chr.21: 29065301-29065301 on Build GRCh38
rs1047764303 Chr.21: 29064879-29064879 on Build GRCh38
rs924684230 Chr.21: 29065261-29065261 on Build GRCh38
rs765803538 Chr.21: 29064921-29064921 on Build GRCh38
rs138452746 Chr.21: 29064950-29064950 on Build GRCh38
rs766694746 Chr.21: 29064942-29064942 on Build GRCh38
rs755999088 Chr.21: 29065055-29065055 on Build GRCh38
rs895263648 Chr.21: 29065322-29065322 on Build GRCh38
rs1002949087 Chr.21: 29065161-29065161 on Build GRCh38
rs188032190 Chr.21: 29065223-29065223 on Build GRCh38
rs879035328 Chr.21: 29065136-29065136 on Build GRCh38
rs903740369 Chr.21: 29065332-29065332 on Build GRCh38
rs977518784 Chr.21: 29064936-29064936 on Build GRCh38
rs527960785 Chr.21: 29064947-29064947 on Build GRCh38
rs543495048 Chr.21: 29065282-29065282 on Build GRCh38
rs367740639 Chr.21: 29065157-29065157 on Build GRCh38
rs958687611 Chr.21: 29064887-29064890 on Build GRCh38
rs1012059410 Chr.21: 29064969-29064969 on Build GRCh38
rs767787828 Chr.21: 29065049-29065049 on Build GRCh38
rs535304949 Chr.21: 29064894-29064894 on Build GRCh38
rs529788710 Chr.21: 29065312-29065312 on Build GRCh38
rs1022122867 Chr.21: 29065324-29065324 on Build GRCh38
rs148840751 Chr.21: 29065035-29065035 on Build GRCh38
rs771775162 Chr.21: 29065011-29065011 on Build GRCh38
rs745566853 Chr.21: 29065009-29065009 on Build GRCh38
rs1036008303 Chr.21: 29064883-29064883 on Build GRCh38
rs745786262 Chr.21: 29065051-29065051 on Build GRCh38
rs762175370 Chr.21: 29064917-29064917 on Build GRCh38
rs991432404 Chr.21: 29065240-29065240 on Build GRCh38
rs567474269 Chr.21: 29065278-29065278 on Build GRCh38
rs755940332 Chr.21: 29064965-29064965 on Build GRCh38
rs142657269 Chr.21: 29065034-29065034 on Build GRCh38
rs991712663 Chr.21: 29064893-29064893 on Build GRCh38
rs769749225 Chr.21: 29065169-29065169 on Build GRCh38
rs762240052 Chr.21: 29065018-29065018 on Build GRCh38
rs968540408 Chr.21: 29065260-29065260 on Build GRCh38
rs944026400 Chr.21: 29064988-29064988 on Build GRCh38
rs1041087387 Chr.21: 29064990-29064990 on Build GRCh38
rs771556080 Chr.21: 29065130-29065130 on Build GRCh38
rs750400684 Chr.21: 29064931-29064931 on Build GRCh38
rs779768213 Chr.21: 29065134-29065134 on Build GRCh38
rs773957587 Chr.21: 29064996-29064996 on Build GRCh38
rs148204307 Chr.21: 29065200-29065200 on Build GRCh38
rs955264743 Chr.21: 29065092-29065092 on Build GRCh38
rs546735886 Chr.21: 29065315-29065315 on Build GRCh38
rs182987613 Chr.21: 29064994-29064994 on Build GRCh38
rs773913046 Chr.21: 29065033-29065033 on Build GRCh38
rs745410286 Chr.21: 29065121-29065121 on Build GRCh38
rs1012026864 Chr.21: 29065323-29065323 on Build GRCh38
rs192821292 Chr.21: 29064961-29064961 on Build GRCh38
rs370283212 Chr.21: 29065265-29065265 on Build GRCh38
rs755212663 Chr.21: 29065048-29065048 on Build GRCh38
rs143385323 Chr.21: 29065043-29065043 on Build GRCh38
rs552427135 Chr.21: 29065314-29065314 on Build GRCh38
rs777570949 Chr.21: 29065076-29065076 on Build GRCh38
rs36080401 Chr.21: 29065297-29065298 on Build GRCh38
rs778343795 Chr.21: 29065117-29065117 on Build GRCh38
rs755168027 Chr.21: 29065031-29065031 on Build GRCh38
rs751465117 Chr.21: 29065260-29065261 on Build GRCh38
rs756733846 Chr.21: 29065111-29065111 on Build GRCh38
rs563945345 Chr.21: 29064985-29064985 on Build GRCh38
rs141148644 Chr.21: 29065277-29065277 on Build GRCh38
rs749260785 Chr.21: 29065100-29065100 on Build GRCh38
rs752884645 Chr.21: 29065242-29065249 on Build GRCh38
rs565031384 Chr.21: 29065217-29065217 on Build GRCh38
rs765605189 Chr.21: 29065023-29065023 on Build GRCh38

View Full Product Details