Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 34446502-34447033 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNE1
Entrez Gene ID
3753
Gene Name
potassium voltage-gated channel subfamily E regulatory subunit 1
Gene Aliases
ISK, JLNS, JLNS2, LQT2/5, LQT5, MinK
UniGene
Hs.121495
Gene Chromosome Location
Chr.21: 34446688-34512275 on Build GRCh38
Transcripts
NM_000219.5 NM_001127668.3 NM_001127669.3 NM_001127670.3 NM_001270402.2 NM_001270403.2 NM_001270404.2 NM_001270405.2 XM_017028342.1 XM_006723946.2 XM_006723949.2

Gene Symbol
KCNE1B
Entrez Gene ID
102723475
Gene Name
potassium voltage-gated channel subfamily E regulatory subunit 1B
Gene Aliases
-
UniGene
Gene Chromosome Location
Chr.21: 7816677-7829632 on Build GRCh38
Transcripts
NM_000219.5 NM_001127668.3 NM_001127669.3 NM_001127670.3 NM_001270402.2 NM_001270403.2 NM_001270404.2 NM_001270405.2 XM_017028342.1 XM_006723946.2 XM_006723949.2

SNP Detail

SNP ID Chromosome Location
rs1039996720 Chr.21: 34446825-34446825 on Build GRCh38
rs747442476 Chr.21: 34446832-34446832 on Build GRCh38
rs936209002 Chr.21: 34446819-34446819 on Build GRCh38
rs552526806 Chr.21: 34446658-34446658 on Build GRCh38
rs528498228 Chr.21: 34446926-34446926 on Build GRCh38
rs1016954150 Chr.21: 34446664-34446664 on Build GRCh38
rs564052299 Chr.21: 34446605-34446605 on Build GRCh38
rs769307001 Chr.21: 34446929-34446929 on Build GRCh38
rs543391377 Chr.21: 34446790-34446790 on Build GRCh38
rs934192023 Chr.21: 34446846-34446846 on Build GRCh38
rs896302148 Chr.21: 34446947-34446947 on Build GRCh38
rs3453 Chr.21: 34446765-34446765 on Build GRCh38
rs200481471 Chr.21: 34446927-34446927 on Build GRCh38
rs73363349 Chr.21: 34446660-34446660 on Build GRCh38
rs2834485 Chr.21: 34446716-34446716 on Build GRCh38
rs1033926419 Chr.21: 34446746-34446746 on Build GRCh38
rs13050198 Chr.21: 34446784-34446784 on Build GRCh38
rs373970167 Chr.21: 34446916-34446916 on Build GRCh38
rs774883610 Chr.21: 34446980-34446980 on Build GRCh38
rs1002136141 Chr.21: 34446717-34446717 on Build GRCh38
rs376395432 Chr.21: 34446668-34446668 on Build GRCh38
rs1051159775 Chr.21: 34446618-34446618 on Build GRCh38
rs567630448 Chr.21: 34446852-34446852 on Build GRCh38
rs745512218 Chr.21: 34446925-34446925 on Build GRCh38
rs368975115 Chr.21: 34446647-34446647 on Build GRCh38
rs1038743204 Chr.21: 34446910-34446910 on Build GRCh38
rs879427774 Chr.21: 34446592-34446592 on Build GRCh38
rs866859424 Chr.21: 34446965-34446965 on Build GRCh38
rs887169387 Chr.21: 34446895-34446895 on Build GRCh38
rs1033631203 Chr.21: 34446944-34446944 on Build GRCh38
rs554562103 Chr.21: 34446767-34446767 on Build GRCh38
rs940123354 Chr.21: 34446909-34446909 on Build GRCh38
rs529749296 Chr.21: 34446540-34446540 on Build GRCh38
rs569634817 Chr.21: 34446575-34446575 on Build GRCh38
rs998223522 Chr.21: 34446585-34446585 on Build GRCh38
rs924978266 Chr.21: 34446811-34446811 on Build GRCh38
rs373184377 Chr.21: 34446723-34446723 on Build GRCh38
rs558859988 Chr.21: 34446897-34446897 on Build GRCh38
rs566164652 Chr.21: 34446737-34446737 on Build GRCh38
rs913676929 Chr.21: 34446770-34446770 on Build GRCh38
rs532307231 Chr.21: 34446551-34446551 on Build GRCh38
rs966414305 Chr.21: 34446993-34446993 on Build GRCh38
rs778082466 Chr.21: 34446786-34446786 on Build GRCh38
rs936278394 Chr.21: 34446524-34446524 on Build GRCh38
rs1004193896 Chr.21: 34446662-34446662 on Build GRCh38
rs1020810519 Chr.21: 34446966-34446966 on Build GRCh38
rs552886835 Chr.21: 34446731-34446731 on Build GRCh38
rs548171786 Chr.21: 34446570-34446570 on Build GRCh38
rs530848740 Chr.21: 34446624-34446624 on Build GRCh38
rs780840352 Chr.21: 34446915-34446915 on Build GRCh38

View Full Product Details