Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 34716613-34717150 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLIC6
Entrez Gene ID
54102
Gene Name
chloride intracellular channel 6
Gene Aliases
CLIC1L
UniGene
Hs.473695
Gene Chromosome Location
Chr.21: 34668953-34718227 on Build GRCh38
Transcripts
NM_001317009.1 NM_053277.2

SNP Detail

SNP ID Chromosome Location
rs879571163 Chr.21: 34717080-34717080 on Build GRCh38
rs1028917824 Chr.21: 34716659-34716659 on Build GRCh38
rs900133609 Chr.21: 34716710-34716710 on Build GRCh38
rs1008848547 Chr.21: 34716879-34716879 on Build GRCh38
rs927539730 Chr.21: 34716831-34716831 on Build GRCh38
rs142359746 Chr.21: 34717010-34717010 on Build GRCh38
rs75433027 Chr.21: 34716900-34716901 on Build GRCh38
rs780769343 Chr.21: 34716862-34716863 on Build GRCh38
rs567028557 Chr.21: 34716767-34716767 on Build GRCh38
rs866201575 Chr.21: 34716859-34716859 on Build GRCh38
rs1001375340 Chr.21: 34717096-34717096 on Build GRCh38
rs777476295 Chr.21: 34716986-34716986 on Build GRCh38
rs60037226 Chr.21: 34716857-34716857 on Build GRCh38
rs151283784 Chr.21: 34717107-34717107 on Build GRCh38
rs142197835 Chr.21: 34716831-34716832 on Build GRCh38
rs774282892 Chr.21: 34716864-34716865 on Build GRCh38
rs959972992 Chr.21: 34717115-34717115 on Build GRCh38
rs931501944 Chr.21: 34716818-34716818 on Build GRCh38
rs371353756 Chr.21: 34716858-34716861 on Build GRCh38
rs960270450 Chr.21: 34716855-34716864 on Build GRCh38
rs2834601 Chr.21: 34716756-34716756 on Build GRCh38
rs1034275583 Chr.21: 34716853-34716853 on Build GRCh38
rs994619492 Chr.21: 34716914-34716914 on Build GRCh38
rs1025732159 Chr.21: 34716922-34716922 on Build GRCh38
rs916877439 Chr.21: 34716994-34716994 on Build GRCh38
rs1004659245 Chr.21: 34716778-34716778 on Build GRCh38
rs10541127 Chr.21: 34716863-34716866 on Build GRCh38
rs992443799 Chr.21: 34716869-34716869 on Build GRCh38
rs1029282728 Chr.21: 34716867-34716867 on Build GRCh38
rs372212799 Chr.21: 34716868-34716868 on Build GRCh38
rs926146017 Chr.21: 34716644-34716644 on Build GRCh38
rs371385137 Chr.21: 34717117-34717117 on Build GRCh38
rs370933701 Chr.21: 34716833-34716833 on Build GRCh38
rs1016082127 Chr.21: 34716783-34716783 on Build GRCh38
rs374861299 Chr.21: 34716988-34716988 on Build GRCh38
rs375167292 Chr.21: 34717018-34717018 on Build GRCh38
rs780269590 Chr.21: 34716859-34716860 on Build GRCh38
rs1037463380 Chr.21: 34717042-34717042 on Build GRCh38
rs150674181 Chr.21: 34716863-34716870 on Build GRCh38
rs1019597265 Chr.21: 34716899-34716899 on Build GRCh38
rs552799972 Chr.21: 34716835-34716836 on Build GRCh38
rs934270092 Chr.21: 34716989-34716989 on Build GRCh38
rs148736581 Chr.21: 34716956-34716956 on Build GRCh38
rs373788888 Chr.21: 34716864-34716864 on Build GRCh38
rs927487909 Chr.21: 34716977-34716977 on Build GRCh38
rs367633959 Chr.21: 34716866-34716866 on Build GRCh38
rs796820318 Chr.21: 34716832-34716845 on Build GRCh38
rs913254079 Chr.21: 34716899-34716900 on Build GRCh38
rs965388520 Chr.21: 34716901-34716901 on Build GRCh38
rs895500979 Chr.21: 34716855-34716855 on Build GRCh38
rs942929865 Chr.21: 34716648-34716648 on Build GRCh38
rs953309270 Chr.21: 34716678-34716678 on Build GRCh38
rs538348744 Chr.21: 34717122-34717122 on Build GRCh38
rs1056706149 Chr.21: 34716835-34716835 on Build GRCh38
rs58117172 Chr.21: 34716860-34716861 on Build GRCh38
rs1000484348 Chr.21: 34716650-34716650 on Build GRCh38
rs952940033 Chr.21: 34716943-34716943 on Build GRCh38
rs1014609493 Chr.21: 34717124-34717124 on Build GRCh38
rs773237192 Chr.21: 34716863-34716864 on Build GRCh38
rs879327160 Chr.21: 34716860-34716860 on Build GRCh38
rs192573042 Chr.21: 34716998-34716998 on Build GRCh38
rs59598168 Chr.21: 34716861-34716861 on Build GRCh38
rs1008794550 Chr.21: 34717095-34717095 on Build GRCh38
rs372671905 Chr.21: 34716862-34716862 on Build GRCh38
rs749558731 Chr.21: 34716789-34716789 on Build GRCh38
rs867530251 Chr.21: 34716860-34716860 on Build GRCh38
rs531310007 Chr.21: 34716801-34716801 on Build GRCh38
rs1019651337 Chr.21: 34716760-34716760 on Build GRCh38
rs984743830 Chr.21: 34716824-34716824 on Build GRCh38
rs945742264 Chr.21: 34716995-34716995 on Build GRCh38
rs75634757 Chr.21: 34717084-34717084 on Build GRCh38
rs935773381 Chr.21: 34717019-34717019 on Build GRCh38
rs571157227 Chr.21: 34717108-34717108 on Build GRCh38
rs1026104315 Chr.21: 34717128-34717128 on Build GRCh38
rs751500768 Chr.21: 34716743-34716743 on Build GRCh38
rs369686913 Chr.21: 34716699-34716699 on Build GRCh38
rs753970537 Chr.21: 34716832-34716835 on Build GRCh38
rs972855028 Chr.21: 34716788-34716788 on Build GRCh38
rs59448002 Chr.21: 34716897-34716900 on Build GRCh38
rs578009161 Chr.21: 34716980-34716980 on Build GRCh38

View Full Product Details