Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 34717037-34717539 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLIC6
Entrez Gene ID
54102
Gene Name
chloride intracellular channel 6
Gene Aliases
CLIC1L
UniGene
Hs.473695
Gene Chromosome Location
Chr.21: 34668953-34718227 on Build GRCh38
Transcripts
NM_001317009.1 NM_053277.2

SNP Detail

SNP ID Chromosome Location
rs1020163809 Chr.21: 34717483-34717483 on Build GRCh38
rs879571163 Chr.21: 34717080-34717080 on Build GRCh38
rs993844796 Chr.21: 34717344-34717344 on Build GRCh38
rs888464953 Chr.21: 34717377-34717377 on Build GRCh38
rs917491110 Chr.21: 34717202-34717202 on Build GRCh38
rs35366741 Chr.21: 34717453-34717454 on Build GRCh38
rs1014375736 Chr.21: 34717472-34717472 on Build GRCh38
rs572003894 Chr.21: 34717494-34717494 on Build GRCh38
rs978092024 Chr.21: 34717501-34717501 on Build GRCh38
rs183288149 Chr.21: 34717403-34717403 on Build GRCh38
rs796736219 Chr.21: 34717410-34717410 on Build GRCh38
rs1001375340 Chr.21: 34717096-34717096 on Build GRCh38
rs907503643 Chr.21: 34717358-34717358 on Build GRCh38
rs140277579 Chr.21: 34717313-34717313 on Build GRCh38
rs1040265708 Chr.21: 34717306-34717306 on Build GRCh38
rs151283784 Chr.21: 34717107-34717107 on Build GRCh38
rs935736521 Chr.21: 34717241-34717241 on Build GRCh38
rs959972992 Chr.21: 34717115-34717115 on Build GRCh38
rs1034458297 Chr.21: 34717182-34717182 on Build GRCh38
rs748121971 Chr.21: 34717146-34717146 on Build GRCh38
rs968076705 Chr.21: 34717217-34717217 on Build GRCh38
rs560362896 Chr.21: 34717308-34717308 on Build GRCh38
rs772183688 Chr.21: 34717238-34717238 on Build GRCh38
rs577908602 Chr.21: 34717185-34717185 on Build GRCh38
rs993006698 Chr.21: 34717198-34717198 on Build GRCh38
rs544901406 Chr.21: 34717271-34717271 on Build GRCh38
rs371385137 Chr.21: 34717117-34717117 on Build GRCh38
rs925640627 Chr.21: 34717227-34717227 on Build GRCh38
rs967362373 Chr.21: 34717180-34717180 on Build GRCh38
rs144123421 Chr.21: 34717508-34717508 on Build GRCh38
rs560560030 Chr.21: 34717396-34717396 on Build GRCh38
rs897738401 Chr.21: 34717343-34717343 on Build GRCh38
rs113291551 Chr.21: 34717514-34717514 on Build GRCh38
rs1020259843 Chr.21: 34717163-34717163 on Build GRCh38
rs958868276 Chr.21: 34717194-34717194 on Build GRCh38
rs1005554693 Chr.21: 34717395-34717395 on Build GRCh38
rs747488601 Chr.21: 34717347-34717347 on Build GRCh38
rs538904262 Chr.21: 34717277-34717277 on Build GRCh38
rs940341440 Chr.21: 34717371-34717371 on Build GRCh38
rs538348744 Chr.21: 34717122-34717122 on Build GRCh38
rs35473052 Chr.21: 34717415-34717416 on Build GRCh38
rs773664111 Chr.21: 34717284-34717284 on Build GRCh38
rs1014609493 Chr.21: 34717124-34717124 on Build GRCh38
rs1014142092 Chr.21: 34717442-34717442 on Build GRCh38
rs977098331 Chr.21: 34717223-34717223 on Build GRCh38
rs556734692 Chr.21: 34717157-34717157 on Build GRCh38
rs894598369 Chr.21: 34717468-34717468 on Build GRCh38
rs542563782 Chr.21: 34717339-34717339 on Build GRCh38
rs1008794550 Chr.21: 34717095-34717095 on Build GRCh38
rs75634757 Chr.21: 34717084-34717084 on Build GRCh38
rs571157227 Chr.21: 34717108-34717108 on Build GRCh38
rs1026104315 Chr.21: 34717128-34717128 on Build GRCh38
rs1032655005 Chr.21: 34717507-34717507 on Build GRCh38
rs1033007039 Chr.21: 34717506-34717506 on Build GRCh38
rs549464915 Chr.21: 34717486-34717486 on Build GRCh38
rs1037412388 Chr.21: 34717391-34717391 on Build GRCh38
rs1006077749 Chr.21: 34717159-34717159 on Build GRCh38
rs910346117 Chr.21: 34717279-34717279 on Build GRCh38
rs983453494 Chr.21: 34717352-34717352 on Build GRCh38
rs895655585 Chr.21: 34717422-34717422 on Build GRCh38

View Full Product Details