Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 36134883-36135388 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CBR3
Entrez Gene ID
874
Gene Name
carbonyl reductase 3
Gene Aliases
HEL-S-25, SDR21C2, hCBR3
UniGene
Hs.154510
Gene Chromosome Location
Chr.21: 36134901-36146562 on Build GRCh38
Transcripts
NM_001236.3 XM_011529772.2 NR_038892.1 NR_038893.1

Gene Symbol
CBR3-AS1
Entrez Gene ID
100506428
Gene Name
CBR3 antisense RNA 1
Gene Aliases
PlncRNA-1, PlncRNA1
UniGene
Hs.651214
Gene Chromosome Location
Chr.21: 36131767-36156308 on Build GRCh38
Transcripts
NM_001236.3 XM_011529772.2 NR_038892.1 NR_038893.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
444425 c.11G>A p.C4Y Chr.21: 36135203-36135203 on Build GRCh38
5741981 c.50T>C p.I17T Chr.21: 36135242-36135242 on Build GRCh38
7312289 c.144G>T p.Q48H Chr.21: 36135336-36135336 on Build GRCh38
1632471 c.156G>A p.A52A Chr.21: 36135348-36135348 on Build GRCh38
7304605 c.122C>T p.A41V Chr.21: 36135314-36135314 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs577004977 Chr.21: 36135120-36135120 on Build GRCh38
rs537344040 Chr.21: 36135108-36135108 on Build GRCh38
rs551927137 Chr.21: 36134944-36134944 on Build GRCh38
rs1012215813 Chr.21: 36134941-36134941 on Build GRCh38
rs775199518 Chr.21: 36135208-36135208 on Build GRCh38
rs778802502 Chr.21: 36135310-36135310 on Build GRCh38
rs988972898 Chr.21: 36135014-36135014 on Build GRCh38
rs888457474 Chr.21: 36134935-36134935 on Build GRCh38
rs756334238 Chr.21: 36135183-36135183 on Build GRCh38
rs763727652 Chr.21: 36135226-36135226 on Build GRCh38
rs113315710 Chr.21: 36135157-36135157 on Build GRCh38
rs974196448 Chr.21: 36134940-36134940 on Build GRCh38
rs945487810 Chr.21: 36135063-36135063 on Build GRCh38
rs1045067564 Chr.21: 36135151-36135151 on Build GRCh38
rs977009601 Chr.21: 36135084-36135084 on Build GRCh38
rs746788841 Chr.21: 36135331-36135331 on Build GRCh38
rs200563007 Chr.21: 36135249-36135249 on Build GRCh38
rs778208355 Chr.21: 36135191-36135191 on Build GRCh38
rs925387280 Chr.21: 36135351-36135351 on Build GRCh38
rs563525294 Chr.21: 36135289-36135289 on Build GRCh38
rs918766631 Chr.21: 36135097-36135097 on Build GRCh38
rs771994596 Chr.21: 36135323-36135323 on Build GRCh38
rs778118640 Chr.21: 36135287-36135287 on Build GRCh38
rs572427756 Chr.21: 36135067-36135067 on Build GRCh38
rs772527690 Chr.21: 36135354-36135354 on Build GRCh38
rs535434585 Chr.21: 36135008-36135008 on Build GRCh38
rs903869374 Chr.21: 36135102-36135102 on Build GRCh38
rs926894523 Chr.21: 36134961-36134961 on Build GRCh38
rs761325372 Chr.21: 36135347-36135347 on Build GRCh38
rs745966181 Chr.21: 36135313-36135313 on Build GRCh38
rs574795759 Chr.21: 36135235-36135235 on Build GRCh38
rs896952920 Chr.21: 36135142-36135142 on Build GRCh38
rs749373377 Chr.21: 36135196-36135196 on Build GRCh38
rs372270313 Chr.21: 36135327-36135327 on Build GRCh38
rs8133052 Chr.21: 36135203-36135203 on Build GRCh38
rs979603464 Chr.21: 36135346-36135346 on Build GRCh38
rs770963323 Chr.21: 36135197-36135197 on Build GRCh38
rs762615026 Chr.21: 36135360-36135360 on Build GRCh38
rs939577102 Chr.21: 36135028-36135028 on Build GRCh38
rs768120895 Chr.21: 36135362-36135362 on Build GRCh38
rs970105109 Chr.21: 36134951-36134951 on Build GRCh38
rs1015390360 Chr.21: 36135135-36135135 on Build GRCh38
rs745745422 Chr.21: 36135198-36135198 on Build GRCh38
rs756604189 Chr.21: 36135285-36135285 on Build GRCh38
rs768139296 Chr.21: 36135274-36135274 on Build GRCh38
rs968392712 Chr.21: 36135041-36135041 on Build GRCh38
rs774046253 Chr.21: 36134988-36134988 on Build GRCh38
rs979775017 Chr.21: 36134960-36134960 on Build GRCh38
rs1035354567 Chr.21: 36135269-36135269 on Build GRCh38
rs45510501 Chr.21: 36134906-36134906 on Build GRCh38
rs985082944 Chr.21: 36135048-36135048 on Build GRCh38
rs558752456 Chr.21: 36135117-36135117 on Build GRCh38
rs1021362849 Chr.21: 36135316-36135316 on Build GRCh38
rs932665880 Chr.21: 36135103-36135103 on Build GRCh38
rs772147691 Chr.21: 36135114-36135114 on Build GRCh38
rs768484134 Chr.21: 36135332-36135332 on Build GRCh38
rs969823249 Chr.21: 36135259-36135259 on Build GRCh38
rs944270766 Chr.21: 36135075-36135075 on Build GRCh38
rs761632509 Chr.21: 36135034-36135034 on Build GRCh38
rs772208537 Chr.21: 36135284-36135304 on Build GRCh38
rs1044170020 Chr.21: 36135101-36135101 on Build GRCh38
rs369773796 Chr.21: 36135162-36135162 on Build GRCh38
rs112226760 Chr.21: 36135345-36135345 on Build GRCh38
rs897026058 Chr.21: 36135246-36135247 on Build GRCh38
rs769497289 Chr.21: 36135352-36135352 on Build GRCh38
rs570053275 Chr.21: 36135056-36135056 on Build GRCh38
rs374238468 Chr.21: 36135339-36135339 on Build GRCh38
rs757528534 Chr.21: 36135295-36135295 on Build GRCh38
rs961149609 Chr.21: 36134922-36134922 on Build GRCh38
rs138199629 Chr.21: 36135282-36135282 on Build GRCh38
rs905764796 Chr.21: 36134952-36134952 on Build GRCh38
rs764614988 Chr.21: 36135236-36135236 on Build GRCh38
rs556642930 Chr.21: 36135032-36135032 on Build GRCh38
rs760530968 Chr.21: 36135216-36135216 on Build GRCh38
rs776198944 Chr.21: 36135230-36135230 on Build GRCh38
rs980211521 Chr.21: 36135260-36135260 on Build GRCh38
rs541190073 Chr.21: 36135123-36135123 on Build GRCh38
rs992376255 Chr.21: 36135309-36135309 on Build GRCh38
rs955870883 Chr.21: 36135010-36135010 on Build GRCh38
rs1008095137 Chr.21: 36135128-36135128 on Build GRCh38

View Full Product Details