Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 44288267-44288776 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AIRE
Entrez Gene ID
326
Gene Name
autoimmune regulator
Gene Aliases
AIRE1, APECED, APS1, APSI, PGA1
UniGene
Hs.129829
Gene Chromosome Location
Chr.21: 44285838-44298219 on Build GRCh38
Transcripts
NM_000383.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4002050 c.588C>T p.S196S Chr.21: 44288394-44288394 on Build GRCh38
6467999 c.637C>T p.Q213* Chr.21: 44288443-44288443 on Build GRCh38
4101920 c.539-1G>T p.? Chr.21: 44288344-44288344 on Build GRCh38
6880202 c.604G>A p.A202T Chr.21: 44288410-44288410 on Build GRCh38
5636717 c.612G>C p.G204G Chr.21: 44288418-44288418 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs572126327 Chr.21: 44288561-44288561 on Build GRCh38
rs768603759 Chr.21: 44288452-44288452 on Build GRCh38
rs765159981 Chr.21: 44288310-44288310 on Build GRCh38
rs917451165 Chr.21: 44288494-44288494 on Build GRCh38
rs780642720 Chr.21: 44288503-44288503 on Build GRCh38
rs41277544 Chr.21: 44288314-44288314 on Build GRCh38
rs543508973 Chr.21: 44288666-44288666 on Build GRCh38
rs369541283 Chr.21: 44288393-44288393 on Build GRCh38
rs929485089 Chr.21: 44288724-44288724 on Build GRCh38
rs922101020 Chr.21: 44288704-44288704 on Build GRCh38
rs140196414 Chr.21: 44288405-44288405 on Build GRCh38
rs887814723 Chr.21: 44288573-44288573 on Build GRCh38
rs751759051 Chr.21: 44288316-44288316 on Build GRCh38
rs574896544 Chr.21: 44288540-44288540 on Build GRCh38
rs771925480 Chr.21: 44288338-44288339 on Build GRCh38
rs1048190979 Chr.21: 44288526-44288526 on Build GRCh38
rs762910691 Chr.21: 44288392-44288392 on Build GRCh38
rs759510873 Chr.21: 44288308-44288308 on Build GRCh38
rs767844747 Chr.21: 44288399-44288399 on Build GRCh38
rs999236123 Chr.21: 44288625-44288625 on Build GRCh38
rs765762534 Chr.21: 44288477-44288477 on Build GRCh38
rs536914673 Chr.21: 44288322-44288322 on Build GRCh38
rs763047196 Chr.21: 44288318-44288319 on Build GRCh38
rs774634852 Chr.21: 44288455-44288455 on Build GRCh38
rs1009106769 Chr.21: 44288383-44288383 on Build GRCh38
rs960513263 Chr.21: 44288635-44288635 on Build GRCh38
rs564316895 Chr.21: 44288633-44288633 on Build GRCh38
rs556758490 Chr.21: 44288484-44288484 on Build GRCh38
rs929284151 Chr.21: 44288523-44288523 on Build GRCh38
rs1011174398 Chr.21: 44288585-44288585 on Build GRCh38
rs143952576 Chr.21: 44288366-44288366 on Build GRCh38
rs775304437 Chr.21: 44288433-44288433 on Build GRCh38
rs773143714 Chr.21: 44288348-44288348 on Build GRCh38
rs34219046 Chr.21: 44288354-44288354 on Build GRCh38
rs765761567 Chr.21: 44288677-44288677 on Build GRCh38
rs772221175 Chr.21: 44288464-44288464 on Build GRCh38
rs1800523 Chr.21: 44288475-44288475 on Build GRCh38
rs757848836 Chr.21: 44288336-44288336 on Build GRCh38
rs199612115 Chr.21: 44288459-44288459 on Build GRCh38
rs74162061 Chr.21: 44288401-44288401 on Build GRCh38
rs769525035 Chr.21: 44288376-44288376 on Build GRCh38
rs867155900 Chr.21: 44288428-44288428 on Build GRCh38
rs943362441 Chr.21: 44288574-44288574 on Build GRCh38
rs369232026 Chr.21: 44288446-44288446 on Build GRCh38
rs1029475010 Chr.21: 44288443-44288443 on Build GRCh38
rs548518643 Chr.21: 44288689-44288689 on Build GRCh38
rs776389119 Chr.21: 44288302-44288302 on Build GRCh38
rs777290325 Chr.21: 44288346-44288346 on Build GRCh38
rs774507800 Chr.21: 44288330-44288330 on Build GRCh38
rs201650973 Chr.21: 44288370-44288370 on Build GRCh38
rs929395671 Chr.21: 44288360-44288360 on Build GRCh38
rs900646672 Chr.21: 44288572-44288572 on Build GRCh38
rs531871611 Chr.21: 44288644-44288644 on Build GRCh38
rs779725226 Chr.21: 44288698-44288698 on Build GRCh38
rs562704578 Chr.21: 44288667-44288667 on Build GRCh38
rs992752920 Chr.21: 44288545-44288545 on Build GRCh38
rs1035087448 Chr.21: 44288628-44288628 on Build GRCh38
rs780838923 Chr.21: 44288422-44288422 on Build GRCh38
rs180838238 Chr.21: 44288751-44288751 on Build GRCh38
rs750100732 Chr.21: 44288508-44288508 on Build GRCh38
rs902178229 Chr.21: 44288624-44288624 on Build GRCh38
rs973964792 Chr.21: 44288651-44288651 on Build GRCh38
rs745459635 Chr.21: 44288425-44288425 on Build GRCh38
rs200261323 Chr.21: 44288325-44288325 on Build GRCh38
rs749744291 Chr.21: 44288334-44288334 on Build GRCh38
rs143522755 Chr.21: 44288682-44288682 on Build GRCh38
rs997581212 Chr.21: 44288637-44288637 on Build GRCh38
rs756768608 Chr.21: 44288499-44288499 on Build GRCh38
rs760027964 Chr.21: 44288473-44288473 on Build GRCh38
rs373277723 Chr.21: 44288465-44288465 on Build GRCh38
rs374858848 Chr.21: 44288313-44288313 on Build GRCh38
rs878081 Chr.21: 44288394-44288394 on Build GRCh38
rs944790496 Chr.21: 44288692-44288692 on Build GRCh38
rs750755686 Chr.21: 44288738-44288738 on Build GRCh38
rs754331960 Chr.21: 44288406-44288406 on Build GRCh38
rs769868098 Chr.21: 44288321-44288321 on Build GRCh38
rs775701048 Chr.21: 44288312-44288312 on Build GRCh38
rs527562882 Chr.21: 44288729-44288729 on Build GRCh38
rs760661721 Chr.21: 44288367-44288368 on Build GRCh38
rs75039713 Chr.21: 44288560-44288560 on Build GRCh38
rs377279425 Chr.21: 44288648-44288648 on Build GRCh38
rs745653031 Chr.21: 44288371-44288371 on Build GRCh38
rs768039280 Chr.21: 44288320-44288320 on Build GRCh38
rs41277546 Chr.21: 44288472-44288472 on Build GRCh38
rs1033319992 Chr.21: 44288688-44288688 on Build GRCh38
rs552196958 Chr.21: 44288740-44288740 on Build GRCh38
rs762238342 Chr.21: 44288640-44288640 on Build GRCh38
rs143784684 Chr.21: 44288421-44288421 on Build GRCh38
rs775297065 Chr.21: 44288386-44288386 on Build GRCh38
rs139969357 Chr.21: 44288571-44288571 on Build GRCh38
rs1039119580 Chr.21: 44288619-44288619 on Build GRCh38
rs372339585 Chr.21: 44288323-44288323 on Build GRCh38
rs1023438054 Chr.21: 44288643-44288643 on Build GRCh38
rs939162622 Chr.21: 44288307-44288307 on Build GRCh38
rs909450890 Chr.21: 44288733-44288733 on Build GRCh38
rs939556900 Chr.21: 44288525-44288525 on Build GRCh38
rs755490967 Chr.21: 44288413-44288413 on Build GRCh38
rs148012328 Chr.21: 44288395-44288395 on Build GRCh38
rs919460951 Chr.21: 44288717-44288717 on Build GRCh38
rs376083924 Chr.21: 44288400-44288400 on Build GRCh38
rs769510002 Chr.21: 44288430-44288430 on Build GRCh38
rs1043964850 Chr.21: 44288620-44288620 on Build GRCh38
rs558470210 Chr.21: 44288559-44288559 on Build GRCh38
rs766189603 Chr.21: 44288369-44288370 on Build GRCh38
rs1009936985 Chr.21: 44288661-44288661 on Build GRCh38
rs75809937 Chr.21: 44288690-44288690 on Build GRCh38
rs200359179 Chr.21: 44288414-44288414 on Build GRCh38

View Full Product Details