Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 44329601-44330097 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C21orf2
Entrez Gene ID
755
Gene Name
chromosome 21 open reading frame 2
Gene Aliases
LRRC76, YF5/A2
UniGene
Hs.517331
Gene Chromosome Location
Chr.21: 44328944-44339417 on Build GRCh38
Transcripts
NM_001271440.1 NM_001271441.1 NM_001271442.1 NM_004928.2 XM_006724051.3 XM_006724053.3 XM_017028470.1 XM_017028471.1 XM_017028472.1

SNP Detail

SNP ID Chromosome Location
rs534722715 Chr.21: 44329912-44329912 on Build GRCh38
rs911651292 Chr.21: 44330074-44330074 on Build GRCh38
rs1038822367 Chr.21: 44329864-44329864 on Build GRCh38
rs1025880256 Chr.21: 44329910-44329910 on Build GRCh38
rs1052717893 Chr.21: 44329651-44329653 on Build GRCh38
rs974582417 Chr.21: 44330011-44330011 on Build GRCh38
rs992963135 Chr.21: 44329812-44329812 on Build GRCh38
rs950440122 Chr.21: 44330052-44330052 on Build GRCh38
rs561098091 Chr.21: 44329669-44329669 on Build GRCh38
rs993120284 Chr.21: 44329640-44329640 on Build GRCh38
rs774845265 Chr.21: 44329780-44329780 on Build GRCh38
rs1011327315 Chr.21: 44329659-44329659 on Build GRCh38
rs73907169 Chr.21: 44329810-44329810 on Build GRCh38
rs964474372 Chr.21: 44329981-44329981 on Build GRCh38
rs913137201 Chr.21: 44330049-44330049 on Build GRCh38
rs530851137 Chr.21: 44329626-44329626 on Build GRCh38
rs983639203 Chr.21: 44329833-44329833 on Build GRCh38
rs569949897 Chr.21: 44329803-44329803 on Build GRCh38
rs891427444 Chr.21: 44329658-44329659 on Build GRCh38
rs1014444075 Chr.21: 44329989-44329989 on Build GRCh38
rs1027896877 Chr.21: 44330051-44330051 on Build GRCh38
rs766675596 Chr.21: 44329776-44329776 on Build GRCh38
rs551740814 Chr.21: 44329768-44329768 on Build GRCh38
rs1056689 Chr.21: 44329978-44329978 on Build GRCh38
rs73907170 Chr.21: 44329821-44329821 on Build GRCh38
rs768002305 Chr.21: 44329876-44329876 on Build GRCh38
rs923110058 Chr.21: 44330023-44330023 on Build GRCh38
rs369516551 Chr.21: 44329660-44329661 on Build GRCh38
rs139131768 Chr.21: 44329734-44329734 on Build GRCh38
rs1047453932 Chr.21: 44329701-44329701 on Build GRCh38
rs543710749 Chr.21: 44329831-44329831 on Build GRCh38
rs995126306 Chr.21: 44330043-44330043 on Build GRCh38
rs1056687 Chr.21: 44329962-44329962 on Build GRCh38
rs911609247 Chr.21: 44329847-44329847 on Build GRCh38
rs916157772 Chr.21: 44329645-44329645 on Build GRCh38
rs577858726 Chr.21: 44329965-44329965 on Build GRCh38
rs374967367 Chr.21: 44329827-44329827 on Build GRCh38
rs960218167 Chr.21: 44329627-44329627 on Build GRCh38
rs372930994 Chr.21: 44329904-44329904 on Build GRCh38
rs552792461 Chr.21: 44329920-44329920 on Build GRCh38
rs895860715 Chr.21: 44329980-44329980 on Build GRCh38
rs528601860 Chr.21: 44329694-44329694 on Build GRCh38
rs1035851733 Chr.21: 44329856-44329857 on Build GRCh38
rs890323245 Chr.21: 44329717-44329717 on Build GRCh38
rs928982772 Chr.21: 44329832-44329832 on Build GRCh38
rs998860290 Chr.21: 44329921-44329921 on Build GRCh38
rs929075520 Chr.21: 44329690-44329690 on Build GRCh38
rs753202185 Chr.21: 44330001-44330001 on Build GRCh38
rs920369924 Chr.21: 44329679-44329679 on Build GRCh38
rs944543095 Chr.21: 44329858-44329858 on Build GRCh38
rs546739223 Chr.21: 44329721-44329721 on Build GRCh38
rs999369920 Chr.21: 44329802-44329802 on Build GRCh38
rs115529052 Chr.21: 44330063-44330063 on Build GRCh38
rs1035966782 Chr.21: 44329696-44329696 on Build GRCh38
rs996823766 Chr.21: 44329746-44329746 on Build GRCh38
rs533156525 Chr.21: 44329739-44329739 on Build GRCh38
rs946666467 Chr.21: 44329891-44329891 on Build GRCh38
rs1023167787 Chr.21: 44329991-44329991 on Build GRCh38
rs973041664 Chr.21: 44329674-44329674 on Build GRCh38
rs1021554365 Chr.21: 44329789-44329789 on Build GRCh38
rs769714219 Chr.21: 44329642-44329642 on Build GRCh38
rs904511047 Chr.21: 44329790-44329790 on Build GRCh38
rs1021769233 Chr.21: 44329667-44329667 on Build GRCh38
rs181149547 Chr.21: 44329877-44329877 on Build GRCh38
rs1038966593 Chr.21: 44329638-44329638 on Build GRCh38
rs545183540 Chr.21: 44330024-44330024 on Build GRCh38
rs911270496 Chr.21: 44329757-44329757 on Build GRCh38
rs573366125 Chr.21: 44329853-44329854 on Build GRCh38
rs960274685 Chr.21: 44329705-44329705 on Build GRCh38
rs542989343 Chr.21: 44329655-44329655 on Build GRCh38
rs756662341 Chr.21: 44330013-44330013 on Build GRCh38
rs1023241528 Chr.21: 44329814-44329814 on Build GRCh38
rs773374302 Chr.21: 44329720-44329720 on Build GRCh38

View Full Product Details