Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 45175621-45176148 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADARB1
Entrez Gene ID
104
Gene Name
adenosine deaminase, RNA specific B1
Gene Aliases
ADAR2, DRABA2, DRADA2, RED1
UniGene
Hs.474018
Gene Chromosome Location
Chr.21: 45074578-45226563 on Build GRCh38
Transcripts
NM_001112.3 NM_001160230.1 NM_015833.3 NM_015834.3 NR_027672.1 NR_027673.1 NR_027674.1 NR_073200.1 XM_006723954.2 XM_011529425.2 XM_011529432.1 XM_017028242.1 XM_017028243.1 XM_017028244.1 XM_017028245.1 XM_017028246.1 XM_017028247.1 XM_017028248.1 XM_017028249.1 XM_017028250.1 XM_017028251.1 XM_017028252.1 XM_017028253.1 XM_017028254.1 XM_017028255.1 XM_017028256.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6594686 c.76T>C p.S26P Chr.21: 45175777-45175777 on Build GRCh38
5677322 c.134G>T p.G45V Chr.21: 45175835-45175835 on Build GRCh38
3551553 c.98C>T p.P33L Chr.21: 45175799-45175799 on Build GRCh38
3551557 c.302C>A p.T101N Chr.21: 45176003-45176003 on Build GRCh38
1414469 c.226C>T p.P76S Chr.21: 45175927-45175927 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs943839960 Chr.21: 45175849-45175849 on Build GRCh38
rs771605100 Chr.21: 45176098-45176098 on Build GRCh38
rs150871717 Chr.21: 45176007-45176007 on Build GRCh38
rs2838798 Chr.21: 45175929-45175929 on Build GRCh38
rs760175149 Chr.21: 45176110-45176110 on Build GRCh38
rs150318827 Chr.21: 45175898-45175898 on Build GRCh38
rs777322692 Chr.21: 45175860-45175860 on Build GRCh38
rs763402307 Chr.21: 45176070-45176070 on Build GRCh38
rs747059653 Chr.21: 45176012-45176012 on Build GRCh38
rs779762821 Chr.21: 45175707-45175707 on Build GRCh38
rs931367429 Chr.21: 45175686-45175686 on Build GRCh38
rs147822904 Chr.21: 45175871-45175871 on Build GRCh38
rs749115732 Chr.21: 45175708-45175708 on Build GRCh38
rs201038678 Chr.21: 45175869-45175869 on Build GRCh38
rs766273696 Chr.21: 45175914-45175914 on Build GRCh38
rs747116737 Chr.21: 45175848-45175848 on Build GRCh38
rs374872814 Chr.21: 45176115-45176115 on Build GRCh38
rs771515782 Chr.21: 45175854-45175854 on Build GRCh38
rs972299463 Chr.21: 45175671-45175671 on Build GRCh38
rs779992203 Chr.21: 45175786-45175786 on Build GRCh38
rs200269366 Chr.21: 45175829-45175829 on Build GRCh38
rs766882884 Chr.21: 45175779-45175779 on Build GRCh38
rs143261942 Chr.21: 45176069-45176069 on Build GRCh38
rs148374145 Chr.21: 45175936-45175936 on Build GRCh38
rs775199908 Chr.21: 45175703-45175704 on Build GRCh38
rs760230700 Chr.21: 45175909-45175909 on Build GRCh38
rs754756681 Chr.21: 45175937-45175937 on Build GRCh38
rs751028540 Chr.21: 45175687-45175687 on Build GRCh38
rs918713208 Chr.21: 45175684-45175684 on Build GRCh38
rs765249261 Chr.21: 45176121-45176121 on Build GRCh38
rs745476919 Chr.21: 45175983-45175983 on Build GRCh38
rs1001134517 Chr.21: 45175645-45175645 on Build GRCh38
rs770342145 Chr.21: 45176032-45176032 on Build GRCh38
rs141594032 Chr.21: 45175804-45175804 on Build GRCh38
rs778516155 Chr.21: 45175938-45175938 on Build GRCh38
rs758507445 Chr.21: 45175993-45175993 on Build GRCh38
rs201356458 Chr.21: 45176019-45176019 on Build GRCh38
rs774487150 Chr.21: 45175724-45175724 on Build GRCh38
rs759943630 Chr.21: 45175868-45175868 on Build GRCh38
rs753361667 Chr.21: 45176118-45176118 on Build GRCh38
rs919946042 Chr.21: 45175809-45175809 on Build GRCh38
rs773097471 Chr.21: 45175837-45175837 on Build GRCh38
rs771170713 Chr.21: 45176017-45176017 on Build GRCh38
rs201786934 Chr.21: 45175895-45175895 on Build GRCh38
rs768820156 Chr.21: 45175714-45175714 on Build GRCh38
rs1034977318 Chr.21: 45176097-45176097 on Build GRCh38
rs879149351 Chr.21: 45176011-45176011 on Build GRCh38
rs201653105 Chr.21: 45175777-45175777 on Build GRCh38
rs777093797 Chr.21: 45175763-45175763 on Build GRCh38
rs763812559 Chr.21: 45175774-45175774 on Build GRCh38
rs779194116 Chr.21: 45175810-45175810 on Build GRCh38
rs759998474 Chr.21: 45175770-45175770 on Build GRCh38
rs749061640 Chr.21: 45175792-45175792 on Build GRCh38
rs892741158 Chr.21: 45176073-45176073 on Build GRCh38
rs748955652 Chr.21: 45175769-45175771 on Build GRCh38
rs370431644 Chr.21: 45175794-45175794 on Build GRCh38
rs956880780 Chr.21: 45175662-45175662 on Build GRCh38
rs367618294 Chr.21: 45176119-45176119 on Build GRCh38
rs772893086 Chr.21: 45176099-45176099 on Build GRCh38
rs115658841 Chr.21: 45176010-45176010 on Build GRCh38
rs375447640 Chr.21: 45175683-45175683 on Build GRCh38
rs375341807 Chr.21: 45175760-45175760 on Build GRCh38
rs756785178 Chr.21: 45175778-45175778 on Build GRCh38
rs370534548 Chr.21: 45175651-45175651 on Build GRCh38
rs745855846 Chr.21: 45175694-45175694 on Build GRCh38
rs756763995 Chr.21: 45175693-45175693 on Build GRCh38
rs35259738 Chr.21: 45176007-45176008 on Build GRCh38
rs143795052 Chr.21: 45175897-45175897 on Build GRCh38
rs942964451 Chr.21: 45175738-45175738 on Build GRCh38
rs908348394 Chr.21: 45175847-45175847 on Build GRCh38
rs748231046 Chr.21: 45175733-45175733 on Build GRCh38
rs78867942 Chr.21: 45176044-45176044 on Build GRCh38
rs756062092 Chr.21: 45175695-45175695 on Build GRCh38
rs766996044 Chr.21: 45175762-45175762 on Build GRCh38
rs775680938 Chr.21: 45176046-45176046 on Build GRCh38
rs576621164 Chr.21: 45175870-45175870 on Build GRCh38
rs781762879 Chr.21: 45176018-45176018 on Build GRCh38
rs144363543 Chr.21: 45175835-45175835 on Build GRCh38
rs142180298 Chr.21: 45175759-45175759 on Build GRCh38
rs146833412 Chr.21: 45176058-45176058 on Build GRCh38
rs1013733289 Chr.21: 45175669-45175669 on Build GRCh38
rs36102117 Chr.21: 45175979-45175980 on Build GRCh38
rs756010542 Chr.21: 45175782-45175782 on Build GRCh38
rs749956447 Chr.21: 45175781-45175781 on Build GRCh38
rs1031562608 Chr.21: 45175657-45175657 on Build GRCh38
rs141269164 Chr.21: 45175880-45175880 on Build GRCh38
rs199805536 Chr.21: 45176062-45176062 on Build GRCh38

View Full Product Details