Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 24011045-24011569 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CABIN1
Entrez Gene ID
23523
Gene Name
calcineurin binding protein 1
Gene Aliases
CAIN, KB-318B8.7, PPP3IN
UniGene
Hs.517478
Gene Chromosome Location
Chr.22: 24011192-24178628 on Build GRCh38
Transcripts
NM_001199281.1 NM_001201429.1 NM_012295.3 XM_005261419.2

SNP Detail

SNP ID Chromosome Location
rs1041889823 Chr.22: 24011107-24011107 on Build GRCh38
rs886168426 Chr.22: 24011069-24011069 on Build GRCh38
rs531178583 Chr.22: 24011246-24011246 on Build GRCh38
rs934505142 Chr.22: 24011162-24011162 on Build GRCh38
rs918165159 Chr.22: 24011329-24011329 on Build GRCh38
rs199531771 Chr.22: 24011157-24011158 on Build GRCh38
rs558617520 Chr.22: 24011197-24011197 on Build GRCh38
rs949687806 Chr.22: 24011335-24011335 on Build GRCh38
rs368701182 Chr.22: 24011304-24011304 on Build GRCh38
rs945155172 Chr.22: 24011167-24011167 on Build GRCh38
rs766340040 Chr.22: 24011411-24011411 on Build GRCh38
rs372125909 Chr.22: 24011434-24011434 on Build GRCh38
rs1009269223 Chr.22: 24011103-24011103 on Build GRCh38
rs762262820 Chr.22: 24011085-24011086 on Build GRCh38
rs765277924 Chr.22: 24011142-24011151 on Build GRCh38
rs753602922 Chr.22: 24011140-24011143 on Build GRCh38
rs115068404 Chr.22: 24011163-24011163 on Build GRCh38
rs759528466 Chr.22: 24011442-24011442 on Build GRCh38
rs532114113 Chr.22: 24011312-24011312 on Build GRCh38
rs536881709 Chr.22: 24011488-24011488 on Build GRCh38
rs868339314 Chr.22: 24011105-24011105 on Build GRCh38
rs960096137 Chr.22: 24011231-24011231 on Build GRCh38
rs983029960 Chr.22: 24011319-24011319 on Build GRCh38
rs915572117 Chr.22: 24011134-24011141 on Build GRCh38
rs368608686 Chr.22: 24011109-24011109 on Build GRCh38
rs927027749 Chr.22: 24011383-24011383 on Build GRCh38
rs199660607 Chr.22: 24011154-24011157 on Build GRCh38
rs974627650 Chr.22: 24011240-24011240 on Build GRCh38
rs565469346 Chr.22: 24011161-24011161 on Build GRCh38
rs959780589 Chr.22: 24011103-24011104 on Build GRCh38
rs899815810 Chr.22: 24011196-24011196 on Build GRCh38
rs1041603682 Chr.22: 24011446-24011446 on Build GRCh38
rs770266641 Chr.22: 24011077-24011086 on Build GRCh38
rs73879031 Chr.22: 24011149-24011149 on Build GRCh38
rs7290013 Chr.22: 24011135-24011135 on Build GRCh38
rs752853864 Chr.22: 24011143-24011144 on Build GRCh38
rs891084662 Chr.22: 24011450-24011450 on Build GRCh38
rs772131458 Chr.22: 24011142-24011157 on Build GRCh38
rs867683306 Chr.22: 24011095-24011095 on Build GRCh38
rs867892938 Chr.22: 24011111-24011111 on Build GRCh38
rs1014346044 Chr.22: 24011430-24011430 on Build GRCh38
rs150112712 Chr.22: 24011131-24011131 on Build GRCh38
rs989084443 Chr.22: 24011330-24011330 on Build GRCh38
rs773495452 Chr.22: 24011083-24011086 on Build GRCh38
rs71866862 Chr.22: 24011077-24011078 on Build GRCh38
rs542401670 Chr.22: 24011227-24011227 on Build GRCh38
rs778277088 Chr.22: 24011158-24011163 on Build GRCh38
rs978403652 Chr.22: 24011436-24011436 on Build GRCh38
rs71320695 Chr.22: 24011165-24011165 on Build GRCh38
rs957350066 Chr.22: 24011099-24011099 on Build GRCh38
rs184631484 Chr.22: 24011207-24011207 on Build GRCh38
rs979369650 Chr.22: 24011152-24011173 on Build GRCh38
rs547926418 Chr.22: 24011441-24011441 on Build GRCh38
rs892948524 Chr.22: 24011433-24011433 on Build GRCh38
rs988725319 Chr.22: 24011504-24011505 on Build GRCh38
rs575001659 Chr.22: 24011222-24011222 on Build GRCh38
rs896274006 Chr.22: 24011101-24011101 on Build GRCh38
rs968970367 Chr.22: 24011148-24011167 on Build GRCh38
rs565393029 Chr.22: 24011408-24011408 on Build GRCh38
rs75568443 Chr.22: 24011147-24011147 on Build GRCh38
rs983363591 Chr.22: 24011542-24011542 on Build GRCh38
rs757832895 Chr.22: 24011158-24011175 on Build GRCh38
rs189903013 Chr.22: 24011509-24011509 on Build GRCh38
rs558793565 Chr.22: 24011508-24011508 on Build GRCh38
rs1022611333 Chr.22: 24011272-24011272 on Build GRCh38
rs7290007 Chr.22: 24011123-24011123 on Build GRCh38
rs140642551 Chr.22: 24011175-24011175 on Build GRCh38
rs1027503729 Chr.22: 24011520-24011520 on Build GRCh38
rs1054320527 Chr.22: 24011198-24011198 on Build GRCh38
rs553475428 Chr.22: 24011208-24011208 on Build GRCh38
rs537347193 Chr.22: 24011177-24011177 on Build GRCh38
rs992535886 Chr.22: 24011245-24011245 on Build GRCh38
rs7290008 Chr.22: 24011129-24011129 on Build GRCh38
rs1041104366 Chr.22: 24011180-24011180 on Build GRCh38
rs899890780 Chr.22: 24011368-24011368 on Build GRCh38
rs1008241647 Chr.22: 24011225-24011225 on Build GRCh38
rs1041138923 Chr.22: 24011340-24011340 on Build GRCh38
rs541427443 Chr.22: 24011464-24011464 on Build GRCh38
rs1047834737 Chr.22: 24011168-24011168 on Build GRCh38
rs1045413048 Chr.22: 24011376-24011376 on Build GRCh38
rs71312989 Chr.22: 24011119-24011120 on Build GRCh38
rs888705216 Chr.22: 24011397-24011397 on Build GRCh38
rs530294817 Chr.22: 24011097-24011097 on Build GRCh38
rs201160099 Chr.22: 24011139-24011140 on Build GRCh38
rs920134224 Chr.22: 24011292-24011292 on Build GRCh38
rs951540587 Chr.22: 24011531-24011531 on Build GRCh38
rs538298606 Chr.22: 24011152-24011152 on Build GRCh38
rs372186531 Chr.22: 24011131-24011132 on Build GRCh38
rs1054628449 Chr.22: 24011427-24011427 on Build GRCh38
rs879008432 Chr.22: 24011170-24011173 on Build GRCh38
rs368371389 Chr.22: 24011146-24011169 on Build GRCh38
rs932658112 Chr.22: 24011373-24011373 on Build GRCh38
rs1024020387 Chr.22: 24011076-24011076 on Build GRCh38
rs55821428 Chr.22: 24011169-24011169 on Build GRCh38
rs777868359 Chr.22: 24011142-24011143 on Build GRCh38
rs577152520 Chr.22: 24011203-24011203 on Build GRCh38
rs920167414 Chr.22: 24011165-24011165 on Build GRCh38
rs1008226811 Chr.22: 24011457-24011457 on Build GRCh38
rs755051824 Chr.22: 24011139-24011142 on Build GRCh38
rs879372288 Chr.22: 24011150-24011165 on Build GRCh38
rs556334424 Chr.22: 24011157-24011157 on Build GRCh38
rs571219823 Chr.22: 24011151-24011151 on Build GRCh38
rs905680623 Chr.22: 24011080-24011080 on Build GRCh38
rs200995428 Chr.22: 24011173-24011174 on Build GRCh38
rs28693002 Chr.22: 24011115-24011115 on Build GRCh38
rs879525548 Chr.22: 24011134-24011143 on Build GRCh38
rs1001327144 Chr.22: 24011091-24011091 on Build GRCh38
rs984744475 Chr.22: 24011145-24011145 on Build GRCh38
rs111718395 Chr.22: 24011342-24011342 on Build GRCh38
rs563876347 Chr.22: 24011237-24011237 on Build GRCh38
rs748017941 Chr.22: 24011505-24011505 on Build GRCh38
rs535560626 Chr.22: 24011171-24011171 on Build GRCh38
rs868350806 Chr.22: 24011113-24011113 on Build GRCh38
rs969707449 Chr.22: 24011283-24011283 on Build GRCh38
rs972124522 Chr.22: 24011133-24011133 on Build GRCh38
rs905654593 Chr.22: 24011192-24011192 on Build GRCh38
rs1013986436 Chr.22: 24011071-24011071 on Build GRCh38
rs760275590 Chr.22: 24011136-24011147 on Build GRCh38
rs546469411 Chr.22: 24011277-24011277 on Build GRCh38
rs146242394 Chr.22: 24011141-24011142 on Build GRCh38
rs944142979 Chr.22: 24011337-24011337 on Build GRCh38
rs879129919 Chr.22: 24011155-24011155 on Build GRCh38
rs547102428 Chr.22: 24011346-24011346 on Build GRCh38
rs11913855 Chr.22: 24011137-24011137 on Build GRCh38
rs951944540 Chr.22: 24011143-24011143 on Build GRCh38
rs753759013 Chr.22: 24011415-24011415 on Build GRCh38
rs11914076 Chr.22: 24011159-24011159 on Build GRCh38
rs1022686534 Chr.22: 24011431-24011431 on Build GRCh38
rs974270618 Chr.22: 24011154-24011154 on Build GRCh38
rs58506551 Chr.22: 24011116-24011117 on Build GRCh38
rs373060220 Chr.22: 24011170-24011171 on Build GRCh38
rs1020640793 Chr.22: 24011209-24011209 on Build GRCh38
rs7290037 Chr.22: 24011173-24011173 on Build GRCh38
rs57294943 Chr.22: 24011144-24011145 on Build GRCh38
rs548821638 Chr.22: 24011125-24011125 on Build GRCh38
rs796614720 Chr.22: 24011404-24011404 on Build GRCh38
rs1025172975 Chr.22: 24011547-24011547 on Build GRCh38
rs563937656 Chr.22: 24011127-24011127 on Build GRCh38

View Full Product Details