Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 28696805-28697344 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHEK2
Entrez Gene ID
11200
Gene Name
checkpoint kinase 2
Gene Aliases
CDS1, CHK2, HuCds1, LFS2, PP1425, RAD53, hCds1
UniGene
Hs.291363
Gene Chromosome Location
Chr.22: 28687743-28741866 on Build GRCh38
Transcripts
NM_001005735.1 NM_001257387.1 NM_007194.3 NM_145862.2 XM_006724114.3 XM_006724116.2 XM_011529839.2 XM_011529840.2 XM_011529841.1 XM_011529842.2 XM_011529843.1 XM_011529844.2 XM_011529845.2 XM_017028560.1 XM_017028561.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5983821 c.1207G>C p.E403Q Chr.22: 28696918-28696918 on Build GRCh38
6942269 c.1067C>T p.S356L Chr.22: 28696929-28696929 on Build GRCh38
7346458 c.1166G>A p.R389H Chr.22: 28696959-28696959 on Build GRCh38
6919729 c.1165C>A p.R389S Chr.22: 28696960-28696960 on Build GRCh38
4428443 c.1042T>C p.L348L Chr.22: 28696954-28696954 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769631429 Chr.22: 28696878-28696878 on Build GRCh38
rs901286019 Chr.22: 28697203-28697203 on Build GRCh38
rs189822221 Chr.22: 28697318-28697318 on Build GRCh38
rs774179198 Chr.22: 28696905-28696905 on Build GRCh38
rs1007907219 Chr.22: 28697077-28697077 on Build GRCh38
rs200020484 Chr.22: 28696882-28696882 on Build GRCh38
rs980053804 Chr.22: 28697167-28697167 on Build GRCh38
rs568693812 Chr.22: 28697161-28697161 on Build GRCh38
rs9625534 Chr.22: 28697290-28697290 on Build GRCh38
rs730881705 Chr.22: 28696972-28696972 on Build GRCh38
rs926134174 Chr.22: 28697179-28697179 on Build GRCh38
rs763174055 Chr.22: 28696994-28696994 on Build GRCh38
rs946179247 Chr.22: 28697007-28697007 on Build GRCh38
rs768172525 Chr.22: 28696900-28696900 on Build GRCh38
rs587782268 Chr.22: 28696943-28696943 on Build GRCh38
rs730881689 Chr.22: 28696951-28696951 on Build GRCh38
rs747655283 Chr.22: 28696977-28696977 on Build GRCh38
rs748898751 Chr.22: 28697023-28697023 on Build GRCh38
rs876659921 Chr.22: 28696903-28696903 on Build GRCh38
rs746134647 Chr.22: 28697169-28697169 on Build GRCh38
rs555234460 Chr.22: 28697009-28697009 on Build GRCh38
rs572668197 Chr.22: 28696913-28696913 on Build GRCh38
rs141502354 Chr.22: 28696969-28696969 on Build GRCh38
rs878854909 Chr.22: 28696924-28696924 on Build GRCh38
rs992427733 Chr.22: 28697123-28697123 on Build GRCh38
rs778518211 Chr.22: 28697128-28697128 on Build GRCh38
rs559280794 Chr.22: 28697204-28697204 on Build GRCh38
rs876658337 Chr.22: 28696918-28696918 on Build GRCh38
rs763381213 Chr.22: 28697303-28697303 on Build GRCh38
rs781640725 Chr.22: 28696866-28696866 on Build GRCh38
rs121908703 Chr.22: 28696929-28696929 on Build GRCh38
rs201206424 Chr.22: 28696960-28696960 on Build GRCh38
rs532199701 Chr.22: 28697092-28697092 on Build GRCh38
rs766158073 Chr.22: 28696989-28696989 on Build GRCh38
rs945819379 Chr.22: 28697250-28697250 on Build GRCh38
rs1028944988 Chr.22: 28697070-28697070 on Build GRCh38
rs540691865 Chr.22: 28697199-28697199 on Build GRCh38
rs786202893 Chr.22: 28696944-28696944 on Build GRCh38
rs771209375 Chr.22: 28697149-28697149 on Build GRCh38
rs773846607 Chr.22: 28696974-28696974 on Build GRCh38
rs869312554 Chr.22: 28697153-28697153 on Build GRCh38
rs371207635 Chr.22: 28696981-28696981 on Build GRCh38
rs771442272 Chr.22: 28696853-28696853 on Build GRCh38
rs757393834 Chr.22: 28697005-28697005 on Build GRCh38
rs864622537 Chr.22: 28696963-28696963 on Build GRCh38
rs28909980 Chr.22: 28696957-28696957 on Build GRCh38
rs764083004 Chr.22: 28696998-28696998 on Build GRCh38
rs374660293 Chr.22: 28696983-28696983 on Build GRCh38
rs4820790 Chr.22: 28697251-28697251 on Build GRCh38
rs876659406 Chr.22: 28696948-28696948 on Build GRCh38
rs755664409 Chr.22: 28697039-28697039 on Build GRCh38
rs919032799 Chr.22: 28697091-28697091 on Build GRCh38
rs749233029 Chr.22: 28697084-28697084 on Build GRCh38
rs377668478 Chr.22: 28696973-28696973 on Build GRCh38
rs781368998 Chr.22: 28697034-28697034 on Build GRCh38
rs757222003 Chr.22: 28697009-28697010 on Build GRCh38
rs1053173462 Chr.22: 28697201-28697201 on Build GRCh38
rs765425451 Chr.22: 28696926-28696926 on Build GRCh38
rs767306337 Chr.22: 28696911-28696911 on Build GRCh38
rs1030239691 Chr.22: 28697032-28697032 on Build GRCh38
rs780408916 Chr.22: 28697017-28697017 on Build GRCh38
rs760449049 Chr.22: 28696920-28696920 on Build GRCh38
rs199859140 Chr.22: 28696915-28696915 on Build GRCh38
rs748002165 Chr.22: 28697131-28697131 on Build GRCh38
rs962169511 Chr.22: 28697105-28697105 on Build GRCh38
rs786201130 Chr.22: 28696933-28696933 on Build GRCh38
rs202051128 Chr.22: 28696964-28696964 on Build GRCh38
rs878854908 Chr.22: 28696953-28696953 on Build GRCh38
rs369497542 Chr.22: 28696869-28696869 on Build GRCh38
rs576700526 Chr.22: 28697027-28697027 on Build GRCh38
rs889786497 Chr.22: 28697260-28697260 on Build GRCh38
rs758434121 Chr.22: 28696938-28696938 on Build GRCh38
rs543588734 Chr.22: 28697082-28697082 on Build GRCh38
rs938389349 Chr.22: 28696931-28696931 on Build GRCh38
rs587782441 Chr.22: 28696984-28696984 on Build GRCh38
rs730881688 Chr.22: 28696959-28696959 on Build GRCh38
rs786202676 Chr.22: 28696956-28696956 on Build GRCh38
rs527300711 Chr.22: 28697172-28697172 on Build GRCh38
rs932235706 Chr.22: 28697304-28697304 on Build GRCh38
rs762414562 Chr.22: 28696889-28696889 on Build GRCh38
rs775593362 Chr.22: 28696993-28696993 on Build GRCh38
rs756520206 Chr.22: 28696961-28696961 on Build GRCh38
rs926927207 Chr.22: 28697180-28697180 on Build GRCh38
rs751749563 Chr.22: 28697003-28697003 on Build GRCh38
rs761681864 Chr.22: 28696907-28696907 on Build GRCh38
rs564832288 Chr.22: 28697085-28697085 on Build GRCh38
rs779769892 Chr.22: 28697079-28697079 on Build GRCh38
rs760502479 Chr.22: 28696985-28696985 on Build GRCh38
rs863224746 Chr.22: 28696968-28696968 on Build GRCh38
rs377730816 Chr.22: 28697016-28697016 on Build GRCh38
rs764181318 Chr.22: 28696941-28696941 on Build GRCh38

View Full Product Details