Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 28783369-28783888 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC117
Entrez Gene ID
150275
Gene Name
coiled-coil domain containing 117
Gene Aliases
dJ366L4.1
UniGene
Hs.406460
Gene Chromosome Location
Chr.22: 28772674-28789301 on Build GRCh38
Transcripts
NM_001284263.1 NM_001284264.1 NM_001284265.1 NM_173510.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
279615 c.590T>G p.M197R Chr.22: 28783633-28783633 on Build GRCh38
6297318 c.526G>T p.V176F Chr.22: 28783569-28783569 on Build GRCh38
350301 c.472G>A p.D158N Chr.22: 28783515-28783515 on Build GRCh38
6004928 c.497A>G p.D166G Chr.22: 28783540-28783540 on Build GRCh38
2936026 c.470T>G p.I157S Chr.22: 28783513-28783513 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs901141327 Chr.22: 28783822-28783822 on Build GRCh38
rs757306032 Chr.22: 28783806-28783806 on Build GRCh38
rs201539039 Chr.22: 28783576-28783576 on Build GRCh38
rs747526714 Chr.22: 28783480-28783480 on Build GRCh38
rs948813337 Chr.22: 28783446-28783446 on Build GRCh38
rs749911759 Chr.22: 28783600-28783600 on Build GRCh38
rs759257197 Chr.22: 28783714-28783714 on Build GRCh38
rs370186831 Chr.22: 28783669-28783669 on Build GRCh38
rs753597791 Chr.22: 28783611-28783611 on Build GRCh38
rs747298850 Chr.22: 28783625-28783625 on Build GRCh38
rs750294956 Chr.22: 28783486-28783486 on Build GRCh38
rs2347447 Chr.22: 28783751-28783751 on Build GRCh38
rs761500029 Chr.22: 28783694-28783694 on Build GRCh38
rs756044555 Chr.22: 28783601-28783601 on Build GRCh38
rs926069114 Chr.22: 28783799-28783799 on Build GRCh38
rs781565758 Chr.22: 28783515-28783520 on Build GRCh38
rs990625969 Chr.22: 28783596-28783596 on Build GRCh38
rs772214168 Chr.22: 28783493-28783493 on Build GRCh38
rs756030945 Chr.22: 28783487-28783487 on Build GRCh38
rs935528913 Chr.22: 28783699-28783699 on Build GRCh38
rs17854014 Chr.22: 28783616-28783616 on Build GRCh38
rs889036433 Chr.22: 28783857-28783857 on Build GRCh38
rs867261813 Chr.22: 28783841-28783841 on Build GRCh38
rs574208461 Chr.22: 28783468-28783468 on Build GRCh38
rs770315729 Chr.22: 28783538-28783538 on Build GRCh38
rs558330098 Chr.22: 28783562-28783562 on Build GRCh38
rs779760624 Chr.22: 28783490-28783490 on Build GRCh38
rs756707922 Chr.22: 28783471-28783471 on Build GRCh38
rs963368410 Chr.22: 28783567-28783567 on Build GRCh38
rs149747242 Chr.22: 28783644-28783644 on Build GRCh38
rs997285728 Chr.22: 28783455-28783455 on Build GRCh38
rs767083565 Chr.22: 28783578-28783578 on Build GRCh38
rs1054202501 Chr.22: 28783700-28783700 on Build GRCh38
rs201017004 Chr.22: 28783695-28783695 on Build GRCh38
rs117327764 Chr.22: 28783839-28783839 on Build GRCh38
rs997323214 Chr.22: 28783787-28783787 on Build GRCh38
rs577588852 Chr.22: 28783726-28783726 on Build GRCh38
rs1030219373 Chr.22: 28783458-28783458 on Build GRCh38
rs140909131 Chr.22: 28783535-28783535 on Build GRCh38
rs747096161 Chr.22: 28783503-28783503 on Build GRCh38
rs79919091 Chr.22: 28783850-28783850 on Build GRCh38
rs948929030 Chr.22: 28783770-28783770 on Build GRCh38
rs769000146 Chr.22: 28783676-28783676 on Build GRCh38
rs781122623 Chr.22: 28783473-28783473 on Build GRCh38
rs894618105 Chr.22: 28783413-28783413 on Build GRCh38
rs904613567 Chr.22: 28783494-28783494 on Build GRCh38
rs763240602 Chr.22: 28783672-28783672 on Build GRCh38
rs532159091 Chr.22: 28783805-28783805 on Build GRCh38
rs760180713 Chr.22: 28783536-28783536 on Build GRCh38
rs371409488 Chr.22: 28783492-28783492 on Build GRCh38
rs780001169 Chr.22: 28783603-28783603 on Build GRCh38
rs775761135 Chr.22: 28783555-28783555 on Build GRCh38
rs777859084 Chr.22: 28783617-28783617 on Build GRCh38
rs778184539 Chr.22: 28783477-28783480 on Build GRCh38
rs970916225 Chr.22: 28783653-28783653 on Build GRCh38
rs774696290 Chr.22: 28783693-28783693 on Build GRCh38
rs771064464 Chr.22: 28783519-28783519 on Build GRCh38
rs553628807 Chr.22: 28783495-28783498 on Build GRCh38
rs915278273 Chr.22: 28783705-28783705 on Build GRCh38
rs373277116 Chr.22: 28783459-28783459 on Build GRCh38
rs140438864 Chr.22: 28783659-28783659 on Build GRCh38
rs879190599 Chr.22: 28783508-28783508 on Build GRCh38
rs971778272 Chr.22: 28783708-28783708 on Build GRCh38
rs776435860 Chr.22: 28783707-28783707 on Build GRCh38
rs916081266 Chr.22: 28783762-28783762 on Build GRCh38
rs766255694 Chr.22: 28783696-28783696 on Build GRCh38
rs773258300 Chr.22: 28783522-28783522 on Build GRCh38
rs1057317301 Chr.22: 28783400-28783400 on Build GRCh38
rs769954968 Chr.22: 28783527-28783532 on Build GRCh38
rs551870030 Chr.22: 28783813-28783813 on Build GRCh38
rs560174394 Chr.22: 28783782-28783782 on Build GRCh38
rs369776611 Chr.22: 28783654-28783654 on Build GRCh38
rs777999369 Chr.22: 28783495-28783495 on Build GRCh38
rs751156576 Chr.22: 28783565-28783565 on Build GRCh38
rs751841963 Chr.22: 28783771-28783771 on Build GRCh38
rs369357032 Chr.22: 28783634-28783634 on Build GRCh38
rs960332010 Chr.22: 28783736-28783736 on Build GRCh38
rs746066434 Chr.22: 28783524-28783526 on Build GRCh38

View Full Product Details