Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 88953084-88953634 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KYAT3
Entrez Gene ID
56267
Gene Name
kynurenine aminotransferase 3
Gene Aliases
CCBL2, KAT3, KATIII
UniGene
Hs.481898
Gene Chromosome Location
Chr.1: 88935773-88992960 on Build GRCh38
Transcripts
NM_001008661.2 NM_001008662.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1162228 c.875C>T p.S292F Chr.1: 88953142-88953142 on Build GRCh38
1162229 c.773C>T p.S258F Chr.1: 88953142-88953142 on Build GRCh38
6376045 c.785A>G p.N262S Chr.1: 88953130-88953130 on Build GRCh38
682803 c.870C>G p.G290G Chr.1: 88953147-88953147 on Build GRCh38
1344715 c.905T>G p.L302* Chr.1: 88953112-88953112 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750512916 Chr.1: 88953237-88953237 on Build GRCh38
rs755346938 Chr.1: 88953357-88953357 on Build GRCh38
rs767990018 Chr.1: 88953392-88953392 on Build GRCh38
rs12740700 Chr.1: 88953407-88953407 on Build GRCh38
rs368642853 Chr.1: 88953111-88953111 on Build GRCh38
rs9428016 Chr.1: 88953160-88953160 on Build GRCh38
rs190057915 Chr.1: 88953303-88953303 on Build GRCh38
rs67537454 Chr.1: 88953546-88953546 on Build GRCh38
rs912264684 Chr.1: 88953228-88953228 on Build GRCh38
rs1000128092 Chr.1: 88953351-88953351 on Build GRCh38
rs950021459 Chr.1: 88953325-88953325 on Build GRCh38
rs201974256 Chr.1: 88953162-88953162 on Build GRCh38
rs370389170 Chr.1: 88953589-88953589 on Build GRCh38
rs1054262644 Chr.1: 88953366-88953366 on Build GRCh38
rs995777958 Chr.1: 88953462-88953462 on Build GRCh38
rs770644022 Chr.1: 88953132-88953139 on Build GRCh38
rs199612220 Chr.1: 88953546-88953547 on Build GRCh38
rs10922527 Chr.1: 88953311-88953311 on Build GRCh38
rs397980755 Chr.1: 88953566-88953566 on Build GRCh38
rs910051795 Chr.1: 88953271-88953271 on Build GRCh38
rs796419288 Chr.1: 88953148-88953148 on Build GRCh38
rs750738467 Chr.1: 88953181-88953181 on Build GRCh38
rs965681205 Chr.1: 88953193-88953193 on Build GRCh38
rs137937024 Chr.1: 88953608-88953609 on Build GRCh38
rs1044389383 Chr.1: 88953573-88953573 on Build GRCh38
rs974414240 Chr.1: 88953231-88953231 on Build GRCh38
rs763021096 Chr.1: 88953244-88953244 on Build GRCh38
rs181758792 Chr.1: 88953485-88953485 on Build GRCh38
rs369650800 Chr.1: 88953524-88953524 on Build GRCh38
rs796673884 Chr.1: 88953412-88953413 on Build GRCh38
rs762977611 Chr.1: 88953110-88953110 on Build GRCh38
rs67571765 Chr.1: 88953390-88953390 on Build GRCh38
rs761112298 Chr.1: 88953182-88953182 on Build GRCh38
rs868215772 Chr.1: 88953600-88953600 on Build GRCh38
rs576582851 Chr.1: 88953232-88953232 on Build GRCh38
rs767847308 Chr.1: 88953168-88953168 on Build GRCh38
rs984786452 Chr.1: 88953477-88953477 on Build GRCh38
rs954315486 Chr.1: 88953238-88953238 on Build GRCh38
rs1028632457 Chr.1: 88953473-88953473 on Build GRCh38
rs184712364 Chr.1: 88953505-88953505 on Build GRCh38
rs969257916 Chr.1: 88953135-88953135 on Build GRCh38
rs775980738 Chr.1: 88953547-88953548 on Build GRCh38
rs972957013 Chr.1: 88953510-88953510 on Build GRCh38
rs919864059 Chr.1: 88953268-88953268 on Build GRCh38
rs774459748 Chr.1: 88953141-88953141 on Build GRCh38
rs965795971 Chr.1: 88953438-88953438 on Build GRCh38
rs151248715 Chr.1: 88953412-88953412 on Build GRCh38
rs112972487 Chr.1: 88953186-88953186 on Build GRCh38
rs917377477 Chr.1: 88953321-88953321 on Build GRCh38
rs1010246595 Chr.1: 88953362-88953362 on Build GRCh38
rs952686221 Chr.1: 88953567-88953567 on Build GRCh38
rs753314332 Chr.1: 88953458-88953458 on Build GRCh38
rs140607347 Chr.1: 88953114-88953114 on Build GRCh38
rs931113843 Chr.1: 88953273-88953273 on Build GRCh38
rs779883303 Chr.1: 88953546-88953548 on Build GRCh38
rs954264241 Chr.1: 88953475-88953475 on Build GRCh38
rs376578285 Chr.1: 88953191-88953191 on Build GRCh38
rs140483656 Chr.1: 88953413-88953413 on Build GRCh38
rs1044440398 Chr.1: 88953338-88953338 on Build GRCh38
rs927287697 Chr.1: 88953596-88953596 on Build GRCh38
rs567423989 Chr.1: 88953587-88953587 on Build GRCh38
rs779761953 Chr.1: 88953340-88953340 on Build GRCh38
rs532543529 Chr.1: 88953395-88953395 on Build GRCh38
rs950131970 Chr.1: 88953570-88953570 on Build GRCh38

View Full Product Details