Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 92247754-92248297 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLMN
Entrez Gene ID
11146
Gene Name
glomulin, FKBP associated protein
Gene Aliases
FAP, FAP48, FAP68, FKBPAP, GLML, GVM, VMGLOM
UniGene
Hs.49105
Gene Chromosome Location
Chr.1: 92245786-92441936 on Build GRCh38
Transcripts
NM_001319683.1 NM_053274.2 NR_135089.1 XM_005270401.3 XM_006710309.2 XM_011540546.2 XM_017000137.1 XM_017000138.1 XM_017000139.1 XM_017000140.1 XM_017000141.1 XM_017000142.1 XM_017000143.1 XM_017000144.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6401924 c.1555C>T p.H519Y Chr.1: 92247908-92247908 on Build GRCh38
1627252 c.1488A>G p.T496T Chr.1: 92247975-92247975 on Build GRCh38
4407967 c.1546T>A p.S516T Chr.1: 92247917-92247917 on Build GRCh38
5978043 c.1506G>C p.E502D Chr.1: 92247957-92247957 on Build GRCh38
3493355 c.1515C>T p.F505F Chr.1: 92247948-92247948 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs150536401 Chr.1: 92247981-92247981 on Build GRCh38
rs964432923 Chr.1: 92248253-92248253 on Build GRCh38
rs768238010 Chr.1: 92247870-92247870 on Build GRCh38
rs1030743292 Chr.1: 92248193-92248193 on Build GRCh38
rs34441399 Chr.1: 92247934-92247935 on Build GRCh38
rs776075245 Chr.1: 92247858-92247858 on Build GRCh38
rs558699815 Chr.1: 92248186-92248186 on Build GRCh38
rs1011481953 Chr.1: 92248188-92248188 on Build GRCh38
rs112414902 Chr.1: 92248070-92248070 on Build GRCh38
rs377485551 Chr.1: 92248015-92248015 on Build GRCh38
rs934189198 Chr.1: 92248022-92248022 on Build GRCh38
rs1031599290 Chr.1: 92247783-92247783 on Build GRCh38
rs188888558 Chr.1: 92247862-92247862 on Build GRCh38
rs780185005 Chr.1: 92248028-92248031 on Build GRCh38
rs762265911 Chr.1: 92247788-92247788 on Build GRCh38
rs1038626224 Chr.1: 92248052-92248052 on Build GRCh38
rs767224524 Chr.1: 92247888-92247888 on Build GRCh38
rs746943075 Chr.1: 92248041-92248041 on Build GRCh38
rs569694943 Chr.1: 92248009-92248009 on Build GRCh38
rs771514512 Chr.1: 92248035-92248035 on Build GRCh38
rs768862381 Chr.1: 92247834-92247834 on Build GRCh38
rs1052843632 Chr.1: 92248179-92248179 on Build GRCh38
rs964338208 Chr.1: 92247885-92247885 on Build GRCh38
rs907250727 Chr.1: 92248163-92248163 on Build GRCh38
rs747716033 Chr.1: 92248029-92248029 on Build GRCh38
rs989775612 Chr.1: 92247860-92247860 on Build GRCh38
rs578092105 Chr.1: 92248268-92248268 on Build GRCh38
rs757404436 Chr.1: 92247980-92247980 on Build GRCh38
rs891627016 Chr.1: 92248185-92248185 on Build GRCh38
rs147009011 Chr.1: 92247881-92247881 on Build GRCh38
rs532719020 Chr.1: 92248050-92248050 on Build GRCh38
rs758215984 Chr.1: 92248026-92248028 on Build GRCh38
rs940295619 Chr.1: 92248051-92248051 on Build GRCh38
rs766022940 Chr.1: 92247922-92247922 on Build GRCh38
rs780234890 Chr.1: 92248007-92248007 on Build GRCh38
rs552303288 Chr.1: 92248053-92248053 on Build GRCh38
rs1044501463 Chr.1: 92248127-92248127 on Build GRCh38
rs772838108 Chr.1: 92247900-92247900 on Build GRCh38
rs185682792 Chr.1: 92248120-92248120 on Build GRCh38
rs977868854 Chr.1: 92248189-92248190 on Build GRCh38
rs1021483928 Chr.1: 92248199-92248199 on Build GRCh38
rs1002715454 Chr.1: 92248178-92248178 on Build GRCh38
rs148335800 Chr.1: 92247962-92247962 on Build GRCh38
rs535813981 Chr.1: 92248001-92248004 on Build GRCh38
rs987341101 Chr.1: 92248027-92248027 on Build GRCh38
rs200719668 Chr.1: 92248008-92248008 on Build GRCh38
rs571387093 Chr.1: 92247818-92247818 on Build GRCh38
rs1022056124 Chr.1: 92247778-92247778 on Build GRCh38
rs960379559 Chr.1: 92248208-92248208 on Build GRCh38
rs753844057 Chr.1: 92247934-92247934 on Build GRCh38
rs547625852 Chr.1: 92247874-92247874 on Build GRCh38
rs555913359 Chr.1: 92248092-92248095 on Build GRCh38
rs762117271 Chr.1: 92247960-92247960 on Build GRCh38
rs760533628 Chr.1: 92247913-92247913 on Build GRCh38
rs1018545486 Chr.1: 92248070-92248070 on Build GRCh38
rs759342815 Chr.1: 92247941-92247941 on Build GRCh38
rs990698542 Chr.1: 92247776-92247776 on Build GRCh38
rs770038768 Chr.1: 92247836-92247836 on Build GRCh38
rs549886420 Chr.1: 92247792-92247792 on Build GRCh38
rs377426579 Chr.1: 92247994-92247994 on Build GRCh38
rs750270670 Chr.1: 92248012-92248014 on Build GRCh38
rs756870316 Chr.1: 92247843-92247843 on Build GRCh38

View Full Product Details