Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 92267830-92268341 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLMN
Entrez Gene ID
11146
Gene Name
glomulin, FKBP associated protein
Gene Aliases
FAP, FAP48, FAP68, FKBPAP, GLML, GVM, VMGLOM
UniGene
Hs.49105
Gene Chromosome Location
Chr.1: 92245786-92441936 on Build GRCh38
Transcripts
NM_001319683.1 NM_053274.2 NR_135089.1 XM_005270401.3 XM_006710309.2 XM_011540546.2 XM_017000137.1 XM_017000138.1 XM_017000139.1 XM_017000140.1 XM_017000141.1 XM_017000142.1 XM_017000143.1 XM_017000144.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7369560 c.1007T>C p.L336S Chr.1: 92268106-92268106 on Build GRCh38
6671663 c.1074C>A p.I358I Chr.1: 92267937-92267937 on Build GRCh38
6127855 c.1019A>T p.E340V Chr.1: 92267992-92267992 on Build GRCh38
2260024 c.989C>G p.S330C Chr.1: 92268124-92268124 on Build GRCh38
6308296 c.1085_1086insG p.T363fs*16 Chr.1: 92267925-92267926 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs546537881 Chr.1: 92268038-92268038 on Build GRCh38
rs1038486352 Chr.1: 92267931-92267931 on Build GRCh38
rs748340543 Chr.1: 92268029-92268029 on Build GRCh38
rs772484965 Chr.1: 92268050-92268052 on Build GRCh38
rs147889200 Chr.1: 92267989-92267989 on Build GRCh38
rs751761547 Chr.1: 92268169-92268169 on Build GRCh38
rs761602030 Chr.1: 92268122-92268122 on Build GRCh38
rs142899415 Chr.1: 92267936-92267936 on Build GRCh38
rs766312022 Chr.1: 92268151-92268151 on Build GRCh38
rs749507662 Chr.1: 92267888-92267889 on Build GRCh38
rs757034430 Chr.1: 92268012-92268012 on Build GRCh38
rs762542965 Chr.1: 92267991-92267991 on Build GRCh38
rs771340851 Chr.1: 92267901-92267905 on Build GRCh38
rs12097184 Chr.1: 92268313-92268313 on Build GRCh38
rs772591484 Chr.1: 92267945-92267945 on Build GRCh38
rs763713387 Chr.1: 92267994-92267994 on Build GRCh38
rs774710697 Chr.1: 92267938-92267938 on Build GRCh38
rs781104597 Chr.1: 92267912-92267912 on Build GRCh38
rs974264327 Chr.1: 92267983-92267983 on Build GRCh38
rs1003769382 Chr.1: 92267962-92267962 on Build GRCh38
rs547011435 Chr.1: 92268152-92268152 on Build GRCh38
rs149792649 Chr.1: 92267954-92267954 on Build GRCh38
rs898237460 Chr.1: 92267932-92267932 on Build GRCh38
rs758189308 Chr.1: 92267890-92267890 on Build GRCh38
rs778139262 Chr.1: 92268055-92268055 on Build GRCh38
rs757232570 Chr.1: 92267899-92267899 on Build GRCh38
rs764580289 Chr.1: 92267990-92267990 on Build GRCh38
rs771630349 Chr.1: 92267979-92267979 on Build GRCh38
rs751518586 Chr.1: 92267894-92267894 on Build GRCh38
rs748283193 Chr.1: 92267938-92267938 on Build GRCh38
rs956881810 Chr.1: 92268020-92268020 on Build GRCh38
rs767505575 Chr.1: 92268028-92268028 on Build GRCh38
rs529413997 Chr.1: 92268248-92268248 on Build GRCh38
rs534587626 Chr.1: 92268199-92268199 on Build GRCh38
rs896484299 Chr.1: 92268255-92268255 on Build GRCh38
rs768357425 Chr.1: 92268114-92268114 on Build GRCh38
rs1013634000 Chr.1: 92268301-92268301 on Build GRCh38
rs758535726 Chr.1: 92268157-92268157 on Build GRCh38
rs749444788 Chr.1: 92267915-92267915 on Build GRCh38
rs35258161 Chr.1: 92268106-92268106 on Build GRCh38
rs1013845641 Chr.1: 92267949-92267949 on Build GRCh38
rs750401776 Chr.1: 92268139-92268139 on Build GRCh38
rs975240301 Chr.1: 92268128-92268128 on Build GRCh38
rs761274225 Chr.1: 92268144-92268149 on Build GRCh38
rs374098674 Chr.1: 92267880-92267880 on Build GRCh38
rs920094946 Chr.1: 92268009-92268009 on Build GRCh38
rs528499611 Chr.1: 92268117-92268117 on Build GRCh38
rs776011972 Chr.1: 92268069-92268070 on Build GRCh38
rs372616217 Chr.1: 92268145-92268145 on Build GRCh38
rs778151590 Chr.1: 92268166-92268166 on Build GRCh38
rs747349623 Chr.1: 92267964-92267964 on Build GRCh38
rs776986944 Chr.1: 92267981-92267981 on Build GRCh38
rs771360797 Chr.1: 92268064-92268064 on Build GRCh38
rs933891160 Chr.1: 92267859-92267859 on Build GRCh38
rs370567051 Chr.1: 92268046-92268046 on Build GRCh38
rs769368413 Chr.1: 92268071-92268071 on Build GRCh38
rs1050744141 Chr.1: 92267881-92267881 on Build GRCh38
rs751393940 Chr.1: 92268002-92268002 on Build GRCh38
rs1023792600 Chr.1: 92267946-92267946 on Build GRCh38
rs907537324 Chr.1: 92268105-92268105 on Build GRCh38
rs568726964 Chr.1: 92268312-92268312 on Build GRCh38
rs374524087 Chr.1: 92268101-92268101 on Build GRCh38
rs1043538848 Chr.1: 92267877-92267877 on Build GRCh38
rs185115434 Chr.1: 92268212-92268212 on Build GRCh38
rs999602528 Chr.1: 92267893-92267893 on Build GRCh38
rs1000034267 Chr.1: 92268228-92268228 on Build GRCh38
rs747433665 Chr.1: 92268063-92268063 on Build GRCh38
rs1023625424 Chr.1: 92268305-92268305 on Build GRCh38
rs755117000 Chr.1: 92268030-92268030 on Build GRCh38
rs746161319 Chr.1: 92267952-92267956 on Build GRCh38
rs115185705 Chr.1: 92268250-92268250 on Build GRCh38
rs187528834 Chr.1: 92267891-92267891 on Build GRCh38
rs754956349 Chr.1: 92267922-92267922 on Build GRCh38

View Full Product Details