Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 92513631-92514062 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EVI5
Entrez Gene ID
7813
Gene Name
ecotropic viral integration site 5
Gene Aliases
EVI-5, NB4S
UniGene
Hs.594434
Gene Chromosome Location
Chr.1: 92508696-92792404 on Build GRCh38
Transcripts
NM_001308248.1 NM_005665.5 XM_017002269.1 XM_017002270.1 XM_017002271.1 XM_017002272.1 XM_017002273.1 XM_017002274.1 XM_017002275.1 XM_017002276.1 XM_017002277.1 XM_017002278.1 XM_017002279.1 XM_017002280.1 XM_017002281.1 XM_017002282.1 XM_017002283.1 XM_017002284.1 XM_017002285.1 XM_017002286.1 XM_017002287.1 XM_017002288.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5449240 c.2152-4A>G p.? Chr.1: 92513974-92513974 on Build GRCh38
4478524 c.2246C>T p.S749F Chr.1: 92513843-92513843 on Build GRCh38
4478525 c.2279C>T p.S760F Chr.1: 92513843-92513843 on Build GRCh38
6334812 c.2219C>A p.S740Y Chr.1: 92513870-92513870 on Build GRCh38
4866493 c.2386C>A p.L796M Chr.1: 92513736-92513736 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753139991 Chr.1: 92513776-92513776 on Build GRCh38
rs201135975 Chr.1: 92513728-92513728 on Build GRCh38
rs757836040 Chr.1: 92513782-92513782 on Build GRCh38
rs747494688 Chr.1: 92513690-92513690 on Build GRCh38
rs762422659 Chr.1: 92513692-92513692 on Build GRCh38
rs202224223 Chr.1: 92513821-92513821 on Build GRCh38
rs529586905 Chr.1: 92513724-92513724 on Build GRCh38
rs201443870 Chr.1: 92513679-92513679 on Build GRCh38
rs768609602 Chr.1: 92513726-92513726 on Build GRCh38
rs748146159 Chr.1: 92513822-92513822 on Build GRCh38
rs140423717 Chr.1: 92513962-92513962 on Build GRCh38
rs745602454 Chr.1: 92513704-92513704 on Build GRCh38
rs769825409 Chr.1: 92513932-92513932 on Build GRCh38
rs969429528 Chr.1: 92513967-92513967 on Build GRCh38
rs144597007 Chr.1: 92513722-92513722 on Build GRCh38
rs533408017 Chr.1: 92513857-92513857 on Build GRCh38
rs779591884 Chr.1: 92513901-92513901 on Build GRCh38
rs373069997 Chr.1: 92513781-92513781 on Build GRCh38
rs199716793 Chr.1: 92513914-92513914 on Build GRCh38
rs757852548 Chr.1: 92513874-92513874 on Build GRCh38
rs368759252 Chr.1: 92513757-92513757 on Build GRCh38
rs755618652 Chr.1: 92513899-92513899 on Build GRCh38
rs199630852 Chr.1: 92513872-92513872 on Build GRCh38
rs201155262 Chr.1: 92513866-92513866 on Build GRCh38
rs6603979 Chr.1: 92513797-92513797 on Build GRCh38
rs969483216 Chr.1: 92513693-92513693 on Build GRCh38
rs766500924 Chr.1: 92513754-92513754 on Build GRCh38
rs755476736 Chr.1: 92513696-92513696 on Build GRCh38
rs200374814 Chr.1: 92513839-92513839 on Build GRCh38
rs199913873 Chr.1: 92513684-92513684 on Build GRCh38
rs201511730 Chr.1: 92513697-92513697 on Build GRCh38
rs769264438 Chr.1: 92513714-92513714 on Build GRCh38
rs74101172 Chr.1: 92514022-92514022 on Build GRCh38
rs201939388 Chr.1: 92514006-92514006 on Build GRCh38
rs748996054 Chr.1: 92513902-92513902 on Build GRCh38
rs41286809 Chr.1: 92513875-92513875 on Build GRCh38
rs139452163 Chr.1: 92513791-92513791 on Build GRCh38
rs200686753 Chr.1: 92513674-92513674 on Build GRCh38
rs138793808 Chr.1: 92513997-92513997 on Build GRCh38
rs61735641 Chr.1: 92513794-92513794 on Build GRCh38
rs772081750 Chr.1: 92513916-92513916 on Build GRCh38
rs199521058 Chr.1: 92513849-92513849 on Build GRCh38
rs373938333 Chr.1: 92513939-92513939 on Build GRCh38
rs200683132 Chr.1: 92513853-92513853 on Build GRCh38
rs773066792 Chr.1: 92513922-92513922 on Build GRCh38
rs752171993 Chr.1: 92513779-92513779 on Build GRCh38
rs142905346 Chr.1: 92513808-92513808 on Build GRCh38
rs138799860 Chr.1: 92513689-92513689 on Build GRCh38
rs567138887 Chr.1: 92514010-92514010 on Build GRCh38
rs200102416 Chr.1: 92513847-92513847 on Build GRCh38
rs778679412 Chr.1: 92513905-92513905 on Build GRCh38
rs915141295 Chr.1: 92514036-92514036 on Build GRCh38
rs755812556 Chr.1: 92513795-92513795 on Build GRCh38
rs746945638 Chr.1: 92513928-92513928 on Build GRCh38
rs202155652 Chr.1: 92513755-92513755 on Build GRCh38
rs774116515 Chr.1: 92513811-92513811 on Build GRCh38
rs762042622 Chr.1: 92513965-92513965 on Build GRCh38
rs146602523 Chr.1: 92513828-92513828 on Build GRCh38
rs988195047 Chr.1: 92514023-92514023 on Build GRCh38
rs893186313 Chr.1: 92513710-92513710 on Build GRCh38
rs147173044 Chr.1: 92513774-92513774 on Build GRCh38
rs183854416 Chr.1: 92513903-92513903 on Build GRCh38
rs761714329 Chr.1: 92513739-92513739 on Build GRCh38
rs566986526 Chr.1: 92513883-92513883 on Build GRCh38
rs144291497 Chr.1: 92513778-92513778 on Build GRCh38
rs972882832 Chr.1: 92513864-92513864 on Build GRCh38
rs201450382 Chr.1: 92513846-92513846 on Build GRCh38
rs371768613 Chr.1: 92513968-92513968 on Build GRCh38
rs768486325 Chr.1: 92513809-92513809 on Build GRCh38
rs747893888 Chr.1: 92513910-92513910 on Build GRCh38

View Full Product Details