Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 28786007-28786507 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC117
Entrez Gene ID
150275
Gene Name
coiled-coil domain containing 117
Gene Aliases
dJ366L4.1
UniGene
Hs.406460
Gene Chromosome Location
Chr.22: 28772674-28789301 on Build GRCh38
Transcripts
NM_001284263.1 NM_001284264.1 NM_001284265.1 NM_173510.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3740385 c.728G>T p.G243V Chr.22: 28786214-28786214 on Build GRCh38
127552 c.630C>A p.L210L Chr.22: 28786116-28786116 on Build GRCh38
3701727 c.603-6T>C p.? Chr.22: 28786083-28786083 on Build GRCh38
3553062 c.652C>T p.L218F Chr.22: 28786138-28786138 on Build GRCh38
1199804 c.831G>A p.M277I Chr.22: 28786317-28786317 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs575148934 Chr.22: 28786401-28786401 on Build GRCh38
rs377732219 Chr.22: 28786179-28786179 on Build GRCh38
rs769663128 Chr.22: 28786471-28786471 on Build GRCh38
rs767124392 Chr.22: 28786045-28786045 on Build GRCh38
rs754989005 Chr.22: 28786286-28786286 on Build GRCh38
rs149069980 Chr.22: 28786255-28786255 on Build GRCh38
rs758891981 Chr.22: 28786297-28786297 on Build GRCh38
rs1019412606 Chr.22: 28786355-28786355 on Build GRCh38
rs764051659 Chr.22: 28786253-28786253 on Build GRCh38
rs754335519 Chr.22: 28786094-28786094 on Build GRCh38
rs780433074 Chr.22: 28786099-28786099 on Build GRCh38
rs767869628 Chr.22: 28786178-28786178 on Build GRCh38
rs745396819 Chr.22: 28786335-28786335 on Build GRCh38
rs776246858 Chr.22: 28786229-28786229 on Build GRCh38
rs749876500 Chr.22: 28786100-28786100 on Build GRCh38
rs754865644 Chr.22: 28786066-28786066 on Build GRCh38
rs753971274 Chr.22: 28786285-28786285 on Build GRCh38
rs148196050 Chr.22: 28786290-28786290 on Build GRCh38
rs932034692 Chr.22: 28786456-28786456 on Build GRCh38
rs760107421 Chr.22: 28786137-28786137 on Build GRCh38
rs139415822 Chr.22: 28786159-28786159 on Build GRCh38
rs763044376 Chr.22: 28786343-28786343 on Build GRCh38
rs748468959 Chr.22: 28786116-28786116 on Build GRCh38
rs563797090 Chr.22: 28786475-28786475 on Build GRCh38
rs771731132 Chr.22: 28786121-28786121 on Build GRCh38
rs377126733 Chr.22: 28786333-28786333 on Build GRCh38
rs749963124 Chr.22: 28786046-28786046 on Build GRCh38
rs17514713 Chr.22: 28786082-28786082 on Build GRCh38
rs745967615 Chr.22: 28786093-28786093 on Build GRCh38
rs756290019 Chr.22: 28786180-28786180 on Build GRCh38
rs189905230 Chr.22: 28786420-28786420 on Build GRCh38
rs373921809 Chr.22: 28786053-28786053 on Build GRCh38
rs774735302 Chr.22: 28786111-28786111 on Build GRCh38
rs758218442 Chr.22: 28786172-28786172 on Build GRCh38
rs1050462867 Chr.22: 28786466-28786468 on Build GRCh38
rs779340051 Chr.22: 28786206-28786206 on Build GRCh38
rs753741058 Chr.22: 28786062-28786062 on Build GRCh38
rs773561052 Chr.22: 28786221-28786221 on Build GRCh38
rs975072274 Chr.22: 28786085-28786085 on Build GRCh38
rs145482585 Chr.22: 28786200-28786200 on Build GRCh38
rs775139472 Chr.22: 28786341-28786341 on Build GRCh38
rs563450116 Chr.22: 28786387-28786387 on Build GRCh38
rs370272489 Chr.22: 28786216-28786216 on Build GRCh38
rs748521988 Chr.22: 28786208-28786208 on Build GRCh38
rs753936751 Chr.22: 28786368-28786368 on Build GRCh38
rs938069705 Chr.22: 28786480-28786480 on Build GRCh38
rs749295014 Chr.22: 28786199-28786199 on Build GRCh38
rs994208940 Chr.22: 28786282-28786282 on Build GRCh38
rs759917186 Chr.22: 28786173-28786173 on Build GRCh38
rs766241307 Chr.22: 28786056-28786056 on Build GRCh38
rs770648979 Chr.22: 28786325-28786325 on Build GRCh38
rs753269320 Chr.22: 28786296-28786296 on Build GRCh38
rs774278635 Chr.22: 28786446-28786446 on Build GRCh38
rs913274011 Chr.22: 28786165-28786165 on Build GRCh38
rs766633062 Chr.22: 28786357-28786357 on Build GRCh38
rs752519765 Chr.22: 28786078-28786078 on Build GRCh38
rs775510141 Chr.22: 28786247-28786247 on Build GRCh38
rs765180063 Chr.22: 28786068-28786068 on Build GRCh38
rs779973566 Chr.22: 28786482-28786482 on Build GRCh38
rs769031781 Chr.22: 28786102-28786102 on Build GRCh38
rs938878113 Chr.22: 28786115-28786115 on Build GRCh38
rs745406977 Chr.22: 28786426-28786426 on Build GRCh38
rs768694822 Chr.22: 28786344-28786344 on Build GRCh38
rs761463911 Chr.22: 28786356-28786356 on Build GRCh38
rs373725869 Chr.22: 28786225-28786225 on Build GRCh38
rs761009679 Chr.22: 28786256-28786256 on Build GRCh38
rs144460803 Chr.22: 28786303-28786303 on Build GRCh38
rs372804266 Chr.22: 28786373-28786373 on Build GRCh38
rs781525792 Chr.22: 28786088-28786088 on Build GRCh38
rs758902783 Chr.22: 28786273-28786273 on Build GRCh38
rs369892620 Chr.22: 28786169-28786169 on Build GRCh38
rs77099742 Chr.22: 28786181-28786181 on Build GRCh38
rs769875611 Chr.22: 28786246-28786246 on Build GRCh38
rs370581950 Chr.22: 28786348-28786348 on Build GRCh38
rs765237278 Chr.22: 28786161-28786161 on Build GRCh38
rs751223225 Chr.22: 28786339-28786339 on Build GRCh38
rs554533204 Chr.22: 28786248-28786248 on Build GRCh38
rs772789093 Chr.22: 28786131-28786131 on Build GRCh38
rs774229679 Chr.22: 28786354-28786354 on Build GRCh38
rs9613680 Chr.22: 28786301-28786301 on Build GRCh38
rs1046533517 Chr.22: 28786454-28786454 on Build GRCh38
rs908788515 Chr.22: 28786425-28786425 on Build GRCh38
rs758929726 Chr.22: 28786236-28786236 on Build GRCh38
rs908666795 Chr.22: 28786150-28786150 on Build GRCh38
rs765979784 Chr.22: 28786156-28786156 on Build GRCh38
rs759569360 Chr.22: 28786160-28786160 on Build GRCh38
rs138007836 Chr.22: 28786224-28786224 on Build GRCh38

View Full Product Details