Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 32149411-32149948 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C22orf42
Entrez Gene ID
150297
Gene Name
chromosome 22 open reading frame 42
Gene Aliases
dJ90G24.6
UniGene
Hs.426551
Gene Chromosome Location
Chr.22: 32149006-32160448 on Build GRCh38
Transcripts
NM_001010859.1 XM_011529922.2 XM_017028629.1 XM_017028630.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5793645 c.655-10C>T p.? Chr.22: 32149790-32149790 on Build GRCh38
726555 c.679C>A p.L227I Chr.22: 32149756-32149756 on Build GRCh38
4555603 c.661G>A p.E221K Chr.22: 32149774-32149774 on Build GRCh38
3726175 c.678C>A p.Y226* Chr.22: 32149757-32149757 on Build GRCh38
6615683 c.677A>G p.Y226C Chr.22: 32149758-32149758 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs945860542 Chr.22: 32149708-32149708 on Build GRCh38
rs748086050 Chr.22: 32149652-32149654 on Build GRCh38
rs376522438 Chr.22: 32149727-32149727 on Build GRCh38
rs374254447 Chr.22: 32149689-32149694 on Build GRCh38
rs67058393 Chr.22: 32149652-32149652 on Build GRCh38
rs45462396 Chr.22: 32149802-32149802 on Build GRCh38
rs373116952 Chr.22: 32149548-32149548 on Build GRCh38
rs773873387 Chr.22: 32149654-32149656 on Build GRCh38
rs759364791 Chr.22: 32149829-32149829 on Build GRCh38
rs944243377 Chr.22: 32149891-32149891 on Build GRCh38
rs557911499 Chr.22: 32149736-32149737 on Build GRCh38
rs749521486 Chr.22: 32149642-32149642 on Build GRCh38
rs768013286 Chr.22: 32149664-32149679 on Build GRCh38
rs770859580 Chr.22: 32149651-32149652 on Build GRCh38
rs370278397 Chr.22: 32149482-32149482 on Build GRCh38
rs994749616 Chr.22: 32149589-32149589 on Build GRCh38
rs916322271 Chr.22: 32149830-32149830 on Build GRCh38
rs1019972639 Chr.22: 32149590-32149590 on Build GRCh38
rs9606913 Chr.22: 32149650-32149650 on Build GRCh38
rs376903043 Chr.22: 32149917-32149917 on Build GRCh38
rs774416885 Chr.22: 32149747-32149747 on Build GRCh38
rs898743555 Chr.22: 32149730-32149730 on Build GRCh38
rs756883426 Chr.22: 32149781-32149781 on Build GRCh38
rs771794018 Chr.22: 32149562-32149562 on Build GRCh38
rs746957023 Chr.22: 32149649-32149650 on Build GRCh38
rs45494991 Chr.22: 32149775-32149775 on Build GRCh38
rs753754217 Chr.22: 32149713-32149713 on Build GRCh38
rs964414656 Chr.22: 32149648-32149649 on Build GRCh38
rs761873724 Chr.22: 32149752-32149752 on Build GRCh38
rs771918850 Chr.22: 32149725-32149725 on Build GRCh38
rs367944982 Chr.22: 32149841-32149841 on Build GRCh38
rs375489236 Chr.22: 32149769-32149769 on Build GRCh38
rs143839138 Chr.22: 32149514-32149514 on Build GRCh38
rs992723398 Chr.22: 32149906-32149906 on Build GRCh38
rs776311321 Chr.22: 32149650-32149651 on Build GRCh38
rs748881470 Chr.22: 32149513-32149513 on Build GRCh38
rs773395563 Chr.22: 32149765-32149765 on Build GRCh38
rs66503637 Chr.22: 32149654-32149654 on Build GRCh38
rs374195653 Chr.22: 32149638-32149638 on Build GRCh38
rs756421877 Chr.22: 32149642-32149643 on Build GRCh38
rs553928285 Chr.22: 32149854-32149854 on Build GRCh38
rs1042105400 Chr.22: 32149484-32149484 on Build GRCh38
rs756045642 Chr.22: 32149502-32149502 on Build GRCh38
rs755886127 Chr.22: 32149703-32149703 on Build GRCh38
rs200515923 Chr.22: 32149654-32149654 on Build GRCh38
rs541455767 Chr.22: 32149648-32149648 on Build GRCh38
rs535937358 Chr.22: 32149766-32149766 on Build GRCh38
rs776357479 Chr.22: 32149823-32149823 on Build GRCh38
rs746689734 Chr.22: 32149779-32149779 on Build GRCh38
rs572578311 Chr.22: 32149641-32149641 on Build GRCh38
rs193128843 Chr.22: 32149472-32149472 on Build GRCh38
rs991902730 Chr.22: 32149825-32149825 on Build GRCh38
rs766559340 Chr.22: 32149815-32149815 on Build GRCh38
rs926648280 Chr.22: 32149872-32149872 on Build GRCh38
rs1037639147 Chr.22: 32149534-32149534 on Build GRCh38
rs911784566 Chr.22: 32149462-32149462 on Build GRCh38
rs58589689 Chr.22: 32149652-32149652 on Build GRCh38
rs777142463 Chr.22: 32149573-32149573 on Build GRCh38
rs975092976 Chr.22: 32149474-32149474 on Build GRCh38
rs142171049 Chr.22: 32149641-32149642 on Build GRCh38
rs772174648 Chr.22: 32149808-32149808 on Build GRCh38
rs879142967 Chr.22: 32149448-32149449 on Build GRCh38
rs566985292 Chr.22: 32149736-32149736 on Build GRCh38
rs765218442 Chr.22: 32149770-32149770 on Build GRCh38
rs748272719 Chr.22: 32149803-32149803 on Build GRCh38
rs72105282 Chr.22: 32149863-32149864 on Build GRCh38
rs915060743 Chr.22: 32149477-32149477 on Build GRCh38
rs544032029 Chr.22: 32149656-32149656 on Build GRCh38
rs759975893 Chr.22: 32149578-32149578 on Build GRCh38
rs955472716 Chr.22: 32149595-32149595 on Build GRCh38
rs559028285 Chr.22: 32149627-32149627 on Build GRCh38
rs895677880 Chr.22: 32149707-32149707 on Build GRCh38
rs775838348 Chr.22: 32149599-32149599 on Build GRCh38
rs28413788 Chr.22: 32149733-32149733 on Build GRCh38
rs923689098 Chr.22: 32149486-32149486 on Build GRCh38
rs746284860 Chr.22: 32149729-32149729 on Build GRCh38
rs750826482 Chr.22: 32149703-32149714 on Build GRCh38
rs1001381628 Chr.22: 32149540-32149540 on Build GRCh38
rs777571331 Chr.22: 32149596-32149596 on Build GRCh38
rs58570639 Chr.22: 32149862-32149863 on Build GRCh38
rs572629908 Chr.22: 32149875-32149875 on Build GRCh38
rs937295808 Chr.22: 32149699-32149699 on Build GRCh38
rs563830942 Chr.22: 32149774-32149774 on Build GRCh38
rs754897457 Chr.22: 32149714-32149714 on Build GRCh38
rs745603810 Chr.22: 32149651-32149654 on Build GRCh38
rs1009968687 Chr.22: 32149582-32149582 on Build GRCh38
rs371767433 Chr.22: 32149862-32149862 on Build GRCh38
rs1008152885 Chr.22: 32149634-32149638 on Build GRCh38
rs753614998 Chr.22: 32149768-32149768 on Build GRCh38
rs941376271 Chr.22: 32149661-32149661 on Build GRCh38
rs1000962915 Chr.22: 32149921-32149921 on Build GRCh38
rs779025446 Chr.22: 32149645-32149646 on Build GRCh38
rs1056931980 Chr.22: 32149547-32149547 on Build GRCh38
rs374984387 Chr.22: 32149814-32149814 on Build GRCh38
rs540390509 Chr.22: 32149592-32149592 on Build GRCh38
rs974163502 Chr.22: 32149852-32149852 on Build GRCh38
rs747596936 Chr.22: 32149723-32149723 on Build GRCh38
rs369086960 Chr.22: 32149861-32149861 on Build GRCh38
rs115275535 Chr.22: 32149539-32149539 on Build GRCh38
rs778303505 Chr.22: 32149719-32149719 on Build GRCh38
rs916249311 Chr.22: 32149726-32149726 on Build GRCh38
rs920160300 Chr.22: 32149853-32149853 on Build GRCh38
rs56938357 Chr.22: 32149654-32149654 on Build GRCh38
rs986765107 Chr.22: 32149449-32149449 on Build GRCh38
rs184880738 Chr.22: 32149594-32149594 on Build GRCh38
rs749597989 Chr.22: 32149786-32149786 on Build GRCh38
rs773084973 Chr.22: 32149708-32149711 on Build GRCh38
rs192373601 Chr.22: 32149667-32149667 on Build GRCh38
rs1039724634 Chr.22: 32149900-32149900 on Build GRCh38
rs138566276 Chr.22: 32149704-32149704 on Build GRCh38
rs775181116 Chr.22: 32149651-32149656 on Build GRCh38
rs745367107 Chr.22: 32149861-32149862 on Build GRCh38
rs769107364 Chr.22: 32149787-32149787 on Build GRCh38
rs183683173 Chr.22: 32149793-32149793 on Build GRCh38
rs200254624 Chr.22: 32149811-32149811 on Build GRCh38
rs767782407 Chr.22: 32149759-32149759 on Build GRCh38
rs926644595 Chr.22: 32149669-32149669 on Build GRCh38
rs747270720 Chr.22: 32149603-32149603 on Build GRCh38
rs780294111 Chr.22: 32149785-32149785 on Build GRCh38

View Full Product Details