Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 32151803-32152299 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C22orf42
Entrez Gene ID
150297
Gene Name
chromosome 22 open reading frame 42
Gene Aliases
dJ90G24.6
UniGene
Hs.426551
Gene Chromosome Location
Chr.22: 32149006-32160448 on Build GRCh38
Transcripts
NM_001010859.1 XM_011529922.2 XM_017028629.1 XM_017028630.1 XM_017028631.1 XM_017028632.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5177497 c.373-9T>G p.? Chr.22: 32152103-32152103 on Build GRCh38
4546189 c.394G>A p.D132N Chr.22: 32152073-32152073 on Build GRCh38
3694032 c.379C>T p.P127S Chr.22: 32152088-32152088 on Build GRCh38
2938930 c.400+1G>A p.? Chr.22: 32152066-32152066 on Build GRCh38
3553701 c.397C>T p.H133Y Chr.22: 32152070-32152070 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs557065491 Chr.22: 32152174-32152174 on Build GRCh38
rs200984618 Chr.22: 32152219-32152220 on Build GRCh38
rs200925446 Chr.22: 32152059-32152060 on Build GRCh38
rs748830010 Chr.22: 32152067-32152067 on Build GRCh38
rs1029558409 Chr.22: 32151973-32151973 on Build GRCh38
rs549475827 Chr.22: 32151856-32151856 on Build GRCh38
rs543816958 Chr.22: 32152141-32152141 on Build GRCh38
rs67223520 Chr.22: 32151997-32151997 on Build GRCh38
rs139618788 Chr.22: 32152066-32152066 on Build GRCh38
rs757197338 Chr.22: 32152043-32152043 on Build GRCh38
rs569470102 Chr.22: 32152050-32152050 on Build GRCh38
rs558377831 Chr.22: 32151983-32151983 on Build GRCh38
rs748912944 Chr.22: 32152023-32152023 on Build GRCh38
rs752765853 Chr.22: 32152059-32152059 on Build GRCh38
rs11272724 Chr.22: 32151875-32151891 on Build GRCh38
rs376249545 Chr.22: 32152138-32152138 on Build GRCh38
rs957152734 Chr.22: 32152272-32152272 on Build GRCh38
rs766886097 Chr.22: 32152047-32152047 on Build GRCh38
rs372759024 Chr.22: 32152074-32152074 on Build GRCh38
rs762544887 Chr.22: 32151996-32151997 on Build GRCh38
rs546519491 Chr.22: 32152239-32152239 on Build GRCh38
rs151297448 Chr.22: 32152073-32152073 on Build GRCh38
rs777006249 Chr.22: 32152082-32152082 on Build GRCh38
rs534470724 Chr.22: 32152063-32152063 on Build GRCh38
rs529712859 Chr.22: 32151850-32151850 on Build GRCh38
rs763006218 Chr.22: 32152100-32152100 on Build GRCh38
rs777250842 Chr.22: 32152049-32152049 on Build GRCh38
rs750171281 Chr.22: 32152111-32152111 on Build GRCh38
rs775497326 Chr.22: 32152088-32152088 on Build GRCh38
rs771454596 Chr.22: 32152078-32152078 on Build GRCh38
rs878982422 Chr.22: 32152253-32152253 on Build GRCh38
rs774524901 Chr.22: 32152103-32152103 on Build GRCh38
rs997499330 Chr.22: 32152162-32152162 on Build GRCh38
rs760049026 Chr.22: 32152050-32152050 on Build GRCh38
rs577642158 Chr.22: 32152187-32152187 on Build GRCh38
rs765440722 Chr.22: 32152060-32152060 on Build GRCh38
rs763091367 Chr.22: 32152062-32152062 on Build GRCh38
rs752722659 Chr.22: 32152107-32152112 on Build GRCh38
rs766330142 Chr.22: 32152124-32152124 on Build GRCh38
rs1029395575 Chr.22: 32152186-32152186 on Build GRCh38
rs901818955 Chr.22: 32151930-32151930 on Build GRCh38
rs781415286 Chr.22: 32152144-32152144 on Build GRCh38
rs376888120 Chr.22: 32152030-32152030 on Build GRCh38
rs201291462 Chr.22: 32152139-32152139 on Build GRCh38
rs985423419 Chr.22: 32151998-32151998 on Build GRCh38
rs756007676 Chr.22: 32152121-32152121 on Build GRCh38
rs772882950 Chr.22: 32151874-32151875 on Build GRCh38
rs771618458 Chr.22: 32152040-32152040 on Build GRCh38
rs1004922848 Chr.22: 32151885-32151887 on Build GRCh38
rs781147335 Chr.22: 32152064-32152064 on Build GRCh38
rs767318752 Chr.22: 32152107-32152107 on Build GRCh38
rs773782369 Chr.22: 32152033-32152033 on Build GRCh38
rs764347862 Chr.22: 32152102-32152102 on Build GRCh38
rs761980011 Chr.22: 32152105-32152105 on Build GRCh38
rs759698424 Chr.22: 32152049-32152050 on Build GRCh38
rs78374783 Chr.22: 32152083-32152083 on Build GRCh38
rs747423244 Chr.22: 32152077-32152077 on Build GRCh38
rs150457508 Chr.22: 32152179-32152179 on Build GRCh38
rs566782039 Chr.22: 32152050-32152051 on Build GRCh38
rs753341562 Chr.22: 32152137-32152137 on Build GRCh38
rs887971934 Chr.22: 32151861-32151861 on Build GRCh38
rs144766464 Chr.22: 32151946-32151946 on Build GRCh38
rs569648517 Chr.22: 32151936-32151936 on Build GRCh38
rs1014997903 Chr.22: 32151916-32151916 on Build GRCh38
rs878910639 Chr.22: 32152263-32152263 on Build GRCh38
rs1007212329 Chr.22: 32152196-32152196 on Build GRCh38
rs13056640 Chr.22: 32152122-32152122 on Build GRCh38
rs1036366898 Chr.22: 32152136-32152136 on Build GRCh38
rs11273161 Chr.22: 32151877-32151893 on Build GRCh38
rs73881618 Chr.22: 32152034-32152034 on Build GRCh38
rs750355295 Chr.22: 32152065-32152065 on Build GRCh38
rs770270912 Chr.22: 32152087-32152087 on Build GRCh38

View Full Product Details