Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 32158898-32159399 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C22orf42
Entrez Gene ID
150297
Gene Name
chromosome 22 open reading frame 42
Gene Aliases
dJ90G24.6
UniGene
Hs.426551
Gene Chromosome Location
Chr.22: 32149006-32160448 on Build GRCh38
Transcripts
NM_001010859.1 XM_011529922.2 XM_017028629.1 XM_017028630.1 XM_017028631.1 XM_017028632.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
213275 c.205A>T p.T69S Chr.22: 32159011-32159011 on Build GRCh38
4463387 c.128C>T p.S43F Chr.22: 32159088-32159088 on Build GRCh38
4667549 c.12A>G p.K4K Chr.22: 32159204-32159204 on Build GRCh38
478902 c.97C>T p.P33S Chr.22: 32159119-32159119 on Build GRCh38
478903 c.82C>T p.H28Y Chr.22: 32159134-32159134 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774132233 Chr.22: 32158976-32158976 on Build GRCh38
rs749031578 Chr.22: 32158962-32158962 on Build GRCh38
rs186813867 Chr.22: 32159300-32159300 on Build GRCh38
rs921977793 Chr.22: 32159016-32159016 on Build GRCh38
rs747747191 Chr.22: 32158998-32158999 on Build GRCh38
rs750456865 Chr.22: 32159050-32159050 on Build GRCh38
rs375236573 Chr.22: 32159269-32159269 on Build GRCh38
rs78036098 Chr.22: 32159010-32159010 on Build GRCh38
rs1009954327 Chr.22: 32159363-32159363 on Build GRCh38
rs79988797 Chr.22: 32159007-32159007 on Build GRCh38
rs940783554 Chr.22: 32159278-32159278 on Build GRCh38
rs762410800 Chr.22: 32159146-32159146 on Build GRCh38
rs765515191 Chr.22: 32159252-32159252 on Build GRCh38
rs758392170 Chr.22: 32159205-32159205 on Build GRCh38
rs537399401 Chr.22: 32159063-32159063 on Build GRCh38
rs777335751 Chr.22: 32159113-32159113 on Build GRCh38
rs370420192 Chr.22: 32159268-32159268 on Build GRCh38
rs780232860 Chr.22: 32159288-32159288 on Build GRCh38
rs748080257 Chr.22: 32158979-32158979 on Build GRCh38
rs972594327 Chr.22: 32159305-32159305 on Build GRCh38
rs5998267 Chr.22: 32158998-32158998 on Build GRCh38
rs756471138 Chr.22: 32159062-32159062 on Build GRCh38
rs754609078 Chr.22: 32159175-32159175 on Build GRCh38
rs769417520 Chr.22: 32159223-32159223 on Build GRCh38
rs890454698 Chr.22: 32159102-32159102 on Build GRCh38
rs373616961 Chr.22: 32159241-32159241 on Build GRCh38
rs780701425 Chr.22: 32159214-32159214 on Build GRCh38
rs770621495 Chr.22: 32159119-32159119 on Build GRCh38
rs752035598 Chr.22: 32159015-32159015 on Build GRCh38
rs749026989 Chr.22: 32158964-32158964 on Build GRCh38
rs755172070 Chr.22: 32159186-32159186 on Build GRCh38
rs924162377 Chr.22: 32159284-32159284 on Build GRCh38
rs772814338 Chr.22: 32159114-32159115 on Build GRCh38
rs376397903 Chr.22: 32158943-32158943 on Build GRCh38
rs759877360 Chr.22: 32159239-32159239 on Build GRCh38
rs372472168 Chr.22: 32159194-32159194 on Build GRCh38
rs771499241 Chr.22: 32159180-32159180 on Build GRCh38
rs370318154 Chr.22: 32159211-32159211 on Build GRCh38
rs933311608 Chr.22: 32159318-32159318 on Build GRCh38
rs147242510 Chr.22: 32159212-32159212 on Build GRCh38
rs557701595 Chr.22: 32159299-32159299 on Build GRCh38
rs768201524 Chr.22: 32158991-32158999 on Build GRCh38
rs755676084 Chr.22: 32158971-32158971 on Build GRCh38
rs889586409 Chr.22: 32158994-32158994 on Build GRCh38
rs143881857 Chr.22: 32159112-32159112 on Build GRCh38
rs748092756 Chr.22: 32159110-32159110 on Build GRCh38
rs970346131 Chr.22: 32159216-32159216 on Build GRCh38
rs931974468 Chr.22: 32159018-32159018 on Build GRCh38
rs1050981147 Chr.22: 32159324-32159324 on Build GRCh38
rs754696040 Chr.22: 32159090-32159090 on Build GRCh38
rs199682684 Chr.22: 32159036-32159036 on Build GRCh38
rs781361748 Chr.22: 32159238-32159238 on Build GRCh38
rs758805085 Chr.22: 32159210-32159210 on Build GRCh38
rs771656766 Chr.22: 32159038-32159040 on Build GRCh38
rs373240877 Chr.22: 32158932-32158932 on Build GRCh38
rs754031579 Chr.22: 32159185-32159185 on Build GRCh38
rs369125574 Chr.22: 32158951-32158951 on Build GRCh38
rs762255537 Chr.22: 32159257-32159257 on Build GRCh38
rs183438705 Chr.22: 32158969-32158969 on Build GRCh38
rs753475927 Chr.22: 32159031-32159031 on Build GRCh38
rs746112432 Chr.22: 32158947-32158947 on Build GRCh38
rs778696817 Chr.22: 32158996-32158997 on Build GRCh38
rs1026736565 Chr.22: 32159151-32159151 on Build GRCh38
rs751711962 Chr.22: 32158938-32158938 on Build GRCh38
rs923235810 Chr.22: 32159310-32159310 on Build GRCh38
rs546445829 Chr.22: 32159346-32159346 on Build GRCh38
rs780000444 Chr.22: 32158960-32158960 on Build GRCh38
rs368043755 Chr.22: 32159266-32159266 on Build GRCh38
rs761292824 Chr.22: 32159173-32159183 on Build GRCh38
rs201710177 Chr.22: 32159215-32159215 on Build GRCh38
rs750587543 Chr.22: 32159164-32159164 on Build GRCh38
rs892993537 Chr.22: 32159357-32159357 on Build GRCh38
rs12160688 Chr.22: 32159009-32159009 on Build GRCh38
rs961490207 Chr.22: 32158958-32158958 on Build GRCh38
rs768921295 Chr.22: 32159137-32159137 on Build GRCh38
rs777146793 Chr.22: 32159233-32159233 on Build GRCh38
rs778577579 Chr.22: 32159189-32159189 on Build GRCh38
rs369034462 Chr.22: 32159129-32159129 on Build GRCh38
rs995713764 Chr.22: 32159116-32159116 on Build GRCh38
rs138905164 Chr.22: 32159006-32159006 on Build GRCh38
rs774715589 Chr.22: 32159143-32159143 on Build GRCh38
rs762357889 Chr.22: 32159026-32159026 on Build GRCh38
rs771594282 Chr.22: 32158997-32158997 on Build GRCh38
rs768133299 Chr.22: 32159160-32159160 on Build GRCh38
rs778723006 Chr.22: 32159103-32159103 on Build GRCh38
rs1054290876 Chr.22: 32159350-32159350 on Build GRCh38
rs757379498 Chr.22: 32158939-32158939 on Build GRCh38
rs889692764 Chr.22: 32159344-32159344 on Build GRCh38

View Full Product Details