Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 37057060-37057555 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCTD17
Entrez Gene ID
79734
Gene Name
potassium channel tetramerization domain containing 17
Gene Aliases
-
UniGene
Hs.517597
Gene Chromosome Location
Chr.22: 37051725-37063390 on Build GRCh38
Transcripts
NM_001282684.1 NM_001282685.1 NM_001282686.1 NM_024681.3 XM_005261741.3 XM_005261742.2 XM_005261743.2 XM_005261744.2 XM_011530374.2 XM_011530377.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4794270 c.447C>T p.C149C Chr.22: 37057433-37057433 on Build GRCh38
7258617 c.391-7C>T p.? Chr.22: 37057391-37057391 on Build GRCh38
4794269 c.426C>T p.C142C Chr.22: 37057433-37057433 on Build GRCh38
5927913 c.433C>T p.R145C Chr.22: 37057419-37057419 on Build GRCh38
4694799 c.497G>A p.R166H Chr.22: 37057483-37057483 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs915858625 Chr.22: 37057094-37057094 on Build GRCh38
rs760448451 Chr.22: 37057388-37057388 on Build GRCh38
rs374876906 Chr.22: 37057404-37057404 on Build GRCh38
rs710185 Chr.22: 37057412-37057412 on Build GRCh38
rs908498437 Chr.22: 37057110-37057110 on Build GRCh38
rs1015582813 Chr.22: 37057267-37057267 on Build GRCh38
rs922676692 Chr.22: 37057161-37057161 on Build GRCh38
rs143231972 Chr.22: 37057338-37057338 on Build GRCh38
rs1041184356 Chr.22: 37057142-37057142 on Build GRCh38
rs574717152 Chr.22: 37057127-37057127 on Build GRCh38
rs1011532654 Chr.22: 37057206-37057206 on Build GRCh38
rs537641981 Chr.22: 37057487-37057487 on Build GRCh38
rs770258767 Chr.22: 37057516-37057516 on Build GRCh38
rs770530808 Chr.22: 37057390-37057390 on Build GRCh38
rs372989339 Chr.22: 37057513-37057513 on Build GRCh38
rs947420043 Chr.22: 37057422-37057422 on Build GRCh38
rs140838957 Chr.22: 37057482-37057482 on Build GRCh38
rs746274419 Chr.22: 37057493-37057493 on Build GRCh38
rs369557121 Chr.22: 37057504-37057504 on Build GRCh38
rs750621399 Chr.22: 37057451-37057451 on Build GRCh38
rs529549036 Chr.22: 37057313-37057313 on Build GRCh38
rs79695422 Chr.22: 37057316-37057316 on Build GRCh38
rs946867418 Chr.22: 37057493-37057493 on Build GRCh38
rs762190959 Chr.22: 37057172-37057172 on Build GRCh38
rs775595228 Chr.22: 37057518-37057518 on Build GRCh38
rs895556716 Chr.22: 37057270-37057270 on Build GRCh38
rs1056698619 Chr.22: 37057521-37057521 on Build GRCh38
rs777747471 Chr.22: 37057507-37057507 on Build GRCh38
rs952632092 Chr.22: 37057095-37057095 on Build GRCh38
rs531917277 Chr.22: 37057248-37057248 on Build GRCh38
rs146668214 Chr.22: 37057448-37057448 on Build GRCh38
rs529895350 Chr.22: 37057311-37057311 on Build GRCh38
rs866414918 Chr.22: 37057440-37057440 on Build GRCh38
rs762198990 Chr.22: 37057442-37057442 on Build GRCh38
rs1043052727 Chr.22: 37057439-37057439 on Build GRCh38
rs778304882 Chr.22: 37057419-37057419 on Build GRCh38
rs768710127 Chr.22: 37057353-37057353 on Build GRCh38
rs934358610 Chr.22: 37057518-37057518 on Build GRCh38
rs2235320 Chr.22: 37057216-37057216 on Build GRCh38
rs756139290 Chr.22: 37057334-37057334 on Build GRCh38
rs150660811 Chr.22: 37057136-37057136 on Build GRCh38
rs932826944 Chr.22: 37057162-37057162 on Build GRCh38
rs759017155 Chr.22: 37057523-37057523 on Build GRCh38
rs752012072 Chr.22: 37057421-37057421 on Build GRCh38
rs181921639 Chr.22: 37057367-37057367 on Build GRCh38
rs710184 Chr.22: 37057219-37057219 on Build GRCh38
rs1012717581 Chr.22: 37057312-37057312 on Build GRCh38
rs746652573 Chr.22: 37057509-37057509 on Build GRCh38
rs368089936 Chr.22: 37057472-37057472 on Build GRCh38
rs770763960 Chr.22: 37057511-37057511 on Build GRCh38
rs201383375 Chr.22: 37057498-37057498 on Build GRCh38
rs973713580 Chr.22: 37057303-37057303 on Build GRCh38
rs1049907806 Chr.22: 37057173-37057173 on Build GRCh38
rs374873176 Chr.22: 37057497-37057497 on Build GRCh38
rs912354728 Chr.22: 37057121-37057121 on Build GRCh38
rs786205860 Chr.22: 37057420-37057420 on Build GRCh38
rs767117645 Chr.22: 37057506-37057506 on Build GRCh38
rs572373930 Chr.22: 37057146-37057146 on Build GRCh38
rs755029917 Chr.22: 37057488-37057488 on Build GRCh38
rs988785744 Chr.22: 37057368-37057368 on Build GRCh38
rs771791144 Chr.22: 37057359-37057359 on Build GRCh38
rs946951989 Chr.22: 37057201-37057201 on Build GRCh38
rs777171270 Chr.22: 37057088-37057088 on Build GRCh38
rs747922184 Chr.22: 37057356-37057356 on Build GRCh38
rs774005791 Chr.22: 37057354-37057354 on Build GRCh38
rs189290757 Chr.22: 37057096-37057096 on Build GRCh38
rs1043146417 Chr.22: 37057207-37057207 on Build GRCh38
rs541768733 Chr.22: 37057131-37057131 on Build GRCh38
rs985643783 Chr.22: 37057430-37057430 on Build GRCh38
rs960444400 Chr.22: 37057250-37057250 on Build GRCh38
rs17852879 Chr.22: 37057463-37057463 on Build GRCh38
rs895589404 Chr.22: 37057522-37057522 on Build GRCh38
rs544074937 Chr.22: 37057264-37057264 on Build GRCh38
rs894060958 Chr.22: 37057184-37057184 on Build GRCh38
rs976728587 Chr.22: 37057411-37057411 on Build GRCh38

View Full Product Details