Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 37062319-37062820 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCTD17
Entrez Gene ID
79734
Gene Name
potassium channel tetramerization domain containing 17
Gene Aliases
-
UniGene
Hs.517597
Gene Chromosome Location
Chr.22: 37051725-37063390 on Build GRCh38
Transcripts
NM_001282684.1 NM_001282685.1 NM_001282686.1 NM_024681.3 XM_005261741.3 XM_005261742.2 XM_005261743.2 XM_005261744.2 XM_011530374.2 XM_011530377.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7318435 c.965G>C p.*322S Chr.22: 37062593-37062593 on Build GRCh38
7373299 c.937C>G p.H313D Chr.22: 37062565-37062565 on Build GRCh38
4103880 c.917C>G p.P306R Chr.22: 37062545-37062545 on Build GRCh38
7318436 c.872G>C p.*291S Chr.22: 37062593-37062593 on Build GRCh38
4103879 c.824C>G p.P275R Chr.22: 37062545-37062545 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768369293 Chr.22: 37062599-37062599 on Build GRCh38
rs764412806 Chr.22: 37062632-37062632 on Build GRCh38
rs747042008 Chr.22: 37062483-37062483 on Build GRCh38
rs751146810 Chr.22: 37062512-37062513 on Build GRCh38
rs763745590 Chr.22: 37062522-37062522 on Build GRCh38
rs776500999 Chr.22: 37062485-37062485 on Build GRCh38
rs772927623 Chr.22: 37062615-37062615 on Build GRCh38
rs539318802 Chr.22: 37062711-37062711 on Build GRCh38
rs568799442 Chr.22: 37062731-37062731 on Build GRCh38
rs991870958 Chr.22: 37062658-37062658 on Build GRCh38
rs765502882 Chr.22: 37062539-37062539 on Build GRCh38
rs766521424 Chr.22: 37062462-37062462 on Build GRCh38
rs776133872 Chr.22: 37062399-37062400 on Build GRCh38
rs556474244 Chr.22: 37062440-37062440 on Build GRCh38
rs369009490 Chr.22: 37062512-37062512 on Build GRCh38
rs371727118 Chr.22: 37062476-37062476 on Build GRCh38
rs544187968 Chr.22: 37062425-37062425 on Build GRCh38
rs1000002552 Chr.22: 37062421-37062421 on Build GRCh38
rs754259514 Chr.22: 37062510-37062511 on Build GRCh38
rs577107502 Chr.22: 37062362-37062362 on Build GRCh38
rs146711968 Chr.22: 37062565-37062565 on Build GRCh38
rs758030293 Chr.22: 37062471-37062471 on Build GRCh38
rs188337530 Chr.22: 37062628-37062628 on Build GRCh38
rs944023896 Chr.22: 37062747-37062747 on Build GRCh38
rs1041688320 Chr.22: 37062509-37062509 on Build GRCh38
rs769546982 Chr.22: 37062510-37062510 on Build GRCh38
rs376293639 Chr.22: 37062640-37062640 on Build GRCh38
rs763640868 Chr.22: 37062513-37062513 on Build GRCh38
rs933012971 Chr.22: 37062717-37062717 on Build GRCh38
rs940223853 Chr.22: 37062487-37062487 on Build GRCh38
rs778013908 Chr.22: 37062544-37062544 on Build GRCh38
rs564681008 Chr.22: 37062655-37062655 on Build GRCh38
rs745801740 Chr.22: 37062550-37062550 on Build GRCh38
rs377623859 Chr.22: 37062792-37062792 on Build GRCh38
rs1053123969 Chr.22: 37062590-37062590 on Build GRCh38
rs967769017 Chr.22: 37062393-37062393 on Build GRCh38
rs577281266 Chr.22: 37062727-37062727 on Build GRCh38
rs372569746 Chr.22: 37062633-37062633 on Build GRCh38
rs775421289 Chr.22: 37062518-37062518 on Build GRCh38
rs751748057 Chr.22: 37062644-37062644 on Build GRCh38
rs754379400 Chr.22: 37062535-37062535 on Build GRCh38
rs888866375 Chr.22: 37062729-37062729 on Build GRCh38
rs908746612 Chr.22: 37062702-37062702 on Build GRCh38
rs755585851 Chr.22: 37062538-37062538 on Build GRCh38
rs1015331233 Chr.22: 37062677-37062677 on Build GRCh38
rs771681455 Chr.22: 37062613-37062613 on Build GRCh38
rs773882736 Chr.22: 37062525-37062525 on Build GRCh38
rs960354987 Chr.22: 37062659-37062659 on Build GRCh38
rs745853104 Chr.22: 37062490-37062490 on Build GRCh38
rs984299291 Chr.22: 37062695-37062695 on Build GRCh38
rs1053160727 Chr.22: 37062769-37062769 on Build GRCh38
rs1041320916 Chr.22: 37062712-37062712 on Build GRCh38
rs916017721 Chr.22: 37062676-37062676 on Build GRCh38
rs758691104 Chr.22: 37062543-37062543 on Build GRCh38
rs779870701 Chr.22: 37062578-37062578 on Build GRCh38
rs749046876 Chr.22: 37062581-37062581 on Build GRCh38
rs370950840 Chr.22: 37062631-37062631 on Build GRCh38
rs765705394 Chr.22: 37062627-37062627 on Build GRCh38
rs143403650 Chr.22: 37062547-37062547 on Build GRCh38
rs991876631 Chr.22: 37062458-37062458 on Build GRCh38
rs367800128 Chr.22: 37062409-37062410 on Build GRCh38
rs199931970 Chr.22: 37062604-37062604 on Build GRCh38
rs903048687 Chr.22: 37062779-37062779 on Build GRCh38
rs116956945 Chr.22: 37062546-37062546 on Build GRCh38
rs771027813 Chr.22: 37062484-37062484 on Build GRCh38
rs967275518 Chr.22: 37062603-37062603 on Build GRCh38
rs749261230 Chr.22: 37062398-37062399 on Build GRCh38
rs142739650 Chr.22: 37062531-37062531 on Build GRCh38
rs146513411 Chr.22: 37062532-37062532 on Build GRCh38
rs550231152 Chr.22: 37062721-37062721 on Build GRCh38
rs1035626764 Chr.22: 37062786-37062786 on Build GRCh38
rs974323996 Chr.22: 37062679-37062679 on Build GRCh38
rs756795934 Chr.22: 37062513-37062514 on Build GRCh38
rs952735383 Chr.22: 37062678-37062678 on Build GRCh38
rs377601872 Chr.22: 37062541-37062541 on Build GRCh38
rs760270622 Chr.22: 37062623-37062623 on Build GRCh38
rs762500530 Chr.22: 37062519-37062519 on Build GRCh38
rs999082076 Chr.22: 37062783-37062783 on Build GRCh38
rs374182123 Chr.22: 37062716-37062716 on Build GRCh38
rs79883813 Chr.22: 37062759-37062759 on Build GRCh38

View Full Product Details